Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 223357-223854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THOC1
Entrez Gene ID
9984
Gene Name
THO complex 1
Gene Aliases
HPR1, P84, P84N5
UniGene
Hs.712543
Gene Chromosome Location
Chr.18: 214348-268059 on Build GRCh38
Transcripts
NM_005131.2 XM_011525772.2 XM_011525773.1 XM_011525774.2 XM_017026103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
86087 c.1363G>C p.E455Q Chr.18: 223447-223447 on Build GRCh38
2807946 c.1343A>T p.N448I Chr.18: 223467-223467 on Build GRCh38
6267402 c.1367C>A p.T456K Chr.18: 223443-223443 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974517139 Chr.18: 223627-223627 on Build GRCh38
rs768489021 Chr.18: 223392-223407 on Build GRCh38
rs886679282 Chr.18: 223414-223414 on Build GRCh38
rs188154103 Chr.18: 223514-223514 on Build GRCh38
rs770852578 Chr.18: 223554-223554 on Build GRCh38
rs1016382431 Chr.18: 223513-223513 on Build GRCh38
rs749138700 Chr.18: 223426-223426 on Build GRCh38
rs543809473 Chr.18: 223619-223619 on Build GRCh38
rs780943675 Chr.18: 223564-223564 on Build GRCh38
rs892770979 Chr.18: 223519-223519 on Build GRCh38
rs1006790717 Chr.18: 223407-223407 on Build GRCh38
rs1039637207 Chr.18: 223787-223787 on Build GRCh38
rs779630943 Chr.18: 223399-223399 on Build GRCh38
rs182019217 Chr.18: 223734-223734 on Build GRCh38
rs768735586 Chr.18: 223490-223490 on Build GRCh38
rs1047712112 Chr.18: 223706-223706 on Build GRCh38
rs186300534 Chr.18: 223809-223809 on Build GRCh38
rs545984692 Chr.18: 223543-223543 on Build GRCh38
rs1011695316 Chr.18: 223541-223541 on Build GRCh38
rs371378283 Chr.18: 223573-223573 on Build GRCh38
rs533519577 Chr.18: 223813-223813 on Build GRCh38
rs576376617 Chr.18: 223582-223582 on Build GRCh38
rs369768173 Chr.18: 223569-223569 on Build GRCh38
rs747921385 Chr.18: 223528-223528 on Build GRCh38
rs938796336 Chr.18: 223705-223710 on Build GRCh38
rs1026296801 Chr.18: 223385-223389 on Build GRCh38
rs375519048 Chr.18: 223546-223546 on Build GRCh38
rs369274638 Chr.18: 223395-223395 on Build GRCh38
rs941362901 Chr.18: 223612-223612 on Build GRCh38
rs745403057 Chr.18: 223757-223757 on Build GRCh38
rs201902953 Chr.18: 223545-223545 on Build GRCh38
rs755568045 Chr.18: 223396-223396 on Build GRCh38
rs529597055 Chr.18: 223655-223655 on Build GRCh38
rs1006387455 Chr.18: 223737-223737 on Build GRCh38
rs562393985 Chr.18: 223653-223653 on Build GRCh38
rs1051192915 Chr.18: 223675-223675 on Build GRCh38
rs76216934 Chr.18: 223509-223509 on Build GRCh38
rs866623318 Chr.18: 223462-223462 on Build GRCh38
rs974482356 Chr.18: 223542-223542 on Build GRCh38
rs907262161 Chr.18: 223696-223696 on Build GRCh38
rs996768992 Chr.18: 223825-223834 on Build GRCh38
rs916377832 Chr.18: 223642-223642 on Build GRCh38
rs990991906 Chr.18: 223578-223578 on Build GRCh38
rs978219161 Chr.18: 223552-223552 on Build GRCh38
rs942520411 Chr.18: 223762-223762 on Build GRCh38

View Full Product Details