Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 332925-333427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC12
Entrez Gene ID
81035
Gene Name
collectin subfamily member 12
Gene Aliases
CLP1, NSR2, SCARA4, SRCL
UniGene
Hs.464422
Gene Chromosome Location
Chr.18: 319355-500729 on Build GRCh38
Transcripts
NM_130386.2 XM_011525741.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7120972 c.1953+1G>A p.? Chr.18: 333006-333006 on Build GRCh38
707924 c.1870G>C p.E624Q Chr.18: 333090-333090 on Build GRCh38
4072019 c.1840T>G p.F614V Chr.18: 333120-333120 on Build GRCh38
988174 c.1876G>T p.E626* Chr.18: 333084-333084 on Build GRCh38
707925 c.1924C>G p.L642V Chr.18: 333036-333036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754546205 Chr.18: 333082-333082 on Build GRCh38
rs917724224 Chr.18: 333274-333274 on Build GRCh38
rs570806163 Chr.18: 333288-333288 on Build GRCh38
rs553068043 Chr.18: 333287-333287 on Build GRCh38
rs1054287137 Chr.18: 333379-333381 on Build GRCh38
rs1005021323 Chr.18: 333040-333040 on Build GRCh38
rs776090490 Chr.18: 333159-333159 on Build GRCh38
rs147524076 Chr.18: 333124-333124 on Build GRCh38
rs746832000 Chr.18: 333132-333132 on Build GRCh38
rs555107505 Chr.18: 333398-333398 on Build GRCh38
rs979825988 Chr.18: 333230-333230 on Build GRCh38
rs764547445 Chr.18: 333185-333185 on Build GRCh38
rs144753454 Chr.18: 333397-333397 on Build GRCh38
rs951374268 Chr.18: 333237-333237 on Build GRCh38
rs149346230 Chr.18: 333336-333336 on Build GRCh38
rs765835744 Chr.18: 332991-332991 on Build GRCh38
rs1045139380 Chr.18: 332984-332984 on Build GRCh38
rs893076488 Chr.18: 333384-333384 on Build GRCh38
rs568120897 Chr.18: 333356-333356 on Build GRCh38
rs927917223 Chr.18: 333231-333231 on Build GRCh38
rs377320854 Chr.18: 333021-333021 on Build GRCh38
rs1019030990 Chr.18: 333084-333084 on Build GRCh38
rs991997974 Chr.18: 333265-333265 on Build GRCh38
rs1025085935 Chr.18: 333332-333332 on Build GRCh38
rs933699187 Chr.18: 333297-333297 on Build GRCh38
rs147318026 Chr.18: 333009-333009 on Build GRCh38
rs947959103 Chr.18: 332962-332962 on Build GRCh38
rs773032503 Chr.18: 333130-333130 on Build GRCh38
rs371151646 Chr.18: 332987-332987 on Build GRCh38
rs771173973 Chr.18: 333136-333136 on Build GRCh38
rs537826782 Chr.18: 333300-333300 on Build GRCh38
rs758777869 Chr.18: 332998-332998 on Build GRCh38
rs759992691 Chr.18: 333238-333238 on Build GRCh38
rs982220917 Chr.18: 333220-333220 on Build GRCh38
rs139504960 Chr.18: 333137-333137 on Build GRCh38
rs187492425 Chr.18: 332988-332988 on Build GRCh38
rs527541745 Chr.18: 333255-333255 on Build GRCh38
rs199908317 Chr.18: 333049-333049 on Build GRCh38
rs753261571 Chr.18: 332996-332996 on Build GRCh38
rs2305028 Chr.18: 332961-332961 on Build GRCh38
rs756048246 Chr.18: 333281-333281 on Build GRCh38
rs548335086 Chr.18: 333158-333158 on Build GRCh38
rs778750998 Chr.18: 333102-333102 on Build GRCh38
rs898389036 Chr.18: 333331-333331 on Build GRCh38
rs138736525 Chr.18: 333115-333115 on Build GRCh38
rs910161258 Chr.18: 333266-333266 on Build GRCh38
rs750002429 Chr.18: 333019-333019 on Build GRCh38
rs752094410 Chr.18: 332953-332953 on Build GRCh38
rs374940948 Chr.18: 333168-333168 on Build GRCh38
rs753375151 Chr.18: 333298-333298 on Build GRCh38
rs968098810 Chr.18: 333176-333176 on Build GRCh38
rs1046686095 Chr.18: 333395-333395 on Build GRCh38
rs902832645 Chr.18: 333318-333318 on Build GRCh38
rs921443072 Chr.18: 333293-333293 on Build GRCh38
rs762231639 Chr.18: 332965-332969 on Build GRCh38
rs539509715 Chr.18: 333241-333241 on Build GRCh38
rs748779809 Chr.18: 333066-333066 on Build GRCh38
rs370191806 Chr.18: 333151-333151 on Build GRCh38

View Full Product Details