Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13730391-13730926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNMT
Entrez Gene ID
8731
Gene Name
RNA guanine-7 methyltransferase
Gene Aliases
CMT1, CMT1c, MET, Met, RG7MT1, cm1p, hCMT1, hMet
UniGene
Hs.592347
Gene Chromosome Location
Chr.18: 13726647-13764556 on Build GRCh38
Transcripts
NM_001308263.1 NM_003799.2 XM_005258162.1 XM_011525749.1 XM_011525750.2 XM_011525751.1 XM_011525753.1 XM_017026061.1

SNP Detail

SNP ID Chromosome Location
rs939096168 Chr.18: 13730615-13730615 on Build GRCh38
rs147024295 Chr.18: 13730810-13730810 on Build GRCh38
rs974175048 Chr.18: 13730536-13730536 on Build GRCh38
rs185155962 Chr.18: 13730876-13730876 on Build GRCh38
rs1033290506 Chr.18: 13730574-13730574 on Build GRCh38
rs558011891 Chr.18: 13730879-13730879 on Build GRCh38
rs1005139637 Chr.18: 13730473-13730473 on Build GRCh38
rs554707644 Chr.18: 13730473-13730474 on Build GRCh38
rs573299555 Chr.18: 13730894-13730894 on Build GRCh38
rs959134195 Chr.18: 13730575-13730575 on Build GRCh38
rs1007584593 Chr.18: 13730686-13730686 on Build GRCh38
rs551214665 Chr.18: 13730622-13730622 on Build GRCh38
rs141541775 Chr.18: 13730667-13730667 on Build GRCh38
rs755679917 Chr.18: 13730845-13730845 on Build GRCh38
rs750250862 Chr.18: 13730831-13730831 on Build GRCh38
rs562359562 Chr.18: 13730423-13730423 on Build GRCh38
rs1016420971 Chr.18: 13730846-13730846 on Build GRCh38
rs757265536 Chr.18: 13730655-13730655 on Build GRCh38
rs962424370 Chr.18: 13730446-13730446 on Build GRCh38
rs909420969 Chr.18: 13730424-13730424 on Build GRCh38
rs527337442 Chr.18: 13730637-13730637 on Build GRCh38
rs899435400 Chr.18: 13730847-13730847 on Build GRCh38
rs377545417 Chr.18: 13730857-13730857 on Build GRCh38
rs751612723 Chr.18: 13730511-13730511 on Build GRCh38
rs781072226 Chr.18: 13730775-13730775 on Build GRCh38
rs886523079 Chr.18: 13730706-13730706 on Build GRCh38
rs1051811699 Chr.18: 13730687-13730687 on Build GRCh38
rs572954369 Chr.18: 13730626-13730626 on Build GRCh38
rs900473282 Chr.18: 13730675-13730675 on Build GRCh38
rs1005498571 Chr.18: 13730754-13730754 on Build GRCh38
rs568721765 Chr.18: 13730666-13730666 on Build GRCh38
rs950921084 Chr.18: 13730791-13730791 on Build GRCh38
rs145726254 Chr.18: 13730643-13730643 on Build GRCh38
rs991835437 Chr.18: 13730614-13730614 on Build GRCh38
rs926403741 Chr.18: 13730602-13730602 on Build GRCh38
rs566459063 Chr.18: 13730632-13730632 on Build GRCh38
rs764108933 Chr.18: 13730422-13730422 on Build GRCh38
rs369070908 Chr.18: 13730479-13730479 on Build GRCh38
rs1039041723 Chr.18: 13730745-13730745 on Build GRCh38
rs1016538938 Chr.18: 13730861-13730861 on Build GRCh38
rs149792398 Chr.18: 13730526-13730526 on Build GRCh38
rs1043659593 Chr.18: 13730649-13730649 on Build GRCh38
rs557552742 Chr.18: 13730743-13730743 on Build GRCh38
rs773117249 Chr.18: 13730871-13730871 on Build GRCh38
rs886573958 Chr.18: 13730468-13730468 on Build GRCh38
rs1026363429 Chr.18: 13730788-13730788 on Build GRCh38
rs11549670 Chr.18: 13730718-13730718 on Build GRCh38
rs901963406 Chr.18: 13730763-13730763 on Build GRCh38
rs1016969008 Chr.18: 13730503-13730503 on Build GRCh38

View Full Product Details