Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14076916-14077426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF519
Entrez Gene ID
162655
Gene Name
zinc finger protein 519
Gene Aliases
HsT2362
UniGene
Hs.352635
Gene Chromosome Location
Chr.18: 14071230-14132494 on Build GRCh38
Transcripts
NR_033354.1 XM_017025562.1 XM_017025563.1 XM_017025564.1 XM_017025565.1

SNP Detail

SNP ID Chromosome Location
rs962977666 Chr.18: 14077260-14077260 on Build GRCh38
rs968358682 Chr.18: 14077318-14077318 on Build GRCh38
rs765629983 Chr.18: 14077298-14077298 on Build GRCh38
rs904411285 Chr.18: 14077050-14077050 on Build GRCh38
rs771471478 Chr.18: 14077010-14077010 on Build GRCh38
rs1047733047 Chr.18: 14077036-14077036 on Build GRCh38
rs16941550 Chr.18: 14077218-14077218 on Build GRCh38
rs1040476583 Chr.18: 14076992-14076992 on Build GRCh38
rs777104718 Chr.18: 14077048-14077048 on Build GRCh38
rs769699950 Chr.18: 14077283-14077283 on Build GRCh38
rs964515519 Chr.18: 14077148-14077148 on Build GRCh38
rs181215101 Chr.18: 14077142-14077142 on Build GRCh38
rs901936547 Chr.18: 14076993-14076993 on Build GRCh38
rs982721127 Chr.18: 14077235-14077235 on Build GRCh38
rs9965456 Chr.18: 14076991-14076991 on Build GRCh38
rs61740406 Chr.18: 14077335-14077335 on Build GRCh38
rs1040190054 Chr.18: 14077381-14077381 on Build GRCh38
rs1048172484 Chr.18: 14077387-14077388 on Build GRCh38
rs9945224 Chr.18: 14077382-14077382 on Build GRCh38
rs1000466567 Chr.18: 14077081-14077081 on Build GRCh38
rs554883715 Chr.18: 14076994-14076994 on Build GRCh38
rs988189090 Chr.18: 14077267-14077267 on Build GRCh38
rs577636067 Chr.18: 14077276-14077276 on Build GRCh38
rs35776417 Chr.18: 14077111-14077112 on Build GRCh38
rs61730227 Chr.18: 14077321-14077321 on Build GRCh38
rs939484365 Chr.18: 14077330-14077330 on Build GRCh38
rs764109821 Chr.18: 14077350-14077350 on Build GRCh38
rs1020232899 Chr.18: 14077047-14077047 on Build GRCh38
rs9954547 Chr.18: 14076973-14076973 on Build GRCh38
rs943169447 Chr.18: 14077338-14077338 on Build GRCh38
rs775326802 Chr.18: 14077344-14077344 on Build GRCh38
rs1034733821 Chr.18: 14077246-14077246 on Build GRCh38
rs543675167 Chr.18: 14077093-14077093 on Build GRCh38
rs142539912 Chr.18: 14077191-14077191 on Build GRCh38
rs960502180 Chr.18: 14077253-14077253 on Build GRCh38
rs576275035 Chr.18: 14077059-14077059 on Build GRCh38
rs377354286 Chr.18: 14077395-14077396 on Build GRCh38
rs144800282 Chr.18: 14077239-14077239 on Build GRCh38
rs1043934161 Chr.18: 14076956-14076956 on Build GRCh38
rs976855449 Chr.18: 14077159-14077159 on Build GRCh38
rs905635378 Chr.18: 14076955-14076955 on Build GRCh38
rs937086139 Chr.18: 14076963-14076963 on Build GRCh38

View Full Product Details