Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14077265-14077806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF519
Entrez Gene ID
162655
Gene Name
zinc finger protein 519
Gene Aliases
HsT2362
UniGene
Hs.352635
Gene Chromosome Location
Chr.18: 14071230-14132494 on Build GRCh38
Transcripts
NR_033354.1 XM_017025562.1 XM_017025563.1 XM_017025564.1 XM_017025565.1

SNP Detail

SNP ID Chromosome Location
rs141860622 Chr.18: 14077624-14077624 on Build GRCh38
rs201465954 Chr.18: 14077484-14077484 on Build GRCh38
rs968358682 Chr.18: 14077318-14077318 on Build GRCh38
rs765629983 Chr.18: 14077298-14077298 on Build GRCh38
rs917033057 Chr.18: 14077655-14077655 on Build GRCh38
rs558425312 Chr.18: 14077708-14077708 on Build GRCh38
rs1057308502 Chr.18: 14077775-14077775 on Build GRCh38
rs945761493 Chr.18: 14077675-14077675 on Build GRCh38
rs960925073 Chr.18: 14077629-14077629 on Build GRCh38
rs9945409 Chr.18: 14077505-14077505 on Build GRCh38
rs780198687 Chr.18: 14077573-14077573 on Build GRCh38
rs1041444350 Chr.18: 14077712-14077712 on Build GRCh38
rs571912346 Chr.18: 14077500-14077500 on Build GRCh38
rs554688036 Chr.18: 14077607-14077607 on Build GRCh38
rs150231488 Chr.18: 14077747-14077748 on Build GRCh38
rs762740575 Chr.18: 14077491-14077491 on Build GRCh38
rs61740406 Chr.18: 14077335-14077335 on Build GRCh38
rs906116583 Chr.18: 14077445-14077445 on Build GRCh38
rs1040190054 Chr.18: 14077381-14077381 on Build GRCh38
rs1003013378 Chr.18: 14077455-14077455 on Build GRCh38
rs1048172484 Chr.18: 14077387-14077388 on Build GRCh38
rs1044706707 Chr.18: 14077437-14077438 on Build GRCh38
rs9945224 Chr.18: 14077382-14077382 on Build GRCh38
rs1041391839 Chr.18: 14077421-14077421 on Build GRCh38
rs9945512 Chr.18: 14077640-14077640 on Build GRCh38
rs963326934 Chr.18: 14077599-14077599 on Build GRCh38
rs1029719691 Chr.18: 14077571-14077571 on Build GRCh38
rs536718008 Chr.18: 14077585-14077585 on Build GRCh38
rs61730227 Chr.18: 14077321-14077321 on Build GRCh38
rs939484365 Chr.18: 14077330-14077330 on Build GRCh38
rs576654643 Chr.18: 14077734-14077734 on Build GRCh38
rs1000115420 Chr.18: 14077479-14077479 on Build GRCh38
rs764109821 Chr.18: 14077350-14077350 on Build GRCh38
rs943169447 Chr.18: 14077338-14077338 on Build GRCh38
rs775326802 Chr.18: 14077344-14077344 on Build GRCh38
rs1035878567 Chr.18: 14077470-14077470 on Build GRCh38
rs942001270 Chr.18: 14077692-14077692 on Build GRCh38
rs9945271 Chr.18: 14077578-14077578 on Build GRCh38
rs377354286 Chr.18: 14077395-14077396 on Build GRCh38
rs138741134 Chr.18: 14077478-14077478 on Build GRCh38
rs546564502 Chr.18: 14077502-14077502 on Build GRCh38
rs981056854 Chr.18: 14077620-14077620 on Build GRCh38
rs909203222 Chr.18: 14077681-14077681 on Build GRCh38

View Full Product Details