Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23237059-23237564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABLES1
Entrez Gene ID
91768
Gene Name
Cdk5 and Abl enzyme substrate 1
Gene Aliases
CABL1, CABLES, HsT2563, IK3-1
UniGene
Hs.11108
Gene Chromosome Location
Chr.18: 23134564-23260470 on Build GRCh38
Transcripts
NM_001100619.2 NM_001256438.1 NM_138375.2 NR_023359.1 XM_017026043.1 XM_017026046.1 XM_017026047.1 XM_017026048.1

Gene Symbol
TMEM241
Entrez Gene ID
85019
Gene Name
transmembrane protein 241
Gene Aliases
C18orf45, hVVT
UniGene
Hs.137562
Gene Chromosome Location
Chr.18: 23237252-23437969 on Build GRCh38
Transcripts
NM_001100619.2 NM_001256438.1 NM_138375.2 NR_023359.1 XM_017026043.1 XM_017026046.1 XM_017026047.1 XM_017026048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853975 c.651G>A p.M217I Chr.18: 23237245-23237245 on Build GRCh38
3821236 c.1433T>C p.F478S Chr.18: 23237232-23237232 on Build GRCh38
987036 c.1385T>C p.L462S Chr.18: 23237184-23237184 on Build GRCh38
3821237 c.638T>C p.F213S Chr.18: 23237232-23237232 on Build GRCh38
4525810 c.560G>T p.G187V Chr.18: 23237154-23237154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202142583 Chr.18: 23237149-23237149 on Build GRCh38
rs771688713 Chr.18: 23237165-23237165 on Build GRCh38
rs764346578 Chr.18: 23237130-23237130 on Build GRCh38
rs1026021052 Chr.18: 23237531-23237531 on Build GRCh38
rs779939432 Chr.18: 23237142-23237142 on Build GRCh38
rs761091853 Chr.18: 23237104-23237104 on Build GRCh38
rs973007011 Chr.18: 23237521-23237521 on Build GRCh38
rs374844780 Chr.18: 23237113-23237113 on Build GRCh38
rs575119100 Chr.18: 23237340-23237340 on Build GRCh38
rs891845365 Chr.18: 23237387-23237387 on Build GRCh38
rs370033761 Chr.18: 23237282-23237282 on Build GRCh38
rs1041246856 Chr.18: 23237382-23237382 on Build GRCh38
rs1051898784 Chr.18: 23237320-23237320 on Build GRCh38
rs956544272 Chr.18: 23237176-23237176 on Build GRCh38
rs930220268 Chr.18: 23237422-23237422 on Build GRCh38
rs777133412 Chr.18: 23237112-23237112 on Build GRCh38
rs45541339 Chr.18: 23237099-23237099 on Build GRCh38
rs751823917 Chr.18: 23237243-23237243 on Build GRCh38
rs775472800 Chr.18: 23237221-23237221 on Build GRCh38
rs912167547 Chr.18: 23237333-23237333 on Build GRCh38
rs771287723 Chr.18: 23237203-23237203 on Build GRCh38
rs77591546 Chr.18: 23237337-23237337 on Build GRCh38
rs763314753 Chr.18: 23237297-23237297 on Build GRCh38
rs760419867 Chr.18: 23237215-23237215 on Build GRCh38
rs12185467 Chr.18: 23237312-23237312 on Build GRCh38
rs747777804 Chr.18: 23237097-23237097 on Build GRCh38
rs901404550 Chr.18: 23237389-23237389 on Build GRCh38
rs559246969 Chr.18: 23237415-23237415 on Build GRCh38
rs1009374342 Chr.18: 23237185-23237185 on Build GRCh38
rs753660686 Chr.18: 23237270-23237270 on Build GRCh38
rs770265368 Chr.18: 23237220-23237220 on Build GRCh38
rs1033533911 Chr.18: 23237515-23237516 on Build GRCh38
rs376624705 Chr.18: 23237283-23237283 on Build GRCh38
rs572528816 Chr.18: 23237294-23237294 on Build GRCh38
rs2289013 Chr.18: 23237309-23237309 on Build GRCh38
rs770353353 Chr.18: 23237290-23237290 on Build GRCh38
rs142158551 Chr.18: 23237158-23237158 on Build GRCh38
rs778065523 Chr.18: 23237140-23237141 on Build GRCh38
rs756389414 Chr.18: 23237113-23237114 on Build GRCh38
rs752776242 Chr.18: 23237119-23237119 on Build GRCh38
rs777467325 Chr.18: 23237184-23237184 on Build GRCh38
rs1003767340 Chr.18: 23237543-23237543 on Build GRCh38
rs1005513707 Chr.18: 23237464-23237464 on Build GRCh38
rs746583023 Chr.18: 23237198-23237198 on Build GRCh38
rs766297106 Chr.18: 23237255-23237255 on Build GRCh38
rs905314053 Chr.18: 23237534-23237534 on Build GRCh38
rs776652851 Chr.18: 23237541-23237541 on Build GRCh38
rs781586497 Chr.18: 23237284-23237284 on Build GRCh38
rs971080498 Chr.18: 23237467-23237467 on Build GRCh38
rs918975685 Chr.18: 23237311-23237311 on Build GRCh38
rs199957517 Chr.18: 23237098-23237098 on Build GRCh38
rs1000176707 Chr.18: 23237369-23237369 on Build GRCh38
rs757939131 Chr.18: 23237278-23237278 on Build GRCh38
rs201780883 Chr.18: 23237082-23237082 on Build GRCh38
rs752207209 Chr.18: 23237273-23237273 on Build GRCh38
rs888415630 Chr.18: 23237451-23237451 on Build GRCh38
rs766926220 Chr.18: 23237105-23237105 on Build GRCh38
rs904644181 Chr.18: 23237140-23237147 on Build GRCh38
rs368459056 Chr.18: 23237150-23237150 on Build GRCh38
rs773957845 Chr.18: 23237092-23237092 on Build GRCh38
rs763088478 Chr.18: 23237222-23237222 on Build GRCh38
rs963026087 Chr.18: 23237515-23237515 on Build GRCh38
rs1002058119 Chr.18: 23237488-23237488 on Build GRCh38
rs45518733 Chr.18: 23237242-23237242 on Build GRCh38
rs1039084918 Chr.18: 23237345-23237345 on Build GRCh38
rs747659049 Chr.18: 23237163-23237163 on Build GRCh38
rs753458944 Chr.18: 23237296-23237296 on Build GRCh38
rs765334150 Chr.18: 23237115-23237115 on Build GRCh38
rs982000139 Chr.18: 23237279-23237279 on Build GRCh38
rs1047778232 Chr.18: 23237448-23237448 on Build GRCh38
rs1023673648 Chr.18: 23237520-23237520 on Build GRCh38
rs368694092 Chr.18: 23237286-23237286 on Build GRCh38
rs778611451 Chr.18: 23237152-23237152 on Build GRCh38
rs529155042 Chr.18: 23237368-23237368 on Build GRCh38
rs763138481 Chr.18: 23237129-23237129 on Build GRCh38
rs950492074 Chr.18: 23237545-23237545 on Build GRCh38
rs1009818386 Chr.18: 23237397-23237397 on Build GRCh38
rs762063341 Chr.18: 23237254-23237254 on Build GRCh38
rs776943205 Chr.18: 23237204-23237204 on Build GRCh38
rs766692279 Chr.18: 23237466-23237466 on Build GRCh38
rs760059139 Chr.18: 23237219-23237219 on Build GRCh38
rs544070219 Chr.18: 23237367-23237367 on Build GRCh38
rs952733283 Chr.18: 23237139-23237139 on Build GRCh38
rs201342490 Chr.18: 23237271-23237271 on Build GRCh38
rs750255561 Chr.18: 23237133-23237133 on Build GRCh38

View Full Product Details