Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35991840-35992387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPRD1A
Entrez Gene ID
55197
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Gene Aliases
HsT3101, P15RS
UniGene
Hs.464912
Gene Chromosome Location
Chr.18: 35982385-36067594 on Build GRCh38
Transcripts
NM_001303411.1 NM_001303412.1 NM_018170.4 XM_011526048.1 XM_011526049.1 XM_011526050.1

SNP Detail

SNP ID Chromosome Location
rs923628228 Chr.18: 35992212-35992212 on Build GRCh38
rs146252108 Chr.18: 35992075-35992075 on Build GRCh38
rs184905507 Chr.18: 35992305-35992305 on Build GRCh38
rs879094978 Chr.18: 35992134-35992134 on Build GRCh38
rs991210191 Chr.18: 35991993-35991993 on Build GRCh38
rs369362883 Chr.18: 35992315-35992315 on Build GRCh38
rs572694916 Chr.18: 35991875-35991875 on Build GRCh38
rs560924413 Chr.18: 35992209-35992209 on Build GRCh38
rs141717307 Chr.18: 35991868-35991868 on Build GRCh38
rs762323703 Chr.18: 35991876-35991879 on Build GRCh38
rs779519891 Chr.18: 35992202-35992202 on Build GRCh38
rs190936133 Chr.18: 35992350-35992350 on Build GRCh38
rs758631578 Chr.18: 35992349-35992349 on Build GRCh38
rs1027646505 Chr.18: 35992058-35992058 on Build GRCh38
rs957941124 Chr.18: 35992240-35992240 on Build GRCh38
rs879257028 Chr.18: 35992102-35992102 on Build GRCh38
rs995384087 Chr.18: 35992319-35992319 on Build GRCh38
rs1019374553 Chr.18: 35992107-35992107 on Build GRCh38
rs544815017 Chr.18: 35991958-35991958 on Build GRCh38
rs140068923 Chr.18: 35992249-35992249 on Build GRCh38
rs938619752 Chr.18: 35992312-35992312 on Build GRCh38
rs977826052 Chr.18: 35992210-35992210 on Build GRCh38
rs757803455 Chr.18: 35992105-35992105 on Build GRCh38
rs928736835 Chr.18: 35992316-35992316 on Build GRCh38
rs979336748 Chr.18: 35992232-35992232 on Build GRCh38
rs1015519842 Chr.18: 35991934-35991934 on Build GRCh38
rs897149603 Chr.18: 35991967-35991967 on Build GRCh38
rs569770377 Chr.18: 35992246-35992246 on Build GRCh38
rs916450444 Chr.18: 35992294-35992294 on Build GRCh38
rs934874622 Chr.18: 35992245-35992245 on Build GRCh38
rs182360162 Chr.18: 35992053-35992053 on Build GRCh38
rs959796455 Chr.18: 35991876-35991876 on Build GRCh38
rs1044469147 Chr.18: 35991865-35991865 on Build GRCh38
rs911863151 Chr.18: 35992297-35992297 on Build GRCh38
rs1037059220 Chr.18: 35992359-35992359 on Build GRCh38
rs564591786 Chr.18: 35992040-35992040 on Build GRCh38

View Full Product Details