Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 58671637-58672147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927322
Entrez Gene ID
101927322
Gene Name
uncharacterized LOC101927322
Gene Aliases
-
UniGene
Hs.644875
Gene Chromosome Location
Chr.18: 58670009-58671873 on Build GRCh38
Transcripts
NM_001289967.1 NM_006785.3 NM_173844.2 XM_011525794.1

Gene Symbol
MALT1
Entrez Gene ID
10892
Gene Name
MALT1 paracaspase
Gene Aliases
IMD12, MLT, MLT1, PCASP1
UniGene
Hs.601217
Gene Chromosome Location
Chr.18: 58671386-58753806 on Build GRCh38
Transcripts
NM_001289967.1 NM_006785.3 NM_173844.2 XM_011525794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6983506 c.12delG p.D6fs*67 Chr.18: 58671655-58671655 on Build GRCh38
6393136 c.20C>T p.P7L Chr.18: 58671663-58671663 on Build GRCh38
5677203 c.158G>T p.G53V Chr.18: 58671801-58671801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003010019 Chr.18: 58671876-58671876 on Build GRCh38
rs966640423 Chr.18: 58671778-58671778 on Build GRCh38
rs994799429 Chr.18: 58671710-58671710 on Build GRCh38
rs1051462565 Chr.18: 58671720-58671720 on Build GRCh38
rs971971018 Chr.18: 58671987-58671987 on Build GRCh38
rs963317512 Chr.18: 58671976-58671976 on Build GRCh38
rs549192343 Chr.18: 58671776-58671776 on Build GRCh38
rs528315278 Chr.18: 58671895-58671895 on Build GRCh38
rs549390447 Chr.18: 58672092-58672092 on Build GRCh38
rs986681540 Chr.18: 58671775-58671775 on Build GRCh38
rs943135898 Chr.18: 58672070-58672070 on Build GRCh38
rs556960858 Chr.18: 58672061-58672061 on Build GRCh38
rs1056811993 Chr.18: 58671959-58671959 on Build GRCh38
rs1020525635 Chr.18: 58671845-58671845 on Build GRCh38
rs909231316 Chr.18: 58671937-58671937 on Build GRCh38
rs1019648492 Chr.18: 58672101-58672101 on Build GRCh38
rs933129506 Chr.18: 58671697-58671697 on Build GRCh38
rs937923201 Chr.18: 58671917-58671917 on Build GRCh38
rs953528575 Chr.18: 58672095-58672095 on Build GRCh38
rs959083747 Chr.18: 58671894-58671894 on Build GRCh38
rs397858172 Chr.18: 58672121-58672121 on Build GRCh38
rs941975162 Chr.18: 58671684-58671684 on Build GRCh38
rs888829305 Chr.18: 58671735-58671735 on Build GRCh38
rs967542589 Chr.18: 58671864-58671864 on Build GRCh38
rs546780839 Chr.18: 58671962-58671962 on Build GRCh38
rs866625194 Chr.18: 58671749-58671749 on Build GRCh38
rs1027827994 Chr.18: 58671713-58671713 on Build GRCh38
rs1053513904 Chr.18: 58672072-58672072 on Build GRCh38
rs919134273 Chr.18: 58671988-58671988 on Build GRCh38
rs1036222536 Chr.18: 58671685-58671685 on Build GRCh38
rs538885990 Chr.18: 58672005-58672005 on Build GRCh38
rs147329646 Chr.18: 58672076-58672076 on Build GRCh38
rs754701003 Chr.18: 58671678-58671678 on Build GRCh38
rs567480682 Chr.18: 58671893-58671893 on Build GRCh38
rs796640136 Chr.18: 58672053-58672054 on Build GRCh38
rs896331836 Chr.18: 58671658-58671658 on Build GRCh38
rs987901122 Chr.18: 58671822-58671822 on Build GRCh38
rs910381874 Chr.18: 58672033-58672033 on Build GRCh38
rs571572816 Chr.18: 58671997-58671997 on Build GRCh38
rs953666345 Chr.18: 58671743-58671743 on Build GRCh38
rs946328697 Chr.18: 58671890-58671890 on Build GRCh38
rs1027629971 Chr.18: 58672091-58672091 on Build GRCh38
rs919101714 Chr.18: 58671691-58671691 on Build GRCh38
rs893459894 Chr.18: 58671798-58671798 on Build GRCh38
rs1009198493 Chr.18: 58672096-58672096 on Build GRCh38
rs1035857224 Chr.18: 58671882-58671882 on Build GRCh38
rs966234450 Chr.18: 58672116-58672116 on Build GRCh38
rs1052066663 Chr.18: 58672017-58672017 on Build GRCh38
rs1011849690 Chr.18: 58671823-58671823 on Build GRCh38

View Full Product Details