Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63335084-63335500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295809 c.845C>T p.A282V Chr.18: 63335291-63335291 on Build GRCh38
5172720 c.827C>T p.A276V Chr.18: 63335309-63335309 on Build GRCh38
7192140 c.802G>T p.G268C Chr.18: 63335334-63335334 on Build GRCh38
6433590 c.778C>T p.Q260* Chr.18: 63335358-63335358 on Build GRCh38
989457 c.818T>C p.M273T Chr.18: 63335318-63335318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746111032 Chr.18: 63335278-63335278 on Build GRCh38
rs1041712635 Chr.18: 63335475-63335475 on Build GRCh38
rs752611378 Chr.18: 63335257-63335257 on Build GRCh38
rs61758854 Chr.18: 63335419-63335419 on Build GRCh38
rs192158889 Chr.18: 63335418-63335418 on Build GRCh38
rs780067439 Chr.18: 63335354-63335354 on Build GRCh38
rs919600678 Chr.18: 63335449-63335449 on Build GRCh38
rs372823909 Chr.18: 63335385-63335385 on Build GRCh38
rs903474670 Chr.18: 63335125-63335125 on Build GRCh38
rs755751272 Chr.18: 63335232-63335232 on Build GRCh38
rs564896316 Chr.18: 63335277-63335277 on Build GRCh38
rs962008550 Chr.18: 63335370-63335370 on Build GRCh38
rs371294721 Chr.18: 63335222-63335222 on Build GRCh38
rs758475771 Chr.18: 63335184-63335184 on Build GRCh38
rs900023088 Chr.18: 63335143-63335143 on Build GRCh38
rs931527156 Chr.18: 63335453-63335453 on Build GRCh38
rs879008336 Chr.18: 63335448-63335448 on Build GRCh38
rs766039383 Chr.18: 63335390-63335390 on Build GRCh38
rs1008794161 Chr.18: 63335134-63335134 on Build GRCh38
rs754621278 Chr.18: 63335382-63335383 on Build GRCh38
rs567222247 Chr.18: 63335405-63335405 on Build GRCh38
rs777579624 Chr.18: 63335271-63335271 on Build GRCh38
rs139372995 Chr.18: 63335438-63335438 on Build GRCh38
rs944274585 Chr.18: 63335463-63335463 on Build GRCh38
rs186054237 Chr.18: 63335342-63335342 on Build GRCh38
rs778987260 Chr.18: 63335253-63335253 on Build GRCh38
rs753410186 Chr.18: 63335361-63335361 on Build GRCh38
rs753364832 Chr.18: 63335246-63335246 on Build GRCh38
rs951679830 Chr.18: 63335220-63335220 on Build GRCh38
rs560475341 Chr.18: 63335311-63335311 on Build GRCh38
rs774845467 Chr.18: 63335353-63335353 on Build GRCh38
rs1028701344 Chr.18: 63335194-63335194 on Build GRCh38
rs753463390 Chr.18: 63335392-63335392 on Build GRCh38
rs891794996 Chr.18: 63335142-63335142 on Build GRCh38
rs755020391 Chr.18: 63335247-63335247 on Build GRCh38
rs372863936 Chr.18: 63335412-63335412 on Build GRCh38
rs773593231 Chr.18: 63335373-63335373 on Build GRCh38
rs759118040 Chr.18: 63335395-63335395 on Build GRCh38
rs774757639 Chr.18: 63335219-63335219 on Build GRCh38
rs765919072 Chr.18: 63335248-63335248 on Build GRCh38
rs765844985 Chr.18: 63335245-63335245 on Build GRCh38
rs1019725166 Chr.18: 63335122-63335122 on Build GRCh38
rs758285669 Chr.18: 63335261-63335261 on Build GRCh38
rs534772111 Chr.18: 63335447-63335447 on Build GRCh38
rs762476341 Chr.18: 63335226-63335229 on Build GRCh38
rs924844437 Chr.18: 63335452-63335452 on Build GRCh38
rs914695125 Chr.18: 63335430-63335430 on Build GRCh38
rs551957193 Chr.18: 63335468-63335468 on Build GRCh38
rs979388871 Chr.18: 63335312-63335312 on Build GRCh38
rs772354840 Chr.18: 63335364-63335364 on Build GRCh38
rs959256437 Chr.18: 63335372-63335372 on Build GRCh38
rs144314185 Chr.18: 63335113-63335113 on Build GRCh38
rs749358661 Chr.18: 63335345-63335345 on Build GRCh38
rs995858201 Chr.18: 63335165-63335165 on Build GRCh38
rs750065103 Chr.18: 63335223-63335223 on Build GRCh38

View Full Product Details