Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 66501482-66501988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH19
Entrez Gene ID
28513
Gene Name
cadherin 19
Gene Aliases
CDH7, CDH7L2
UniGene
Hs.42771
Gene Chromosome Location
Chr.18: 66501187-66604241 on Build GRCh38
Transcripts
NM_001271028.1 NM_021153.3 NR_073130.1 XM_011525931.2 XM_017025711.1

SNP Detail

SNP ID Chromosome Location
rs775188720 Chr.18: 66501517-66501517 on Build GRCh38
rs556438964 Chr.18: 66501923-66501923 on Build GRCh38
rs963183467 Chr.18: 66501880-66501880 on Build GRCh38
rs1003863346 Chr.18: 66501846-66501847 on Build GRCh38
rs929003816 Chr.18: 66501913-66501913 on Build GRCh38
rs949014853 Chr.18: 66501920-66501920 on Build GRCh38
rs897499294 Chr.18: 66501642-66501642 on Build GRCh38
rs980658532 Chr.18: 66501885-66501885 on Build GRCh38
rs909798017 Chr.18: 66501536-66501536 on Build GRCh38
rs746899099 Chr.18: 66501523-66501523 on Build GRCh38
rs192523245 Chr.18: 66501691-66501691 on Build GRCh38
rs928953357 Chr.18: 66501644-66501644 on Build GRCh38
rs566196927 Chr.18: 66501736-66501736 on Build GRCh38
rs1056522135 Chr.18: 66501627-66501627 on Build GRCh38
rs112763761 Chr.18: 66501724-66501724 on Build GRCh38
rs114416011 Chr.18: 66501823-66501823 on Build GRCh38
rs113883158 Chr.18: 66501560-66501560 on Build GRCh38
rs1039227828 Chr.18: 66501958-66501958 on Build GRCh38
rs956241872 Chr.18: 66501815-66501815 on Build GRCh38
rs933181188 Chr.18: 66501577-66501577 on Build GRCh38
rs2097020 Chr.18: 66501779-66501779 on Build GRCh38
rs151072903 Chr.18: 66501933-66501933 on Build GRCh38
rs148994062 Chr.18: 66501792-66501792 on Build GRCh38
rs988465022 Chr.18: 66501552-66501552 on Build GRCh38
rs904094829 Chr.18: 66501778-66501778 on Build GRCh38
rs954632086 Chr.18: 66501846-66501846 on Build GRCh38
rs530125730 Chr.18: 66501702-66501702 on Build GRCh38
rs537356308 Chr.18: 66501809-66501809 on Build GRCh38
rs565166067 Chr.18: 66501670-66501670 on Build GRCh38
rs145813194 Chr.18: 66501760-66501760 on Build GRCh38
rs985978787 Chr.18: 66501583-66501583 on Build GRCh38
rs913926625 Chr.18: 66501660-66501660 on Build GRCh38
rs142919788 Chr.18: 66501852-66501852 on Build GRCh38
rs776417782 Chr.18: 66501676-66501676 on Build GRCh38
rs531662666 Chr.18: 66501616-66501616 on Build GRCh38
rs973158042 Chr.18: 66501905-66501905 on Build GRCh38
rs572125066 Chr.18: 66501909-66501909 on Build GRCh38
rs941745514 Chr.18: 66501623-66501623 on Build GRCh38
rs560888666 Chr.18: 66501603-66501603 on Build GRCh38
rs770662667 Chr.18: 66501604-66501604 on Build GRCh38
rs1023079073 Chr.18: 66501713-66501713 on Build GRCh38
rs910398633 Chr.18: 66501860-66501860 on Build GRCh38
rs960522183 Chr.18: 66501894-66501894 on Build GRCh38
rs773952443 Chr.18: 66501782-66501782 on Build GRCh38
rs141287931 Chr.18: 66501962-66501962 on Build GRCh38
rs1037425785 Chr.18: 66501630-66501630 on Build GRCh38
rs543696311 Chr.18: 66501669-66501669 on Build GRCh38
rs896067438 Chr.18: 66501683-66501683 on Build GRCh38
rs992693639 Chr.18: 66501897-66501897 on Build GRCh38
rs565203840 Chr.18: 66501945-66501945 on Build GRCh38
rs926444115 Chr.18: 66501891-66501891 on Build GRCh38
rs117595206 Chr.18: 66501918-66501918 on Build GRCh38
rs567623719 Chr.18: 66501848-66501848 on Build GRCh38
rs184526704 Chr.18: 66501832-66501832 on Build GRCh38
rs1000285078 Chr.18: 66501961-66501961 on Build GRCh38
rs866874432 Chr.18: 66501836-66501836 on Build GRCh38
rs943627574 Chr.18: 66501935-66501935 on Build GRCh38
rs538338079 Chr.18: 66501914-66501914 on Build GRCh38
rs558864373 Chr.18: 66501816-66501816 on Build GRCh38

View Full Product Details