Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40991574-40991981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001301237.1 NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909509 c.684G>T p.E228D Chr.1: 40991809-40991809 on Build GRCh38
5342418 c.694A>G p.M232V Chr.1: 40991819-40991819 on Build GRCh38
2169414 c.692C>G p.S231* Chr.1: 40991817-40991817 on Build GRCh38
5342420 c.715A>G p.M239V Chr.1: 40991819-40991819 on Build GRCh38
5342419 c.694A>G p.M232V Chr.1: 40991819-40991819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778163725 Chr.1: 40991852-40991852 on Build GRCh38
rs778894938 Chr.1: 40991869-40991869 on Build GRCh38
rs918240530 Chr.1: 40991611-40991611 on Build GRCh38
rs769748811 Chr.1: 40991955-40991955 on Build GRCh38
rs763746296 Chr.1: 40991737-40991737 on Build GRCh38
rs761815773 Chr.1: 40991957-40991957 on Build GRCh38
rs745857706 Chr.1: 40991888-40991888 on Build GRCh38
rs567455441 Chr.1: 40991617-40991617 on Build GRCh38
rs984357112 Chr.1: 40991711-40991711 on Build GRCh38
rs770898513 Chr.1: 40991862-40991862 on Build GRCh38
rs756348375 Chr.1: 40991840-40991840 on Build GRCh38
rs78152902 Chr.1: 40991923-40991923 on Build GRCh38
rs951022539 Chr.1: 40991662-40991662 on Build GRCh38
rs760251053 Chr.1: 40991735-40991735 on Build GRCh38
rs375310353 Chr.1: 40991725-40991725 on Build GRCh38
rs865927710 Chr.1: 40991917-40991917 on Build GRCh38
rs748716482 Chr.1: 40991776-40991776 on Build GRCh38
rs899930970 Chr.1: 40991639-40991639 on Build GRCh38
rs781197940 Chr.1: 40991783-40991783 on Build GRCh38
rs976426632 Chr.1: 40991609-40991609 on Build GRCh38
rs755074273 Chr.1: 40991778-40991778 on Build GRCh38
rs867705252 Chr.1: 40991918-40991918 on Build GRCh38
rs776725479 Chr.1: 40991646-40991646 on Build GRCh38
rs752058241 Chr.1: 40991774-40991774 on Build GRCh38
rs752830298 Chr.1: 40991800-40991800 on Build GRCh38
rs74068298 Chr.1: 40991608-40991608 on Build GRCh38
rs775201825 Chr.1: 40991731-40991731 on Build GRCh38
rs759063799 Chr.1: 40991723-40991723 on Build GRCh38
rs530062089 Chr.1: 40991656-40991656 on Build GRCh38
rs766882222 Chr.1: 40991753-40991753 on Build GRCh38
rs750733212 Chr.1: 40991738-40991738 on Build GRCh38
rs1011953955 Chr.1: 40991692-40991692 on Build GRCh38
rs749060604 Chr.1: 40991855-40991855 on Build GRCh38
rs550001215 Chr.1: 40991949-40991949 on Build GRCh38
rs371960232 Chr.1: 40991721-40991721 on Build GRCh38
rs763344393 Chr.1: 40991749-40991749 on Build GRCh38

View Full Product Details