Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41017909-41018415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFNL1
Entrez Gene ID
200172
Gene Name
schlafen like 1
Gene Aliases
-
UniGene
Hs.194609
Gene Chromosome Location
Chr.1: 41015597-41023444 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2169472 c.665C>T p.T222I Chr.1: 41017927-41017927 on Build GRCh38
5959526 c.580G>A p.G194S Chr.1: 41018012-41018012 on Build GRCh38
4007984 c.618C>T p.I206I Chr.1: 41017974-41017974 on Build GRCh38
3944061 c.667C>A p.R223S Chr.1: 41017925-41017925 on Build GRCh38
1730940 c.613C>A p.Q205K Chr.1: 41017979-41017979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41268105 Chr.1: 41018021-41018021 on Build GRCh38
rs867750331 Chr.1: 41018099-41018099 on Build GRCh38
rs751378966 Chr.1: 41018059-41018059 on Build GRCh38
rs1034035817 Chr.1: 41018154-41018156 on Build GRCh38
rs759690123 Chr.1: 41018145-41018153 on Build GRCh38
rs769311448 Chr.1: 41017948-41017948 on Build GRCh38
rs985714435 Chr.1: 41018283-41018283 on Build GRCh38
rs772975519 Chr.1: 41017956-41017956 on Build GRCh38
rs964293275 Chr.1: 41018124-41018124 on Build GRCh38
rs750550948 Chr.1: 41018053-41018053 on Build GRCh38
rs779945577 Chr.1: 41018000-41018000 on Build GRCh38
rs564836842 Chr.1: 41018236-41018236 on Build GRCh38
rs35547639 Chr.1: 41018245-41018246 on Build GRCh38
rs762228548 Chr.1: 41018041-41018041 on Build GRCh38
rs777259636 Chr.1: 41018024-41018024 on Build GRCh38
rs556560244 Chr.1: 41017966-41017966 on Build GRCh38
rs754934380 Chr.1: 41018002-41018002 on Build GRCh38
rs190072621 Chr.1: 41018153-41018153 on Build GRCh38
rs758706343 Chr.1: 41017983-41017983 on Build GRCh38
rs11209361 Chr.1: 41018279-41018279 on Build GRCh38
rs558801329 Chr.1: 41018012-41018012 on Build GRCh38
rs142668626 Chr.1: 41017941-41017941 on Build GRCh38
rs888276470 Chr.1: 41018156-41018156 on Build GRCh38
rs1055044234 Chr.1: 41018380-41018380 on Build GRCh38
rs377035951 Chr.1: 41018058-41018058 on Build GRCh38
rs768996233 Chr.1: 41018161-41018161 on Build GRCh38
rs34386830 Chr.1: 41018108-41018108 on Build GRCh38
rs560848778 Chr.1: 41018286-41018286 on Build GRCh38
rs988936047 Chr.1: 41018305-41018305 on Build GRCh38
rs773273530 Chr.1: 41017997-41017998 on Build GRCh38
rs763744549 Chr.1: 41018343-41018343 on Build GRCh38
rs538664602 Chr.1: 41018334-41018334 on Build GRCh38
rs531228234 Chr.1: 41018185-41018185 on Build GRCh38
rs527338365 Chr.1: 41018322-41018322 on Build GRCh38
rs768735444 Chr.1: 41017937-41017937 on Build GRCh38
rs764695402 Chr.1: 41017972-41017972 on Build GRCh38
rs761123920 Chr.1: 41017969-41017969 on Build GRCh38
rs781241264 Chr.1: 41018006-41018006 on Build GRCh38
rs1005376440 Chr.1: 41018169-41018169 on Build GRCh38
rs1022960701 Chr.1: 41018112-41018112 on Build GRCh38
rs766432124 Chr.1: 41018134-41018136 on Build GRCh38
rs997043032 Chr.1: 41018352-41018352 on Build GRCh38
rs888331959 Chr.1: 41018385-41018385 on Build GRCh38
rs1009268933 Chr.1: 41017990-41017990 on Build GRCh38
rs545359197 Chr.1: 41018009-41018009 on Build GRCh38
rs558865803 Chr.1: 41018386-41018386 on Build GRCh38
rs376280126 Chr.1: 41018001-41018001 on Build GRCh38
rs377572340 Chr.1: 41017962-41017962 on Build GRCh38
rs372325772 Chr.1: 41018179-41018179 on Build GRCh38
rs748885819 Chr.1: 41018020-41018020 on Build GRCh38
rs908581349 Chr.1: 41018230-41018230 on Build GRCh38
rs1015357826 Chr.1: 41018389-41018389 on Build GRCh38
rs142089953 Chr.1: 41018262-41018262 on Build GRCh38
rs369745596 Chr.1: 41017968-41017968 on Build GRCh38
rs147944502 Chr.1: 41018013-41018013 on Build GRCh38
rs536581223 Chr.1: 41018297-41018297 on Build GRCh38
rs777290519 Chr.1: 41018093-41018093 on Build GRCh38
rs148667156 Chr.1: 41018210-41018210 on Build GRCh38
rs367665847 Chr.1: 41018023-41018023 on Build GRCh38
rs374360767 Chr.1: 41018296-41018296 on Build GRCh38
rs770543585 Chr.1: 41018096-41018096 on Build GRCh38
rs765302179 Chr.1: 41018049-41018049 on Build GRCh38
rs368721328 Chr.1: 41018270-41018270 on Build GRCh38
rs547631112 Chr.1: 41018269-41018269 on Build GRCh38
rs763038202 Chr.1: 41018010-41018038 on Build GRCh38
rs780989062 Chr.1: 41018068-41018068 on Build GRCh38
rs149541948 Chr.1: 41018091-41018091 on Build GRCh38
rs757413470 Chr.1: 41017974-41017974 on Build GRCh38
rs910668239 Chr.1: 41018021-41018038 on Build GRCh38
rs748832601 Chr.1: 41018094-41018094 on Build GRCh38
rs770233255 Chr.1: 41018208-41018208 on Build GRCh38
rs754860594 Chr.1: 41018067-41018067 on Build GRCh38
rs1052180716 Chr.1: 41018312-41018312 on Build GRCh38
rs777154246 Chr.1: 41018016-41018016 on Build GRCh38
rs750588926 Chr.1: 41017980-41017980 on Build GRCh38
rs1053523284 Chr.1: 41018092-41018092 on Build GRCh38
rs1033489020 Chr.1: 41018010-41018010 on Build GRCh38
rs947926466 Chr.1: 41018277-41018277 on Build GRCh38
rs926224891 Chr.1: 41018284-41018284 on Build GRCh38
rs982812666 Chr.1: 41018226-41018226 on Build GRCh38
rs906203866 Chr.1: 41018148-41018153 on Build GRCh38
rs763227125 Chr.1: 41018056-41018056 on Build GRCh38
rs974069801 Chr.1: 41018237-41018237 on Build GRCh38
rs945116826 Chr.1: 41018330-41018330 on Build GRCh38
rs751622686 Chr.1: 41018133-41018134 on Build GRCh38
rs977259917 Chr.1: 41018254-41018254 on Build GRCh38
rs897009032 Chr.1: 41018394-41018394 on Build GRCh38
rs767807067 Chr.1: 41017965-41017965 on Build GRCh38
rs1029809797 Chr.1: 41018368-41018368 on Build GRCh38
rs937712787 Chr.1: 41018369-41018369 on Build GRCh38
rs753836544 Chr.1: 41017973-41017973 on Build GRCh38
rs368399441 Chr.1: 41018329-41018329 on Build GRCh38

View Full Product Details