Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 617559-618060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLRMT
Entrez Gene ID
5442
Gene Name
RNA polymerase mitochondrial
Gene Aliases
APOLMT, MTRNAP, MTRPOL, h-mtRPOL
UniGene
Hs.254113
Gene Chromosome Location
Chr.19: 617221-633592 on Build GRCh38
Transcripts
NM_005035.3 XM_005259580.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7402240 c.3622T>A p.S1208T Chr.19: 617574-617574 on Build GRCh38
4082519 c.3451G>A p.D1151N Chr.19: 617821-617821 on Build GRCh38
5576967 c.3528C>T p.S1176S Chr.19: 617789-617789 on Build GRCh38
5745367 c.3574G>A p.E1192K Chr.19: 617622-617622 on Build GRCh38
5786662 c.3502C>T p.R1168W Chr.19: 617649-617649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56263332 Chr.19: 617988-617988 on Build GRCh38
rs766021518 Chr.19: 617645-617645 on Build GRCh38
rs1056616253 Chr.19: 617929-617929 on Build GRCh38
rs749370066 Chr.19: 617658-617658 on Build GRCh38
rs964862358 Chr.19: 618032-618032 on Build GRCh38
rs41551212 Chr.19: 617614-617614 on Build GRCh38
rs746517573 Chr.19: 617967-617967 on Build GRCh38
rs937756682 Chr.19: 617936-617936 on Build GRCh38
rs201558272 Chr.19: 617838-617838 on Build GRCh38
rs374257003 Chr.19: 617834-617834 on Build GRCh38
rs762591427 Chr.19: 617757-617757 on Build GRCh38
rs370155132 Chr.19: 617673-617673 on Build GRCh38
rs761217365 Chr.19: 617806-617806 on Build GRCh38
rs147740567 Chr.19: 617634-617634 on Build GRCh38
rs745682275 Chr.19: 617801-617801 on Build GRCh38
rs376900734 Chr.19: 617799-617799 on Build GRCh38
rs753341461 Chr.19: 617647-617647 on Build GRCh38
rs780465105 Chr.19: 617669-617670 on Build GRCh38
rs771895150 Chr.19: 617739-617739 on Build GRCh38
rs752275492 Chr.19: 617686-617686 on Build GRCh38
rs772812075 Chr.19: 617674-617674 on Build GRCh38
rs761580693 Chr.19: 617594-617594 on Build GRCh38
rs770120131 Chr.19: 617590-617596 on Build GRCh38
rs975268001 Chr.19: 617976-617976 on Build GRCh38
rs41554212 Chr.19: 617769-617769 on Build GRCh38
rs762169550 Chr.19: 617690-617690 on Build GRCh38
rs375923605 Chr.19: 617915-617915 on Build GRCh38
rs992553565 Chr.19: 617637-617637 on Build GRCh38
rs200249697 Chr.19: 617861-617861 on Build GRCh38
rs41563414 Chr.19: 617924-617924 on Build GRCh38
rs765557938 Chr.19: 617774-617774 on Build GRCh38
rs202213737 Chr.19: 617656-617656 on Build GRCh38
rs764610763 Chr.19: 617683-617683 on Build GRCh38
rs955508277 Chr.19: 617994-617994 on Build GRCh38
rs759104301 Chr.19: 617648-617648 on Build GRCh38
rs765354862 Chr.19: 617828-617828 on Build GRCh38
rs942527538 Chr.19: 617744-617744 on Build GRCh38
rs1025765099 Chr.19: 617960-617960 on Build GRCh38
rs571929257 Chr.19: 617682-617682 on Build GRCh38
rs774214180 Chr.19: 617632-617632 on Build GRCh38
rs749542005 Chr.19: 617672-617672 on Build GRCh38
rs745636992 Chr.19: 617840-617840 on Build GRCh38
rs896727808 Chr.19: 618033-618033 on Build GRCh38
rs947397632 Chr.19: 617959-617959 on Build GRCh38
rs774782721 Chr.19: 617759-617760 on Build GRCh38
rs748729176 Chr.19: 617803-617803 on Build GRCh38
rs946744185 Chr.19: 618019-618019 on Build GRCh38
rs755262121 Chr.19: 617734-617734 on Build GRCh38
rs781364079 Chr.19: 617653-617653 on Build GRCh38
rs777745973 Chr.19: 617745-617745 on Build GRCh38
rs755793386 Chr.19: 617599-617599 on Build GRCh38
rs748934263 Chr.19: 617848-617848 on Build GRCh38
rs770557680 Chr.19: 617677-617677 on Build GRCh38
rs776313032 Chr.19: 617681-617681 on Build GRCh38
rs750832613 Chr.19: 617655-617655 on Build GRCh38
rs890476184 Chr.19: 617989-617989 on Build GRCh38
rs528834580 Chr.19: 617759-617759 on Build GRCh38
rs541061783 Chr.19: 618009-618009 on Build GRCh38
rs183246923 Chr.19: 617748-617748 on Build GRCh38
rs755990734 Chr.19: 617844-617844 on Build GRCh38
rs768074385 Chr.19: 617691-617691 on Build GRCh38
rs760259302 Chr.19: 617644-617644 on Build GRCh38
rs371354135 Chr.19: 617593-617593 on Build GRCh38
rs368080596 Chr.19: 617885-617885 on Build GRCh38
rs769082897 Chr.19: 617689-617692 on Build GRCh38
rs954810337 Chr.19: 617699-617699 on Build GRCh38
rs904903034 Chr.19: 617927-617927 on Build GRCh38
rs1056588536 Chr.19: 618032-618033 on Build GRCh38
rs200224860 Chr.19: 617813-617813 on Build GRCh38
rs1043759200 Chr.19: 618030-618030 on Build GRCh38
rs570198637 Chr.19: 617586-617586 on Build GRCh38
rs772690140 Chr.19: 617873-617876 on Build GRCh38
rs942626606 Chr.19: 617701-617701 on Build GRCh38
rs770154364 Chr.19: 617873-617873 on Build GRCh38
rs896752964 Chr.19: 617931-617931 on Build GRCh38
rs767186851 Chr.19: 617596-617596 on Build GRCh38
rs371613474 Chr.19: 617807-617807 on Build GRCh38
rs377569522 Chr.19: 617768-617768 on Build GRCh38
rs1046129304 Chr.19: 617797-617797 on Build GRCh38
rs779545393 Chr.19: 617846-617846 on Build GRCh38
rs913592519 Chr.19: 617909-617909 on Build GRCh38
rs748261773 Chr.19: 617738-617738 on Build GRCh38
rs113833322 Chr.19: 617862-617862 on Build GRCh38
rs778496835 Chr.19: 617858-617858 on Build GRCh38
rs1037908653 Chr.19: 617955-617955 on Build GRCh38
rs200460541 Chr.19: 617792-617792 on Build GRCh38
rs148585853 Chr.19: 617613-617613 on Build GRCh38
rs563479140 Chr.19: 617972-617983 on Build GRCh38
rs755085446 Chr.19: 617773-617773 on Build GRCh38
rs559370508 Chr.19: 618031-618031 on Build GRCh38
rs774797564 Chr.19: 617754-617754 on Build GRCh38
rs760526807 Chr.19: 617675-617675 on Build GRCh38
rs151178174 Chr.19: 617833-617833 on Build GRCh38
rs777128308 Chr.19: 617821-617821 on Build GRCh38
rs993743235 Chr.19: 617817-617817 on Build GRCh38
rs768447989 Chr.19: 617854-617854 on Build GRCh38
rs377402063 Chr.19: 617721-617721 on Build GRCh38
rs765825072 Chr.19: 617603-617603 on Build GRCh38
rs762220644 Chr.19: 617825-617826 on Build GRCh38
rs55791972 Chr.19: 617981-617981 on Build GRCh38
rs563922410 Chr.19: 617737-617737 on Build GRCh38
rs750418584 Chr.19: 617920-617920 on Build GRCh38
rs545557652 Chr.19: 617703-617703 on Build GRCh38
rs144281668 Chr.19: 617804-617804 on Build GRCh38
rs778207346 Chr.19: 617609-617609 on Build GRCh38
rs41546612 Chr.19: 617963-617963 on Build GRCh38
rs775746912 Chr.19: 617585-617585 on Build GRCh38
rs775657210 Chr.19: 617619-617619 on Build GRCh38
rs533719840 Chr.19: 618037-618037 on Build GRCh38
rs75913143 Chr.19: 617837-617837 on Build GRCh38
rs745935945 Chr.19: 617583-617583 on Build GRCh38
rs553663334 Chr.19: 617923-617923 on Build GRCh38
rs112271909 Chr.19: 617886-617886 on Build GRCh38
rs149355349 Chr.19: 617608-617608 on Build GRCh38
rs373904638 Chr.19: 617866-617866 on Build GRCh38
rs534986984 Chr.19: 617913-617913 on Build GRCh38
rs757206165 Chr.19: 617793-617793 on Build GRCh38
rs150648879 Chr.19: 617582-617582 on Build GRCh38
rs1000678728 Chr.19: 618034-618034 on Build GRCh38
rs913851137 Chr.19: 618013-618017 on Build GRCh38
rs199912442 Chr.19: 617623-617623 on Build GRCh38
rs757607880 Chr.19: 617611-617611 on Build GRCh38
rs775738468 Chr.19: 617881-617881 on Build GRCh38
rs373082226 Chr.19: 617695-617695 on Build GRCh38
rs377209245 Chr.19: 617649-617649 on Build GRCh38
rs375762747 Chr.19: 617598-617598 on Build GRCh38
rs1023788509 Chr.19: 617710-617735 on Build GRCh38
rs766996006 Chr.19: 617808-617808 on Build GRCh38
rs374409008 Chr.19: 617789-617789 on Build GRCh38
rs557577694 Chr.19: 617966-617966 on Build GRCh38
rs769501263 Chr.19: 617750-617750 on Build GRCh38
rs764144041 Chr.19: 617587-617587 on Build GRCh38
rs780405645 Chr.19: 617697-617697 on Build GRCh38
rs555413985 Chr.19: 617993-617993 on Build GRCh38
rs780495579 Chr.19: 617657-617657 on Build GRCh38
rs754156760 Chr.19: 617707-617707 on Build GRCh38
rs142357282 Chr.19: 617749-617749 on Build GRCh38
rs942642442 Chr.19: 617592-617592 on Build GRCh38
rs575759646 Chr.19: 617746-617746 on Build GRCh38
rs761939486 Chr.19: 617900-617900 on Build GRCh38
rs752702101 Chr.19: 617788-617788 on Build GRCh38
rs138709888 Chr.19: 617584-617584 on Build GRCh38
rs778880802 Chr.19: 617668-617668 on Build GRCh38
rs768669931 Chr.19: 617666-617666 on Build GRCh38
rs757816473 Chr.19: 617650-617650 on Build GRCh38
rs761647797 Chr.19: 617633-617633 on Build GRCh38
rs753002965 Chr.19: 617831-617831 on Build GRCh38
rs754265267 Chr.19: 617770-617770 on Build GRCh38
rs1026546171 Chr.19: 617970-617970 on Build GRCh38
rs146355392 Chr.19: 617822-617822 on Build GRCh38
rs1032203969 Chr.19: 618035-618035 on Build GRCh38
rs751635676 Chr.19: 617895-617895 on Build GRCh38
rs769327958 Chr.19: 617802-617802 on Build GRCh38
rs753359532 Chr.19: 617604-617604 on Build GRCh38

View Full Product Details