Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 899954-900459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HDM4
Entrez Gene ID
91300
Gene Name
R3H domain containing 4
Gene Aliases
C19orf22
UniGene
Hs.557655
Gene Chromosome Location
Chr.19: 896503-913225 on Build GRCh38
Transcripts
NM_138774.3 XM_011528416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1003658 c.484G>A p.A162T Chr.19: 900138-900138 on Build GRCh38
379156 c.530G>A p.R177Q Chr.19: 900092-900092 on Build GRCh38
4719746 c.535G>A p.V179I Chr.19: 900087-900087 on Build GRCh38
2730937 c.483C>T p.P161P Chr.19: 900139-900139 on Build GRCh38
2730936 c.497G>A p.R166H Chr.19: 900125-900125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748300180 Chr.19: 900049-900049 on Build GRCh38
rs372127843 Chr.19: 900003-900003 on Build GRCh38
rs979780774 Chr.19: 900261-900261 on Build GRCh38
rs7253155 Chr.19: 900061-900061 on Build GRCh38
rs781175964 Chr.19: 900111-900111 on Build GRCh38
rs199817764 Chr.19: 900139-900139 on Build GRCh38
rs771544143 Chr.19: 900176-900176 on Build GRCh38
rs747079024 Chr.19: 900102-900102 on Build GRCh38
rs758583731 Chr.19: 900100-900100 on Build GRCh38
rs200112514 Chr.19: 900124-900124 on Build GRCh38
rs1023536901 Chr.19: 900216-900216 on Build GRCh38
rs777666939 Chr.19: 900101-900101 on Build GRCh38
rs745675321 Chr.19: 900120-900120 on Build GRCh38
rs773787621 Chr.19: 900085-900085 on Build GRCh38
rs375919968 Chr.19: 900196-900196 on Build GRCh38
rs763552609 Chr.19: 900076-900076 on Build GRCh38
rs1048929516 Chr.19: 900077-900077 on Build GRCh38
rs146459823 Chr.19: 900186-900186 on Build GRCh38
rs539357422 Chr.19: 900019-900019 on Build GRCh38
rs756530864 Chr.19: 900037-900037 on Build GRCh38
rs543115291 Chr.19: 900353-900353 on Build GRCh38
rs368367301 Chr.19: 900092-900092 on Build GRCh38
rs924073375 Chr.19: 900004-900004 on Build GRCh38
rs760072395 Chr.19: 900180-900180 on Build GRCh38
rs1014628587 Chr.19: 900231-900231 on Build GRCh38
rs775062281 Chr.19: 900071-900071 on Build GRCh38
rs761384140 Chr.19: 900135-900135 on Build GRCh38
rs940377542 Chr.19: 900090-900090 on Build GRCh38
rs929906226 Chr.19: 900312-900312 on Build GRCh38
rs754211163 Chr.19: 900041-900041 on Build GRCh38
rs112274080 Chr.19: 900022-900022 on Build GRCh38
rs992440854 Chr.19: 900289-900289 on Build GRCh38
rs112403370 Chr.19: 900039-900039 on Build GRCh38
rs761137352 Chr.19: 900088-900088 on Build GRCh38
rs770783798 Chr.19: 900063-900063 on Build GRCh38
rs965600047 Chr.19: 899997-899997 on Build GRCh38
rs938785390 Chr.19: 900364-900364 on Build GRCh38
rs975680558 Chr.19: 900093-900093 on Build GRCh38
rs202174132 Chr.19: 900144-900144 on Build GRCh38
rs953055697 Chr.19: 900272-900272 on Build GRCh38
rs947071989 Chr.19: 900030-900030 on Build GRCh38
rs768263109 Chr.19: 900170-900170 on Build GRCh38
rs753805069 Chr.19: 900018-900019 on Build GRCh38
rs776379157 Chr.19: 900012-900012 on Build GRCh38
rs780992860 Chr.19: 900161-900161 on Build GRCh38
rs1012888239 Chr.19: 900414-900414 on Build GRCh38
rs764234809 Chr.19: 900187-900187 on Build GRCh38
rs140866425 Chr.19: 900223-900223 on Build GRCh38
rs975796677 Chr.19: 900241-900241 on Build GRCh38
rs957063293 Chr.19: 900118-900118 on Build GRCh38
rs984393516 Chr.19: 900321-900322 on Build GRCh38
rs780345265 Chr.19: 900108-900109 on Build GRCh38
rs894471081 Chr.19: 900208-900208 on Build GRCh38
rs375154265 Chr.19: 900157-900157 on Build GRCh38
rs769492622 Chr.19: 900068-900068 on Build GRCh38
rs778618754 Chr.19: 900171-900171 on Build GRCh38
rs537357318 Chr.19: 900098-900098 on Build GRCh38
rs777141353 Chr.19: 900138-900138 on Build GRCh38
rs751648618 Chr.19: 900155-900155 on Build GRCh38
rs73507505 Chr.19: 900044-900044 on Build GRCh38
rs909678587 Chr.19: 900323-900323 on Build GRCh38
rs140670608 Chr.19: 900137-900137 on Build GRCh38
rs1012599319 Chr.19: 900209-900209 on Build GRCh38
rs145420343 Chr.19: 900094-900094 on Build GRCh38
rs752271599 Chr.19: 900020-900020 on Build GRCh38
rs768022273 Chr.19: 900023-900023 on Build GRCh38
rs777310157 Chr.19: 900179-900179 on Build GRCh38
rs924699225 Chr.19: 900359-900359 on Build GRCh38
rs572633918 Chr.19: 900112-900112 on Build GRCh38
rs777858028 Chr.19: 900056-900056 on Build GRCh38
rs372866218 Chr.19: 900054-900054 on Build GRCh38
rs752991735 Chr.19: 900142-900142 on Build GRCh38
rs371777297 Chr.19: 900141-900141 on Build GRCh38
rs11880031 Chr.19: 900096-900096 on Build GRCh38
rs894494344 Chr.19: 900395-900395 on Build GRCh38
rs147694333 Chr.19: 900110-900110 on Build GRCh38
rs914802146 Chr.19: 900283-900283 on Build GRCh38
rs1050696876 Chr.19: 900129-900129 on Build GRCh38
rs947556017 Chr.19: 900329-900329 on Build GRCh38
rs369345406 Chr.19: 900167-900167 on Build GRCh38
rs2306718 Chr.19: 899978-899978 on Build GRCh38
rs922968605 Chr.19: 900250-900250 on Build GRCh38
rs1036068543 Chr.19: 900188-900188 on Build GRCh38
rs776329473 Chr.19: 900066-900066 on Build GRCh38
rs918522729 Chr.19: 900308-900313 on Build GRCh38
rs533734705 Chr.19: 900125-900125 on Build GRCh38
rs1057217170 Chr.19: 900392-900392 on Build GRCh38
rs779277651 Chr.19: 900046-900046 on Build GRCh38
rs763317594 Chr.19: 900143-900143 on Build GRCh38
rs768271869 Chr.19: 900126-900126 on Build GRCh38
rs750317177 Chr.19: 900164-900164 on Build GRCh38
rs375768350 Chr.19: 900072-900072 on Build GRCh38
rs575909114 Chr.19: 900024-900024 on Build GRCh38
rs745670393 Chr.19: 900067-900067 on Build GRCh38
rs968878575 Chr.19: 900251-900251 on Build GRCh38
rs770404179 Chr.19: 900181-900181 on Build GRCh38
rs749053721 Chr.19: 900169-900169 on Build GRCh38
rs755384421 Chr.19: 900097-900097 on Build GRCh38
rs986816861 Chr.19: 900277-900277 on Build GRCh38
rs878957764 Chr.19: 900232-900232 on Build GRCh38
rs951983808 Chr.19: 900222-900222 on Build GRCh38
rs759158734 Chr.19: 900017-900017 on Build GRCh38
rs923554262 Chr.19: 900099-900099 on Build GRCh38

View Full Product Details