Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1011785-1012275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438442 c.786C>T p.Y262Y Chr.19: 1012121-1012121 on Build GRCh38
5527616 c.777C>T p.F259F Chr.19: 1012130-1012130 on Build GRCh38
5587340 c.887G>T p.S296I Chr.19: 1011947-1011947 on Build GRCh38
5572545 c.875A>G p.Y292C Chr.19: 1011959-1011959 on Build GRCh38
990359 c.942C>T p.F314F Chr.19: 1011892-1011892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018242058 Chr.19: 1011978-1011978 on Build GRCh38
rs571419166 Chr.19: 1012104-1012104 on Build GRCh38
rs372779173 Chr.19: 1012149-1012149 on Build GRCh38
rs762318089 Chr.19: 1011977-1011977 on Build GRCh38
rs763113473 Chr.19: 1012048-1012048 on Build GRCh38
rs376508766 Chr.19: 1012115-1012115 on Build GRCh38
rs770312147 Chr.19: 1012170-1012170 on Build GRCh38
rs767887576 Chr.19: 1011922-1011922 on Build GRCh38
rs772151792 Chr.19: 1011952-1011952 on Build GRCh38
rs373045338 Chr.19: 1012079-1012079 on Build GRCh38
rs372664489 Chr.19: 1011934-1011934 on Build GRCh38
rs779036356 Chr.19: 1011939-1011939 on Build GRCh38
rs768416321 Chr.19: 1011841-1011841 on Build GRCh38
rs756970497 Chr.19: 1012016-1012016 on Build GRCh38
rs750272262 Chr.19: 1012137-1012137 on Build GRCh38
rs746737331 Chr.19: 1011823-1011824 on Build GRCh38
rs756632385 Chr.19: 1012223-1012223 on Build GRCh38
rs555699990 Chr.19: 1012182-1012182 on Build GRCh38
rs868142017 Chr.19: 1012158-1012158 on Build GRCh38
rs750693343 Chr.19: 1011923-1011923 on Build GRCh38
rs777376395 Chr.19: 1012221-1012221 on Build GRCh38
rs758363429 Chr.19: 1011947-1011947 on Build GRCh38
rs376288077 Chr.19: 1011998-1011998 on Build GRCh38
rs763058552 Chr.19: 1012185-1012186 on Build GRCh38
rs752715881 Chr.19: 1011944-1011944 on Build GRCh38
rs746285002 Chr.19: 1012166-1012166 on Build GRCh38
rs778043353 Chr.19: 1012229-1012229 on Build GRCh38
rs993140126 Chr.19: 1011808-1011808 on Build GRCh38
rs779171731 Chr.19: 1012239-1012239 on Build GRCh38
rs7145 Chr.19: 1012121-1012121 on Build GRCh38
rs765714375 Chr.19: 1011823-1011823 on Build GRCh38
rs750688476 Chr.19: 1011872-1011872 on Build GRCh38
rs775601880 Chr.19: 1012032-1012033 on Build GRCh38
rs773574166 Chr.19: 1012030-1012030 on Build GRCh38
rs774357289 Chr.19: 1012180-1012180 on Build GRCh38
rs368144549 Chr.19: 1012062-1012062 on Build GRCh38
rs546409591 Chr.19: 1012088-1012088 on Build GRCh38
rs756185577 Chr.19: 1011836-1011836 on Build GRCh38
rs766501808 Chr.19: 1011932-1011932 on Build GRCh38
rs761935720 Chr.19: 1012181-1012181 on Build GRCh38
rs773528026 Chr.19: 1012111-1012111 on Build GRCh38
rs373820239 Chr.19: 1012015-1012015 on Build GRCh38
rs779954772 Chr.19: 1011837-1011837 on Build GRCh38
rs749859272 Chr.19: 1012006-1012006 on Build GRCh38
rs755090111 Chr.19: 1011935-1011935 on Build GRCh38
rs759853360 Chr.19: 1012042-1012042 on Build GRCh38
rs777142421 Chr.19: 1012092-1012092 on Build GRCh38
rs114980668 Chr.19: 1012025-1012025 on Build GRCh38
rs900181278 Chr.19: 1012086-1012086 on Build GRCh38
rs913854154 Chr.19: 1011866-1011866 on Build GRCh38
rs751864414 Chr.19: 1011833-1011833 on Build GRCh38
rs984460731 Chr.19: 1011943-1011943 on Build GRCh38
rs767228248 Chr.19: 1012060-1012060 on Build GRCh38
rs758954365 Chr.19: 1011909-1011909 on Build GRCh38
rs371837146 Chr.19: 1011858-1011858 on Build GRCh38
rs557204155 Chr.19: 1012130-1012130 on Build GRCh38
rs971337670 Chr.19: 1012020-1012020 on Build GRCh38
rs375642389 Chr.19: 1012009-1012009 on Build GRCh38
rs770631832 Chr.19: 1011824-1011825 on Build GRCh38
rs113138803 Chr.19: 1011857-1011857 on Build GRCh38
rs768814792 Chr.19: 1012236-1012236 on Build GRCh38
rs768868585 Chr.19: 1012218-1012218 on Build GRCh38
rs202085162 Chr.19: 1011957-1011957 on Build GRCh38
rs769808146 Chr.19: 1012033-1012040 on Build GRCh38
rs757904664 Chr.19: 1012220-1012220 on Build GRCh38
rs376521538 Chr.19: 1011820-1011820 on Build GRCh38
rs368276870 Chr.19: 1011834-1011834 on Build GRCh38
rs745540430 Chr.19: 1012024-1012024 on Build GRCh38
rs541183075 Chr.19: 1011972-1011972 on Build GRCh38
rs778133543 Chr.19: 1011829-1011829 on Build GRCh38
rs747496294 Chr.19: 1012085-1012085 on Build GRCh38
rs781761315 Chr.19: 1012165-1012165 on Build GRCh38
rs770683826 Chr.19: 1011964-1011964 on Build GRCh38
rs370000710 Chr.19: 1011810-1011810 on Build GRCh38
rs999915895 Chr.19: 1012090-1012090 on Build GRCh38
rs367589078 Chr.19: 1012141-1012141 on Build GRCh38
rs536573821 Chr.19: 1012175-1012175 on Build GRCh38
rs780686224 Chr.19: 1011966-1011966 on Build GRCh38
rs770199719 Chr.19: 1012099-1012099 on Build GRCh38
rs116854930 Chr.19: 1012000-1012000 on Build GRCh38
rs901307190 Chr.19: 1011831-1011831 on Build GRCh38
rs569127401 Chr.19: 1012144-1012144 on Build GRCh38
rs775474352 Chr.19: 1011812-1011812 on Build GRCh38
rs760146161 Chr.19: 1011855-1011855 on Build GRCh38
rs746698506 Chr.19: 1011900-1011900 on Build GRCh38
rs747628492 Chr.19: 1012159-1012159 on Build GRCh38
rs567321900 Chr.19: 1011815-1011815 on Build GRCh38
rs777638879 Chr.19: 1011956-1011956 on Build GRCh38
rs367572388 Chr.19: 1011974-1011974 on Build GRCh38
rs761146613 Chr.19: 1012031-1012031 on Build GRCh38
rs200423335 Chr.19: 1012213-1012213 on Build GRCh38
rs559473498 Chr.19: 1012235-1012235 on Build GRCh38
rs2240161 Chr.19: 1011824-1011824 on Build GRCh38
rs764633987 Chr.19: 1011910-1011910 on Build GRCh38
rs112523716 Chr.19: 1012005-1012005 on Build GRCh38
rs771912461 Chr.19: 1011848-1011848 on Build GRCh38
rs753512044 Chr.19: 1012147-1012147 on Build GRCh38
rs1010017655 Chr.19: 1012140-1012140 on Build GRCh38
rs376540839 Chr.19: 1011859-1011859 on Build GRCh38
rs781081522 Chr.19: 1012000-1012003 on Build GRCh38
rs758793008 Chr.19: 1011828-1011828 on Build GRCh38
rs750149443 Chr.19: 1012067-1012067 on Build GRCh38
rs372803823 Chr.19: 1012046-1012046 on Build GRCh38
rs187323927 Chr.19: 1011809-1011809 on Build GRCh38
rs774236130 Chr.19: 1011816-1011816 on Build GRCh38
rs780569470 Chr.19: 1012232-1012232 on Build GRCh38
rs776210172 Chr.19: 1011908-1011908 on Build GRCh38
rs898893951 Chr.19: 1011916-1011916 on Build GRCh38
rs774931387 Chr.19: 1011971-1011971 on Build GRCh38
rs756239009 Chr.19: 1011875-1011875 on Build GRCh38
rs555802063 Chr.19: 1011880-1011880 on Build GRCh38
rs759115798 Chr.19: 1012208-1012208 on Build GRCh38
rs749484472 Chr.19: 1012177-1012177 on Build GRCh38
rs140018971 Chr.19: 1012179-1012179 on Build GRCh38
rs745422323 Chr.19: 1011967-1011967 on Build GRCh38
rs200991222 Chr.19: 1012078-1012078 on Build GRCh38
rs753525081 Chr.19: 1012148-1012148 on Build GRCh38
rs772690124 Chr.19: 1012029-1012029 on Build GRCh38
rs368644367 Chr.19: 1012193-1012193 on Build GRCh38
rs775822771 Chr.19: 1012100-1012100 on Build GRCh38
rs117246624 Chr.19: 1012003-1012003 on Build GRCh38
rs753978179 Chr.19: 1012007-1012007 on Build GRCh38
rs1053072193 Chr.19: 1011845-1011845 on Build GRCh38
rs773098768 Chr.19: 1012187-1012187 on Build GRCh38
rs778303512 Chr.19: 1012154-1012154 on Build GRCh38
rs756359340 Chr.19: 1011929-1011929 on Build GRCh38
rs778449137 Chr.19: 1012082-1012082 on Build GRCh38
rs73497158 Chr.19: 1011895-1011895 on Build GRCh38
rs755828495 Chr.19: 1012070-1012070 on Build GRCh38
rs898845386 Chr.19: 1011807-1011807 on Build GRCh38
rs750987357 Chr.19: 1012222-1012222 on Build GRCh38
rs370964094 Chr.19: 1012037-1012037 on Build GRCh38
rs10420656 Chr.19: 1011847-1011847 on Build GRCh38
rs752768946 Chr.19: 1012010-1012010 on Build GRCh38
rs1052602516 Chr.19: 1012004-1012004 on Build GRCh38
rs201158183 Chr.19: 1012192-1012192 on Build GRCh38
rs1047422841 Chr.19: 1011945-1011945 on Build GRCh38
rs774422753 Chr.19: 1012238-1012238 on Build GRCh38
rs978880443 Chr.19: 1011850-1011850 on Build GRCh38
rs749267921 Chr.19: 1011840-1011840 on Build GRCh38
rs371078138 Chr.19: 1012012-1012012 on Build GRCh38
rs762694280 Chr.19: 1011814-1011814 on Build GRCh38
rs983603704 Chr.19: 1012215-1012215 on Build GRCh38
rs751940225 Chr.19: 1012214-1012214 on Build GRCh38
rs762266839 Chr.19: 1011917-1011917 on Build GRCh38
rs748213934 Chr.19: 1012249-1012249 on Build GRCh38
rs754010810 Chr.19: 1011933-1011933 on Build GRCh38
rs563678719 Chr.19: 1012014-1012014 on Build GRCh38
rs142318806 Chr.19: 1011983-1011983 on Build GRCh38
rs745778928 Chr.19: 1012029-1012036 on Build GRCh38
rs56377946 Chr.19: 1011811-1011811 on Build GRCh38
rs771965595 Chr.19: 1011892-1011892 on Build GRCh38
rs745982074 Chr.19: 1011835-1011835 on Build GRCh38
rs774806886 Chr.19: 1011915-1011915 on Build GRCh38
rs368626774 Chr.19: 1011869-1011869 on Build GRCh38
rs374586188 Chr.19: 1011854-1011854 on Build GRCh38
rs765491287 Chr.19: 1012045-1012045 on Build GRCh38
rs770433599 Chr.19: 1011907-1011907 on Build GRCh38
rs767764383 Chr.19: 1011870-1011870 on Build GRCh38
rs748007995 Chr.19: 1011881-1011881 on Build GRCh38
rs777089729 Chr.19: 1012038-1012038 on Build GRCh38
rs370976619 Chr.19: 1011821-1011821 on Build GRCh38
rs780997838 Chr.19: 1012018-1012018 on Build GRCh38
rs761566819 Chr.19: 1011819-1011819 on Build GRCh38
rs185585241 Chr.19: 1012051-1012051 on Build GRCh38
rs764101442 Chr.19: 1012112-1012112 on Build GRCh38
rs1005794782 Chr.19: 1012233-1012233 on Build GRCh38
rs775945585 Chr.19: 1012171-1012171 on Build GRCh38
rs749256164 Chr.19: 1011876-1011876 on Build GRCh38
rs779663084 Chr.19: 1012075-1012075 on Build GRCh38
rs528151720 Chr.19: 1012056-1012056 on Build GRCh38
rs751977137 Chr.19: 1011868-1011868 on Build GRCh38
rs538906148 Chr.19: 1011842-1011842 on Build GRCh38
rs201675498 Chr.19: 1012052-1012052 on Build GRCh38
rs113403992 Chr.19: 1011994-1011994 on Build GRCh38

View Full Product Details