Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1609483-1609975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF3
Entrez Gene ID
6929
Gene Name
transcription factor 3
Gene Aliases
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21
UniGene
Hs.371282
Gene Chromosome Location
Chr.19: 1609290-1652590 on Build GRCh38
Transcripts
NM_001136139.2 NM_003200.3 XM_006722852.3 XM_006722853.3 XM_006722855.3 XM_006722856.3 XM_006722857.3 XM_006722858.3 XM_011528215.2 XM_011528216.2 XM_011528217.2 XM_011528219.2 XM_011528220.2 XM_011528221.2 XM_011528223.2 XM_011528225.2 XM_011528226.2 XM_011528227.2 XM_017027177.1 XM_017027178.1 XM_017027179.1 XM_017027180.1 XM_017027181.1 XM_017027182.1 XM_017027183.1

SNP Detail

SNP ID Chromosome Location
rs892701152 Chr.19: 1609618-1609618 on Build GRCh38
rs773633934 Chr.19: 1609703-1609703 on Build GRCh38
rs914028193 Chr.19: 1609849-1609849 on Build GRCh38
rs143589450 Chr.19: 1609668-1609668 on Build GRCh38
rs550821675 Chr.19: 1609734-1609734 on Build GRCh38
rs934843661 Chr.19: 1609674-1609674 on Build GRCh38
rs965144170 Chr.19: 1609786-1609786 on Build GRCh38
rs557406314 Chr.19: 1609819-1609819 on Build GRCh38
rs752088590 Chr.19: 1609953-1609953 on Build GRCh38
rs978388820 Chr.19: 1609816-1609816 on Build GRCh38
rs767530206 Chr.19: 1609629-1609629 on Build GRCh38
rs774223768 Chr.19: 1609535-1609535 on Build GRCh38
rs759550545 Chr.19: 1609530-1609532 on Build GRCh38
rs1046655762 Chr.19: 1609745-1609745 on Build GRCh38
rs899184126 Chr.19: 1609945-1609945 on Build GRCh38
rs956168281 Chr.19: 1609818-1609819 on Build GRCh38
rs188025273 Chr.19: 1609525-1609525 on Build GRCh38
rs929353696 Chr.19: 1609954-1609954 on Build GRCh38
rs879844727 Chr.19: 1609701-1609701 on Build GRCh38
rs896916384 Chr.19: 1609927-1609927 on Build GRCh38
rs542863373 Chr.19: 1609578-1609578 on Build GRCh38
rs905604318 Chr.19: 1609743-1609743 on Build GRCh38
rs964639692 Chr.19: 1609795-1609795 on Build GRCh38
rs982320333 Chr.19: 1609877-1609877 on Build GRCh38
rs536404733 Chr.19: 1609759-1609759 on Build GRCh38
rs527644879 Chr.19: 1609634-1609634 on Build GRCh38
rs905159028 Chr.19: 1609750-1609750 on Build GRCh38
rs988946227 Chr.19: 1609842-1609842 on Build GRCh38
rs1009198269 Chr.19: 1609768-1609768 on Build GRCh38
rs914095428 Chr.19: 1609731-1609731 on Build GRCh38
rs899236375 Chr.19: 1609760-1609760 on Build GRCh38
rs1048300396 Chr.19: 1609936-1609936 on Build GRCh38
rs770886833 Chr.19: 1609526-1609526 on Build GRCh38
rs1016081878 Chr.19: 1609523-1609523 on Build GRCh38
rs905212039 Chr.19: 1609631-1609631 on Build GRCh38
rs72618599 Chr.19: 1609761-1609761 on Build GRCh38
rs1026543811 Chr.19: 1609762-1609762 on Build GRCh38
rs987530027 Chr.19: 1609542-1609542 on Build GRCh38
rs41275834 Chr.19: 1609617-1609617 on Build GRCh38
rs112793810 Chr.19: 1609673-1609673 on Build GRCh38
rs1016364342 Chr.19: 1609636-1609636 on Build GRCh38
rs919166605 Chr.19: 1609527-1609527 on Build GRCh38
rs1023096381 Chr.19: 1609560-1609560 on Build GRCh38
rs987425328 Chr.19: 1609684-1609684 on Build GRCh38
rs545052874 Chr.19: 1609554-1609554 on Build GRCh38
rs902091591 Chr.19: 1609524-1609524 on Build GRCh38
rs1032667228 Chr.19: 1609821-1609821 on Build GRCh38
rs949644567 Chr.19: 1609744-1609744 on Build GRCh38
rs890310749 Chr.19: 1609767-1609767 on Build GRCh38
rs374868920 Chr.19: 1609843-1609843 on Build GRCh38
rs562682379 Chr.19: 1609515-1609515 on Build GRCh38
rs577683162 Chr.19: 1609823-1609823 on Build GRCh38
rs1040582574 Chr.19: 1609512-1609512 on Build GRCh38
rs575889938 Chr.19: 1609951-1609951 on Build GRCh38
rs532030020 Chr.19: 1609719-1609719 on Build GRCh38
rs181274027 Chr.19: 1609751-1609751 on Build GRCh38
rs1042803266 Chr.19: 1609600-1609600 on Build GRCh38
rs868286785 Chr.19: 1609540-1609540 on Build GRCh38
rs1026870806 Chr.19: 1609763-1609763 on Build GRCh38
rs964357671 Chr.19: 1609702-1609702 on Build GRCh38
rs543241041 Chr.19: 1609952-1609952 on Build GRCh38
rs943506959 Chr.19: 1609670-1609670 on Build GRCh38
rs925422604 Chr.19: 1609605-1609605 on Build GRCh38
rs987366057 Chr.19: 1609787-1609787 on Build GRCh38
rs910191654 Chr.19: 1609695-1609695 on Build GRCh38
rs1037688069 Chr.19: 1609919-1609919 on Build GRCh38
rs993996115 Chr.19: 1609934-1609934 on Build GRCh38
rs76820530 Chr.19: 1609891-1609891 on Build GRCh38
rs538660191 Chr.19: 1609890-1609890 on Build GRCh38
rs575746424 Chr.19: 1609549-1609549 on Build GRCh38
rs1052918 Chr.19: 1609670-1609670 on Build GRCh38
rs1017628936 Chr.19: 1609778-1609778 on Build GRCh38
rs926858824 Chr.19: 1609887-1609887 on Build GRCh38
rs905619938 Chr.19: 1609595-1609595 on Build GRCh38
rs949587536 Chr.19: 1609858-1609858 on Build GRCh38
rs200468115 Chr.19: 1609525-1609525 on Build GRCh38
rs913121582 Chr.19: 1609802-1609802 on Build GRCh38
rs888344170 Chr.19: 1609950-1609950 on Build GRCh38
rs941415402 Chr.19: 1609752-1609782 on Build GRCh38
rs937996388 Chr.19: 1609757-1609757 on Build GRCh38
rs755532654 Chr.19: 1609851-1609851 on Build GRCh38
rs947699754 Chr.19: 1609876-1609876 on Build GRCh38
rs567417124 Chr.19: 1609776-1609776 on Build GRCh38
rs954769503 Chr.19: 1609671-1609671 on Build GRCh38
rs1053018771 Chr.19: 1609729-1609729 on Build GRCh38
rs936846351 Chr.19: 1609609-1609609 on Build GRCh38
rs370154030 Chr.19: 1609818-1609818 on Build GRCh38
rs759447852 Chr.19: 1609574-1609574 on Build GRCh38
rs549439126 Chr.19: 1609669-1609669 on Build GRCh38
rs976165100 Chr.19: 1609791-1609792 on Build GRCh38
rs951989380 Chr.19: 1609640-1609640 on Build GRCh38
rs1056865703 Chr.19: 1609926-1609926 on Build GRCh38
rs577934428 Chr.19: 1609879-1609879 on Build GRCh38
rs1046809046 Chr.19: 1609630-1609630 on Build GRCh38
rs763374680 Chr.19: 1609718-1609718 on Build GRCh38
rs531853135 Chr.19: 1609732-1609732 on Build GRCh38

View Full Product Details