Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2110615-2111118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3D1
Entrez Gene ID
8943
Gene Name
adaptor related protein complex 3 delta 1 subunit
Gene Aliases
ADTD, hBLVR
UniGene
Hs.512815
Gene Chromosome Location
Chr.19: 2100988-2151620 on Build GRCh38
Transcripts
NM_001261826.1 NM_003938.6 XM_006722932.2 XM_017027422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449220 c.2923A>G p.R975G Chr.19: 2110773-2110773 on Build GRCh38
3404005 c.3154G>A p.V1052M Chr.19: 2110728-2110728 on Build GRCh38
5449221 c.2800A>G p.R934G Chr.19: 2110773-2110773 on Build GRCh38
4606431 c.2739G>C p.E913D Chr.19: 2110834-2110834 on Build GRCh38
4783757 c.3145G>A p.G1049S Chr.19: 2110737-2110737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771195476 Chr.19: 2110936-2110936 on Build GRCh38
rs749324068 Chr.19: 2110935-2110935 on Build GRCh38
rs1026438538 Chr.19: 2110649-2110649 on Build GRCh38
rs748298616 Chr.19: 2110828-2110828 on Build GRCh38
rs367890742 Chr.19: 2110900-2110900 on Build GRCh38
rs551959777 Chr.19: 2110808-2110808 on Build GRCh38
rs572208167 Chr.19: 2111023-2111023 on Build GRCh38
rs1044196267 Chr.19: 2111005-2111005 on Build GRCh38
rs751860280 Chr.19: 2110774-2110774 on Build GRCh38
rs371993830 Chr.19: 2110870-2110870 on Build GRCh38
rs573454363 Chr.19: 2110736-2110736 on Build GRCh38
rs776803135 Chr.19: 2110751-2110751 on Build GRCh38
rs73512347 Chr.19: 2111013-2111013 on Build GRCh38
rs753440657 Chr.19: 2110794-2110794 on Build GRCh38
rs139281900 Chr.19: 2110883-2110883 on Build GRCh38
rs201069215 Chr.19: 2110764-2110764 on Build GRCh38
rs75337979 Chr.19: 2111064-2111064 on Build GRCh38
rs200189042 Chr.19: 2110795-2110795 on Build GRCh38
rs755674952 Chr.19: 2110734-2110734 on Build GRCh38
rs764258279 Chr.19: 2110672-2110672 on Build GRCh38
rs1018246074 Chr.19: 2111050-2111050 on Build GRCh38
rs200937321 Chr.19: 2110896-2110896 on Build GRCh38
rs20567 Chr.19: 2110747-2110747 on Build GRCh38
rs979386577 Chr.19: 2111068-2111068 on Build GRCh38
rs772188359 Chr.19: 2110662-2110662 on Build GRCh38
rs772676874 Chr.19: 2110691-2110692 on Build GRCh38
rs77605654 Chr.19: 2111039-2111039 on Build GRCh38
rs564181671 Chr.19: 2110810-2110810 on Build GRCh38
rs775540607 Chr.19: 2110669-2110669 on Build GRCh38
rs558977794 Chr.19: 2110741-2110741 on Build GRCh38
rs947139375 Chr.19: 2111004-2111004 on Build GRCh38
rs774723231 Chr.19: 2110860-2110860 on Build GRCh38
rs201197577 Chr.19: 2110760-2110760 on Build GRCh38
rs375258290 Chr.19: 2110697-2110697 on Build GRCh38
rs535472544 Chr.19: 2110907-2110907 on Build GRCh38
rs780575228 Chr.19: 2110728-2110728 on Build GRCh38
rs372877711 Chr.19: 2110687-2110687 on Build GRCh38
rs559127027 Chr.19: 2110955-2110955 on Build GRCh38
rs774528663 Chr.19: 2110946-2110946 on Build GRCh38
rs778410062 Chr.19: 2110737-2110737 on Build GRCh38
rs767204653 Chr.19: 2110663-2110663 on Build GRCh38
rs79288884 Chr.19: 2111036-2111036 on Build GRCh38
rs4806830 Chr.19: 2110842-2110842 on Build GRCh38
rs1030629281 Chr.19: 2111073-2111073 on Build GRCh38
rs528887337 Chr.19: 2111046-2111046 on Build GRCh38
rs752054956 Chr.19: 2110729-2110729 on Build GRCh38
rs746761305 Chr.19: 2110811-2110811 on Build GRCh38
rs541896236 Chr.19: 2110773-2110773 on Build GRCh38
rs571915943 Chr.19: 2110725-2110725 on Build GRCh38
rs534349795 Chr.19: 2110930-2110930 on Build GRCh38
rs968977581 Chr.19: 2111085-2111085 on Build GRCh38
rs1012361793 Chr.19: 2110694-2110694 on Build GRCh38
rs1043548686 Chr.19: 2110638-2110638 on Build GRCh38
rs911666720 Chr.19: 2111044-2111058 on Build GRCh38
rs368781853 Chr.19: 2110913-2110913 on Build GRCh38
rs201595321 Chr.19: 2110724-2110724 on Build GRCh38
rs367631461 Chr.19: 2110746-2110746 on Build GRCh38
rs766517518 Chr.19: 2110857-2110857 on Build GRCh38
rs201737398 Chr.19: 2110845-2110845 on Build GRCh38
rs115359236 Chr.19: 2110990-2110990 on Build GRCh38
rs142622935 Chr.19: 2110975-2110976 on Build GRCh38
rs758594290 Chr.19: 2110788-2110788 on Build GRCh38
rs200108136 Chr.19: 2110740-2110740 on Build GRCh38
rs771585825 Chr.19: 2110687-2110688 on Build GRCh38
rs751570038 Chr.19: 2110822-2110822 on Build GRCh38
rs560258409 Chr.19: 2110791-2110791 on Build GRCh38
rs753803636 Chr.19: 2110679-2110679 on Build GRCh38
rs1021856415 Chr.19: 2111081-2111081 on Build GRCh38
rs565822197 Chr.19: 2110929-2110929 on Build GRCh38
rs768660885 Chr.19: 2110816-2110816 on Build GRCh38
rs371989768 Chr.19: 2110903-2110903 on Build GRCh38
rs544252608 Chr.19: 2110759-2110759 on Build GRCh38
rs955986470 Chr.19: 2111095-2111095 on Build GRCh38
rs760906099 Chr.19: 2110670-2110670 on Build GRCh38
rs745503252 Chr.19: 2110800-2110800 on Build GRCh38
rs545653461 Chr.19: 2111034-2111034 on Build GRCh38
rs755477169 Chr.19: 2110789-2110789 on Build GRCh38
rs960766488 Chr.19: 2110647-2110647 on Build GRCh38
rs911186794 Chr.19: 2110829-2110829 on Build GRCh38
rs753143663 Chr.19: 2110884-2110884 on Build GRCh38
rs768412125 Chr.19: 2110748-2110748 on Build GRCh38
rs565733361 Chr.19: 2111077-2111077 on Build GRCh38
rs896222462 Chr.19: 2110686-2110686 on Build GRCh38
rs373655875 Chr.19: 2110735-2110735 on Build GRCh38
rs902324792 Chr.19: 2110648-2110648 on Build GRCh38
rs773479033 Chr.19: 2110843-2110843 on Build GRCh38
rs556606645 Chr.19: 2110997-2110997 on Build GRCh38
rs574486632 Chr.19: 2111010-2111010 on Build GRCh38
rs747716229 Chr.19: 2110667-2110668 on Build GRCh38
rs201248095 Chr.19: 2110909-2110909 on Build GRCh38
rs914268057 Chr.19: 2111000-2111000 on Build GRCh38
rs994782041 Chr.19: 2111024-2111024 on Build GRCh38
rs577430414 Chr.19: 2110983-2110983 on Build GRCh38
rs750821648 Chr.19: 2110707-2110707 on Build GRCh38
rs561882028 Chr.19: 2111045-2111045 on Build GRCh38
rs749888438 Chr.19: 2110888-2110888 on Build GRCh38
rs961515446 Chr.19: 2110762-2110762 on Build GRCh38
rs1056638213 Chr.19: 2110671-2110671 on Build GRCh38
rs560170643 Chr.19: 2111014-2111014 on Build GRCh38
rs768113479 Chr.19: 2110871-2110871 on Build GRCh38
rs377111065 Chr.19: 2110714-2110714 on Build GRCh38
rs896475933 Chr.19: 2110895-2110895 on Build GRCh38
rs574773348 Chr.19: 2110771-2110771 on Build GRCh38
rs756850780 Chr.19: 2110885-2110885 on Build GRCh38
rs759360356 Chr.19: 2110661-2110661 on Build GRCh38
rs748019491 Chr.19: 2110925-2110925 on Build GRCh38
rs1045139467 Chr.19: 2110972-2110972 on Build GRCh38
rs192625683 Chr.19: 2111038-2111038 on Build GRCh38
rs753610242 Chr.19: 2110763-2110763 on Build GRCh38

View Full Product Details