Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2272614-2273105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAZ1
Entrez Gene ID
4946
Gene Name
ornithine decarboxylase antizyme 1
Gene Aliases
AZI, OAZ
UniGene
Hs.446427
Gene Chromosome Location
Chr.19: 2269486-2273488 on Build GRCh38
Transcripts
NM_001301020.1 NM_004152.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095412 c.652G>A p.E218K Chr.19: 2273076-2273076 on Build GRCh38
5516802 c.652_655delGAGA p.R219fs*>8 Chr.19: 2273076-2273079 on Build GRCh38
7078850 c.604C>T p.P202S Chr.19: 2273028-2273028 on Build GRCh38
6732105 c.636C>A p.F212L Chr.19: 2273060-2273060 on Build GRCh38
6587401 c.505G>A p.D169N Chr.19: 2272778-2272778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752639411 Chr.19: 2273043-2273043 on Build GRCh38
rs998387767 Chr.19: 2272861-2272861 on Build GRCh38
rs761083193 Chr.19: 2272961-2272961 on Build GRCh38
rs998679187 Chr.19: 2272851-2272851 on Build GRCh38
rs746850721 Chr.19: 2272923-2272923 on Build GRCh38
rs564384269 Chr.19: 2272974-2272974 on Build GRCh38
rs993846802 Chr.19: 2272958-2272958 on Build GRCh38
rs770289416 Chr.19: 2272932-2272933 on Build GRCh38
rs574613106 Chr.19: 2272873-2272873 on Build GRCh38
rs750540706 Chr.19: 2272709-2272709 on Build GRCh38
rs919301388 Chr.19: 2272649-2272649 on Build GRCh38
rs777917096 Chr.19: 2273046-2273046 on Build GRCh38
rs764372147 Chr.19: 2273016-2273016 on Build GRCh38
rs749092327 Chr.19: 2273048-2273048 on Build GRCh38
rs749835378 Chr.19: 2273075-2273076 on Build GRCh38
rs910554501 Chr.19: 2272901-2272901 on Build GRCh38
rs760094093 Chr.19: 2272844-2272844 on Build GRCh38
rs774587546 Chr.19: 2272946-2272946 on Build GRCh38
rs761819044 Chr.19: 2272725-2272725 on Build GRCh38
rs373508776 Chr.19: 2273026-2273026 on Build GRCh38
rs771614583 Chr.19: 2272782-2272782 on Build GRCh38
rs112906547 Chr.19: 2272688-2272688 on Build GRCh38
rs929888316 Chr.19: 2272915-2272915 on Build GRCh38
rs960860113 Chr.19: 2272786-2272786 on Build GRCh38
rs896930385 Chr.19: 2272953-2272953 on Build GRCh38
rs757159726 Chr.19: 2273051-2273051 on Build GRCh38
rs768291790 Chr.19: 2272803-2272803 on Build GRCh38
rs771266572 Chr.19: 2272988-2272988 on Build GRCh38
rs895781765 Chr.19: 2272657-2272657 on Build GRCh38
rs772095687 Chr.19: 2272997-2272997 on Build GRCh38
rs750128990 Chr.19: 2272857-2272857 on Build GRCh38
rs768648832 Chr.19: 2272926-2272926 on Build GRCh38
rs947133624 Chr.19: 2272768-2272768 on Build GRCh38
rs949189822 Chr.19: 2272962-2272962 on Build GRCh38
rs748084896 Chr.19: 2272975-2272975 on Build GRCh38
rs1019919662 Chr.19: 2272890-2272890 on Build GRCh38
rs28384684 Chr.19: 2272932-2272932 on Build GRCh38
rs754511396 Chr.19: 2272973-2272973 on Build GRCh38
rs375387391 Chr.19: 2272918-2272918 on Build GRCh38
rs772155239 Chr.19: 2273065-2273065 on Build GRCh38
rs761881099 Chr.19: 2272820-2272820 on Build GRCh38
rs972324428 Chr.19: 2272789-2272789 on Build GRCh38
rs375993479 Chr.19: 2272777-2272777 on Build GRCh38
rs531300010 Chr.19: 2273044-2273044 on Build GRCh38
rs781643247 Chr.19: 2272758-2272758 on Build GRCh38
rs367569769 Chr.19: 2272727-2272727 on Build GRCh38
rs1039801243 Chr.19: 2272963-2272963 on Build GRCh38
rs749464270 Chr.19: 2272987-2272987 on Build GRCh38
rs28384683 Chr.19: 2272904-2272904 on Build GRCh38
rs759142199 Chr.19: 2273076-2273076 on Build GRCh38
rs776665304 Chr.19: 2272683-2272683 on Build GRCh38
rs1044350197 Chr.19: 2273070-2273070 on Build GRCh38
rs567037358 Chr.19: 2272799-2272799 on Build GRCh38
rs777186507 Chr.19: 2273071-2273071 on Build GRCh38
rs751208991 Chr.19: 2273033-2273033 on Build GRCh38
rs1022637215 Chr.19: 2272869-2272869 on Build GRCh38
rs534549628 Chr.19: 2272809-2272809 on Build GRCh38
rs1008031546 Chr.19: 2272686-2272687 on Build GRCh38
rs917137560 Chr.19: 2272912-2272912 on Build GRCh38
rs1057150 Chr.19: 2273080-2273080 on Build GRCh38
rs970090308 Chr.19: 2272871-2272871 on Build GRCh38
rs549863494 Chr.19: 2272692-2272692 on Build GRCh38
rs888466461 Chr.19: 2272853-2272853 on Build GRCh38
rs771687568 Chr.19: 2273078-2273085 on Build GRCh38
rs372096490 Chr.19: 2272830-2272830 on Build GRCh38
rs576779461 Chr.19: 2272854-2272854 on Build GRCh38
rs756504917 Chr.19: 2272855-2272855 on Build GRCh38
rs1029906151 Chr.19: 2272874-2272874 on Build GRCh38
rs776102704 Chr.19: 2272955-2272956 on Build GRCh38
rs771068252 Chr.19: 2272945-2272945 on Build GRCh38
rs755924220 Chr.19: 2272976-2272976 on Build GRCh38
rs376446506 Chr.19: 2272872-2272872 on Build GRCh38
rs960890878 Chr.19: 2272879-2272879 on Build GRCh38
rs779176036 Chr.19: 2273052-2273052 on Build GRCh38
rs756948907 Chr.19: 2272762-2272762 on Build GRCh38
rs370045809 Chr.19: 2273072-2273072 on Build GRCh38
rs937168376 Chr.19: 2272902-2272902 on Build GRCh38
rs746103860 Chr.19: 2273056-2273056 on Build GRCh38
rs910512783 Chr.19: 2272813-2272813 on Build GRCh38
rs754487159 Chr.19: 2273039-2273039 on Build GRCh38
rs755275396 Chr.19: 2272748-2272748 on Build GRCh38
rs957060411 Chr.19: 2272747-2272747 on Build GRCh38
rs774005199 Chr.19: 2273075-2273075 on Build GRCh38
rs767718584 Chr.19: 2272852-2272852 on Build GRCh38
rs376496505 Chr.19: 2273067-2273067 on Build GRCh38
rs758047877 Chr.19: 2272967-2272967 on Build GRCh38
rs1026143515 Chr.19: 2272675-2272675 on Build GRCh38
rs778597507 Chr.19: 2272776-2272776 on Build GRCh38
rs976197326 Chr.19: 2272907-2272907 on Build GRCh38
rs777715067 Chr.19: 2272979-2272979 on Build GRCh38
rs968798613 Chr.19: 2272895-2272895 on Build GRCh38
rs968409965 Chr.19: 2273040-2273040 on Build GRCh38
rs762220087 Chr.19: 2273021-2273021 on Build GRCh38
rs1018265002 Chr.19: 2272980-2272980 on Build GRCh38
rs761122600 Chr.19: 2273010-2273010 on Build GRCh38
rs183701220 Chr.19: 2272856-2272856 on Build GRCh38
rs745931817 Chr.19: 2272995-2272995 on Build GRCh38
rs776990263 Chr.19: 2273020-2273020 on Build GRCh38
rs947202532 Chr.19: 2273069-2273069 on Build GRCh38
rs988817565 Chr.19: 2272888-2272888 on Build GRCh38
rs112007698 Chr.19: 2272969-2272969 on Build GRCh38
rs556203566 Chr.19: 2272859-2272859 on Build GRCh38
rs934504455 Chr.19: 2272914-2272914 on Build GRCh38
rs978442820 Chr.19: 2272896-2272896 on Build GRCh38
rs536418152 Chr.19: 2272712-2272712 on Build GRCh38
rs752073808 Chr.19: 2272842-2272842 on Build GRCh38
rs927086414 Chr.19: 2272897-2272897 on Build GRCh38
rs776020059 Chr.19: 2273003-2273003 on Build GRCh38
rs766615001 Chr.19: 2272836-2272836 on Build GRCh38
rs191605637 Chr.19: 2272721-2272721 on Build GRCh38
rs763093721 Chr.19: 2272832-2272832 on Build GRCh38
rs371945440 Chr.19: 2272940-2272940 on Build GRCh38
rs769515860 Chr.19: 2272937-2272937 on Build GRCh38
rs781046261 Chr.19: 2272929-2272929 on Build GRCh38
rs551868965 Chr.19: 2272662-2272662 on Build GRCh38
rs541636712 Chr.19: 2272878-2272878 on Build GRCh38
rs200109760 Chr.19: 2272991-2272991 on Build GRCh38
rs564119554 Chr.19: 2272677-2272677 on Build GRCh38
rs779566415 Chr.19: 2272865-2272865 on Build GRCh38
rs775431989 Chr.19: 2273066-2273066 on Build GRCh38
rs768066242 Chr.19: 2272952-2272952 on Build GRCh38
rs753188317 Chr.19: 2272955-2272955 on Build GRCh38
rs779949107 Chr.19: 2272790-2272790 on Build GRCh38
rs766987818 Chr.19: 2272713-2272713 on Build GRCh38
rs201200204 Chr.19: 2273083-2273083 on Build GRCh38
rs568178484 Chr.19: 2272695-2272695 on Build GRCh38
rs1040441492 Chr.19: 2272832-2272833 on Build GRCh38
rs374764733 Chr.19: 2272968-2272968 on Build GRCh38
rs752135849 Chr.19: 2273081-2273081 on Build GRCh38
rs764468507 Chr.19: 2272964-2272964 on Build GRCh38
rs759594251 Chr.19: 2272951-2272951 on Build GRCh38
rs201632586 Chr.19: 2272759-2272759 on Build GRCh38
rs765160857 Chr.19: 2272691-2272691 on Build GRCh38
rs768490699 Chr.19: 2272866-2272866 on Build GRCh38
rs368465089 Chr.19: 2272823-2272823 on Build GRCh38
rs954517561 Chr.19: 2272724-2272724 on Build GRCh38
rs969846397 Chr.19: 2272773-2272773 on Build GRCh38
rs901716042 Chr.19: 2272841-2272841 on Build GRCh38
rs763290443 Chr.19: 2272941-2272941 on Build GRCh38

View Full Product Details