Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3056176-3056675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AES
Entrez Gene ID
166
Gene Name
amino-terminal enhancer of split
Gene Aliases
AES-1, AES-2, ESP1, GRG, GRG5, Grg-5, TLE5
UniGene
Hs.515053
Gene Chromosome Location
Chr.19: 3052910-3063107 on Build GRCh38
Transcripts
NM_001130.5 NM_198969.1 NM_198970.1 XM_006722664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4975821 c.396C>G p.Y132* Chr.19: 3056351-3056351 on Build GRCh38
6207660 c.435+9A>C p.? Chr.19: 3056303-3056303 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375769156 Chr.19: 3056382-3056382 on Build GRCh38
rs747975447 Chr.19: 3056262-3056262 on Build GRCh38
rs973802595 Chr.19: 3056523-3056524 on Build GRCh38
rs1037709795 Chr.19: 3056219-3056219 on Build GRCh38
rs769652060 Chr.19: 3056269-3056269 on Build GRCh38
rs967694098 Chr.19: 3056432-3056432 on Build GRCh38
rs762197057 Chr.19: 3056485-3056485 on Build GRCh38
rs578094319 Chr.19: 3056586-3056586 on Build GRCh38
rs748335954 Chr.19: 3056655-3056655 on Build GRCh38
rs764507020 Chr.19: 3056299-3056299 on Build GRCh38
rs773828348 Chr.19: 3056275-3056276 on Build GRCh38
rs1046659610 Chr.19: 3056545-3056545 on Build GRCh38
rs946165168 Chr.19: 3056405-3056405 on Build GRCh38
rs556096421 Chr.19: 3056409-3056409 on Build GRCh38
rs912993530 Chr.19: 3056617-3056617 on Build GRCh38
rs756160679 Chr.19: 3056644-3056644 on Build GRCh38
rs1002565436 Chr.19: 3056289-3056289 on Build GRCh38
rs3888377 Chr.19: 3056215-3056215 on Build GRCh38
rs773139865 Chr.19: 3056276-3056276 on Build GRCh38
rs776896680 Chr.19: 3056469-3056469 on Build GRCh38
rs754214877 Chr.19: 3056307-3056307 on Build GRCh38
rs564114196 Chr.19: 3056431-3056431 on Build GRCh38
rs954496592 Chr.19: 3056454-3056454 on Build GRCh38
rs781361278 Chr.19: 3056273-3056281 on Build GRCh38
rs528278821 Chr.19: 3056277-3056277 on Build GRCh38
rs767196410 Chr.19: 3056589-3056589 on Build GRCh38
rs954612201 Chr.19: 3056654-3056654 on Build GRCh38
rs921920885 Chr.19: 3056256-3056256 on Build GRCh38
rs1025698273 Chr.19: 3056503-3056503 on Build GRCh38
rs376096556 Chr.19: 3056245-3056245 on Build GRCh38
rs774586861 Chr.19: 3056408-3056408 on Build GRCh38
rs919303566 Chr.19: 3056537-3056537 on Build GRCh38
rs775541123 Chr.19: 3056423-3056423 on Build GRCh38
rs769145666 Chr.19: 3056405-3056406 on Build GRCh38
rs568909948 Chr.19: 3056373-3056373 on Build GRCh38
rs545515726 Chr.19: 3056196-3056196 on Build GRCh38
rs756419198 Chr.19: 3056272-3056273 on Build GRCh38
rs58280427 Chr.19: 3056287-3056287 on Build GRCh38
rs979379455 Chr.19: 3056639-3056639 on Build GRCh38
rs772076353 Chr.19: 3056415-3056415 on Build GRCh38
rs759620683 Chr.19: 3056290-3056290 on Build GRCh38
rs775919452 Chr.19: 3056297-3056297 on Build GRCh38
rs780531836 Chr.19: 3056273-3056278 on Build GRCh38
rs761034112 Chr.19: 3056298-3056298 on Build GRCh38
rs746473109 Chr.19: 3056598-3056598 on Build GRCh38
rs988444777 Chr.19: 3056609-3056609 on Build GRCh38
rs998814665 Chr.19: 3056450-3056450 on Build GRCh38
rs765969023 Chr.19: 3056553-3056553 on Build GRCh38
rs557025863 Chr.19: 3056600-3056600 on Build GRCh38
rs781290830 Chr.19: 3056368-3056368 on Build GRCh38
rs933275501 Chr.19: 3056263-3056263 on Build GRCh38
rs546748288 Chr.19: 3056279-3056279 on Build GRCh38
rs927475607 Chr.19: 3056640-3056640 on Build GRCh38
rs929542100 Chr.19: 3056541-3056541 on Build GRCh38
rs895802961 Chr.19: 3056448-3056448 on Build GRCh38
rs986241230 Chr.19: 3056198-3056198 on Build GRCh38
rs369218360 Chr.19: 3056552-3056552 on Build GRCh38
rs758032593 Chr.19: 3056308-3056308 on Build GRCh38
rs766023444 Chr.19: 3056312-3056312 on Build GRCh38
rs947117175 Chr.19: 3056634-3056634 on Build GRCh38
rs771093965 Chr.19: 3056402-3056402 on Build GRCh38
rs534920862 Chr.19: 3056451-3056451 on Build GRCh38
rs771166049 Chr.19: 3056286-3056286 on Build GRCh38
rs142071716 Chr.19: 3056257-3056257 on Build GRCh38
rs754538819 Chr.19: 3056576-3056576 on Build GRCh38
rs372540435 Chr.19: 3056296-3056296 on Build GRCh38
rs749617428 Chr.19: 3056367-3056368 on Build GRCh38
rs763841764 Chr.19: 3056273-3056275 on Build GRCh38
rs751165254 Chr.19: 3056566-3056566 on Build GRCh38
rs180907880 Chr.19: 3056339-3056339 on Build GRCh38
rs538795279 Chr.19: 3056587-3056587 on Build GRCh38
rs536211461 Chr.19: 3056396-3056396 on Build GRCh38
rs777478977 Chr.19: 3056395-3056395 on Build GRCh38
rs1011620394 Chr.19: 3056429-3056429 on Build GRCh38
rs542553369 Chr.19: 3056244-3056244 on Build GRCh38
rs530811499 Chr.19: 3056218-3056218 on Build GRCh38
rs553564174 Chr.19: 3056455-3056455 on Build GRCh38
rs1057088959 Chr.19: 3056328-3056328 on Build GRCh38
rs577233984 Chr.19: 3056529-3056529 on Build GRCh38
rs1043269010 Chr.19: 3056505-3056505 on Build GRCh38
rs779581339 Chr.19: 3056454-3056454 on Build GRCh38
rs751061760 Chr.19: 3056333-3056333 on Build GRCh38

View Full Product Details