Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3589697-3590194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC3
Entrez Gene ID
126326
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
C19orf64, DFNB15, DFNB72, DFNB95
UniGene
Hs.266873
Gene Chromosome Location
Chr.19: 3585531-3593541 on Build GRCh38
Transcripts
NM_133261.2 XM_005259492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725848 c.779C>T p.P260L Chr.19: 3589904-3589904 on Build GRCh38
3532538 c.831G>A p.Q277Q Chr.19: 3590082-3590082 on Build GRCh38
3532539 c.855C>T p.S285S Chr.19: 3590106-3590106 on Build GRCh38
5594628 c.787+2T>A p.? Chr.19: 3589914-3589914 on Build GRCh38
7205831 c.856G>A p.V286I Chr.19: 3590107-3590107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138339125 Chr.19: 3590107-3590107 on Build GRCh38
rs1014244209 Chr.19: 3589890-3589890 on Build GRCh38
rs746265287 Chr.19: 3590078-3590078 on Build GRCh38
rs371622800 Chr.19: 3590004-3590004 on Build GRCh38
rs755288298 Chr.19: 3590064-3590064 on Build GRCh38
rs976428102 Chr.19: 3589753-3589753 on Build GRCh38
rs141419242 Chr.19: 3590058-3590058 on Build GRCh38
rs763402532 Chr.19: 3589995-3589995 on Build GRCh38
rs761135287 Chr.19: 3589825-3589825 on Build GRCh38
rs149647721 Chr.19: 3590116-3590116 on Build GRCh38
rs764287192 Chr.19: 3590121-3590121 on Build GRCh38
rs368529519 Chr.19: 3589723-3589723 on Build GRCh38
rs570485115 Chr.19: 3589906-3589906 on Build GRCh38
rs374114426 Chr.19: 3589944-3589944 on Build GRCh38
rs747730761 Chr.19: 3590092-3590092 on Build GRCh38
rs777097815 Chr.19: 3589942-3589943 on Build GRCh38
rs766192644 Chr.19: 3590106-3590106 on Build GRCh38
rs377024386 Chr.19: 3589993-3589993 on Build GRCh38
rs763654385 Chr.19: 3589999-3589999 on Build GRCh38
rs367940867 Chr.19: 3590119-3590119 on Build GRCh38
rs536060024 Chr.19: 3589755-3589755 on Build GRCh38
rs368101606 Chr.19: 3589777-3589777 on Build GRCh38
rs751286562 Chr.19: 3590061-3590061 on Build GRCh38
rs772816799 Chr.19: 3590159-3590159 on Build GRCh38
rs746316685 Chr.19: 3590133-3590133 on Build GRCh38
rs376993020 Chr.19: 3589946-3589946 on Build GRCh38
rs778730345 Chr.19: 3589950-3589950 on Build GRCh38
rs752914293 Chr.19: 3590071-3590071 on Build GRCh38
rs758088199 Chr.19: 3589956-3589956 on Build GRCh38
rs779309926 Chr.19: 3590131-3590131 on Build GRCh38
rs773206289 Chr.19: 3590046-3590046 on Build GRCh38
rs142749099 Chr.19: 3590166-3590166 on Build GRCh38
rs370361498 Chr.19: 3589780-3589780 on Build GRCh38
rs748204063 Chr.19: 3590013-3590013 on Build GRCh38
rs368548938 Chr.19: 3590029-3590029 on Build GRCh38
rs149028750 Chr.19: 3590072-3590072 on Build GRCh38
rs561617762 Chr.19: 3589960-3589960 on Build GRCh38
rs756062198 Chr.19: 3589786-3589786 on Build GRCh38
rs150473379 Chr.19: 3590070-3590070 on Build GRCh38
rs775216343 Chr.19: 3589997-3589997 on Build GRCh38
rs756042355 Chr.19: 3589859-3589859 on Build GRCh38
rs774571972 Chr.19: 3589818-3589818 on Build GRCh38
rs199951984 Chr.19: 3590122-3590122 on Build GRCh38
rs746137482 Chr.19: 3589882-3589882 on Build GRCh38
rs769307495 Chr.19: 3590158-3590158 on Build GRCh38
rs372890939 Chr.19: 3589822-3589822 on Build GRCh38
rs765760364 Chr.19: 3589921-3589921 on Build GRCh38
rs114801367 Chr.19: 3589955-3589955 on Build GRCh38
rs999714618 Chr.19: 3589765-3589765 on Build GRCh38
rs780392905 Chr.19: 3590136-3590136 on Build GRCh38
rs200909888 Chr.19: 3589856-3589856 on Build GRCh38
rs78077103 Chr.19: 3589943-3589943 on Build GRCh38
rs764630987 Chr.19: 3589826-3589826 on Build GRCh38
rs541403018 Chr.19: 3589992-3589992 on Build GRCh38
rs387907000 Chr.19: 3590154-3590154 on Build GRCh38
rs772581175 Chr.19: 3590094-3590094 on Build GRCh38
rs371599478 Chr.19: 3590093-3590093 on Build GRCh38
rs755235018 Chr.19: 3590008-3590008 on Build GRCh38
rs751260364 Chr.19: 3589854-3589854 on Build GRCh38
rs868241983 Chr.19: 3589852-3589852 on Build GRCh38
rs778677360 Chr.19: 3589878-3589878 on Build GRCh38
rs535252941 Chr.19: 3590156-3590156 on Build GRCh38
rs773696215 Chr.19: 3589938-3589938 on Build GRCh38
rs772055400 Chr.19: 3590167-3590167 on Build GRCh38
rs894024107 Chr.19: 3589974-3589974 on Build GRCh38
rs777588962 Chr.19: 3589787-3589787 on Build GRCh38
rs760318465 Chr.19: 3589998-3589998 on Build GRCh38
rs757685416 Chr.19: 3590076-3590076 on Build GRCh38
rs778361350 Chr.19: 3590019-3590019 on Build GRCh38
rs763530294 Chr.19: 3589945-3589945 on Build GRCh38
rs549625647 Chr.19: 3590007-3590007 on Build GRCh38
rs781415653 Chr.19: 3589989-3589989 on Build GRCh38
rs28532669 Chr.19: 3589770-3589770 on Build GRCh38
rs199684070 Chr.19: 3590002-3590002 on Build GRCh38
rs201853780 Chr.19: 3590134-3590134 on Build GRCh38
rs757053567 Chr.19: 3589864-3589864 on Build GRCh38
rs549658028 Chr.19: 3589764-3589764 on Build GRCh38
rs1045788097 Chr.19: 3589908-3589908 on Build GRCh38
rs759423626 Chr.19: 3590115-3590115 on Build GRCh38
rs372583605 Chr.19: 3590035-3590035 on Build GRCh38
rs200925508 Chr.19: 3589823-3589824 on Build GRCh38
rs764592721 Chr.19: 3590040-3590040 on Build GRCh38
rs762994379 Chr.19: 3590099-3590099 on Build GRCh38
rs377299384 Chr.19: 3590006-3590006 on Build GRCh38
rs759985843 Chr.19: 3589943-3589943 on Build GRCh38
rs749169212 Chr.19: 3589862-3589862 on Build GRCh38
rs759156541 Chr.19: 3589940-3589940 on Build GRCh38
rs528822982 Chr.19: 3590034-3590034 on Build GRCh38
rs387906999 Chr.19: 3589910-3589910 on Build GRCh38
rs749117629 Chr.19: 3589792-3589792 on Build GRCh38
rs781468743 Chr.19: 3590010-3590010 on Build GRCh38
rs749866587 Chr.19: 3590073-3590073 on Build GRCh38
rs572039441 Chr.19: 3589982-3589982 on Build GRCh38
rs35385787 Chr.19: 3589996-3589996 on Build GRCh38
rs876657816 Chr.19: 3589917-3589917 on Build GRCh38
rs775627552 Chr.19: 3589824-3589824 on Build GRCh38
rs778310822 Chr.19: 3589762-3589762 on Build GRCh38
rs772316969 Chr.19: 3589897-3589897 on Build GRCh38
rs768860108 Chr.19: 3589963-3589963 on Build GRCh38
rs747142666 Chr.19: 3589961-3589961 on Build GRCh38
rs765640676 Chr.19: 3590023-3590023 on Build GRCh38
rs777289758 Chr.19: 3589905-3589905 on Build GRCh38
rs758225438 Chr.19: 3590005-3590005 on Build GRCh38
rs768807311 Chr.19: 3589901-3589901 on Build GRCh38
rs1045095383 Chr.19: 3589983-3589983 on Build GRCh38
rs140960269 Chr.19: 3590157-3590157 on Build GRCh38
rs727503065 Chr.19: 3590126-3590126 on Build GRCh38
rs762290371 Chr.19: 3589846-3589846 on Build GRCh38
rs774440451 Chr.19: 3590164-3590164 on Build GRCh38
rs748098027 Chr.19: 3589785-3589785 on Build GRCh38
rs779376305 Chr.19: 3590077-3590077 on Build GRCh38
rs977235380 Chr.19: 3590062-3590062 on Build GRCh38
rs761336297 Chr.19: 3590039-3590039 on Build GRCh38
rs771396764 Chr.19: 3590027-3590027 on Build GRCh38
rs776280043 Chr.19: 3590089-3590089 on Build GRCh38
rs777236848 Chr.19: 3589835-3589835 on Build GRCh38
rs769951289 Chr.19: 3590015-3590015 on Build GRCh38
rs747159691 Chr.19: 3589865-3589865 on Build GRCh38
rs371957896 Chr.19: 3589724-3589724 on Build GRCh38
rs775474512 Chr.19: 3589898-3589898 on Build GRCh38
rs370162131 Chr.19: 3589994-3589994 on Build GRCh38
rs112949777 Chr.19: 3589819-3589819 on Build GRCh38
rs771290947 Chr.19: 3589941-3589942 on Build GRCh38
rs971972097 Chr.19: 3589884-3589884 on Build GRCh38
rs748777903 Chr.19: 3590163-3590163 on Build GRCh38
rs200355546 Chr.19: 3589848-3589848 on Build GRCh38
rs201619343 Chr.19: 3590030-3590030 on Build GRCh38
rs770668040 Chr.19: 3589804-3589804 on Build GRCh38
rs34156653 Chr.19: 3590000-3590000 on Build GRCh38
rs747144389 Chr.19: 3589900-3589900 on Build GRCh38
rs766991127 Chr.19: 3589858-3589858 on Build GRCh38
rs762794106 Chr.19: 3590057-3590057 on Build GRCh38
rs776771676 Chr.19: 3589918-3589918 on Build GRCh38
rs906418662 Chr.19: 3589933-3589934 on Build GRCh38
rs772373224 Chr.19: 3590079-3590079 on Build GRCh38
rs387907001 Chr.19: 3589892-3589892 on Build GRCh38
rs749694987 Chr.19: 3590024-3590024 on Build GRCh38
rs763904089 Chr.19: 3589731-3589731 on Build GRCh38
rs776126285 Chr.19: 3590052-3590052 on Build GRCh38
rs777715304 Chr.19: 3589860-3589860 on Build GRCh38

View Full Product Details