Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4322906-4323410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1
Entrez Gene ID
79187
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
GLFND, MIR1
UniGene
Hs.28144
Gene Chromosome Location
Chr.19: 4304594-4323846 on Build GRCh38
Transcripts
NM_024333.2 XM_005259644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446387 c.1162G>A p.A388T Chr.19: 4323108-4323108 on Build GRCh38
3404302 c.1114G>A p.G372S Chr.19: 4323060-4323060 on Build GRCh38
7251825 c.1296C>G p.L432L Chr.19: 4323352-4323352 on Build GRCh38
5687644 c.1126C>A p.R376S Chr.19: 4323072-4323072 on Build GRCh38
5697027 c.1253T>A p.V418E Chr.19: 4323199-4323199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751698287 Chr.19: 4323075-4323075 on Build GRCh38
rs67511110 Chr.19: 4322993-4322993 on Build GRCh38
rs924630650 Chr.19: 4322930-4322930 on Build GRCh38
rs761082727 Chr.19: 4323038-4323038 on Build GRCh38
rs747522449 Chr.19: 4322968-4322969 on Build GRCh38
rs780498419 Chr.19: 4323326-4323326 on Build GRCh38
rs751672012 Chr.19: 4323188-4323188 on Build GRCh38
rs768350624 Chr.19: 4323154-4323154 on Build GRCh38
rs776886218 Chr.19: 4323376-4323376 on Build GRCh38
rs116268486 Chr.19: 4323260-4323260 on Build GRCh38
rs140560499 Chr.19: 4323005-4323005 on Build GRCh38
rs1017320504 Chr.19: 4323271-4323271 on Build GRCh38
rs773984214 Chr.19: 4323014-4323014 on Build GRCh38
rs780033085 Chr.19: 4323223-4323223 on Build GRCh38
rs200289353 Chr.19: 4323372-4323372 on Build GRCh38
rs746323027 Chr.19: 4323359-4323359 on Build GRCh38
rs571504763 Chr.19: 4322926-4322926 on Build GRCh38
rs775668607 Chr.19: 4323300-4323300 on Build GRCh38
rs774880132 Chr.19: 4323340-4323340 on Build GRCh38
rs368026425 Chr.19: 4323192-4323192 on Build GRCh38
rs776188536 Chr.19: 4323158-4323158 on Build GRCh38
rs148882494 Chr.19: 4323033-4323033 on Build GRCh38
rs764389765 Chr.19: 4323315-4323315 on Build GRCh38
rs182090774 Chr.19: 4323003-4323003 on Build GRCh38
rs780491960 Chr.19: 4322966-4322966 on Build GRCh38
rs779568298 Chr.19: 4323334-4323334 on Build GRCh38
rs550333648 Chr.19: 4322981-4322981 on Build GRCh38
rs764552245 Chr.19: 4322946-4322946 on Build GRCh38
rs954253820 Chr.19: 4323214-4323214 on Build GRCh38
rs1029732884 Chr.19: 4323206-4323206 on Build GRCh38
rs143776863 Chr.19: 4323053-4323053 on Build GRCh38
rs769495047 Chr.19: 4323252-4323252 on Build GRCh38
rs746492451 Chr.19: 4323231-4323231 on Build GRCh38
rs767149220 Chr.19: 4323002-4323002 on Build GRCh38
rs573182650 Chr.19: 4323011-4323011 on Build GRCh38
rs372241443 Chr.19: 4323194-4323194 on Build GRCh38
rs893064222 Chr.19: 4323073-4323073 on Build GRCh38
rs755566755 Chr.19: 4323259-4323260 on Build GRCh38
rs762344071 Chr.19: 4323057-4323057 on Build GRCh38
rs762217479 Chr.19: 4323167-4323167 on Build GRCh38
rs763491624 Chr.19: 4323183-4323183 on Build GRCh38
rs780778980 Chr.19: 4323243-4323243 on Build GRCh38
rs754417370 Chr.19: 4322954-4322954 on Build GRCh38
rs778886532 Chr.19: 4323129-4323129 on Build GRCh38
rs754185016 Chr.19: 4323013-4323013 on Build GRCh38
rs772639106 Chr.19: 4323365-4323365 on Build GRCh38
rs536864651 Chr.19: 4322975-4322975 on Build GRCh38
rs558468739 Chr.19: 4323302-4323302 on Build GRCh38
rs1044214861 Chr.19: 4322999-4322999 on Build GRCh38
rs1029701698 Chr.19: 4323006-4323006 on Build GRCh38
rs918724570 Chr.19: 4323047-4323047 on Build GRCh38
rs867424541 Chr.19: 4323055-4323055 on Build GRCh38
rs775374693 Chr.19: 4323369-4323369 on Build GRCh38
rs757039989 Chr.19: 4323128-4323128 on Build GRCh38
rs542792436 Chr.19: 4323186-4323186 on Build GRCh38
rs575863653 Chr.19: 4323050-4323050 on Build GRCh38
rs755704635 Chr.19: 4323336-4323336 on Build GRCh38
rs745755160 Chr.19: 4323134-4323134 on Build GRCh38
rs760741886 Chr.19: 4323303-4323303 on Build GRCh38
rs370463498 Chr.19: 4323108-4323108 on Build GRCh38
rs914661813 Chr.19: 4323090-4323090 on Build GRCh38
rs200879429 Chr.19: 4323198-4323198 on Build GRCh38
rs749643079 Chr.19: 4323337-4323337 on Build GRCh38
rs759753782 Chr.19: 4323190-4323190 on Build GRCh38
rs144527122 Chr.19: 4323024-4323024 on Build GRCh38
rs766530976 Chr.19: 4323063-4323063 on Build GRCh38
rs758681742 Chr.19: 4322965-4322965 on Build GRCh38
rs751357114 Chr.19: 4323381-4323381 on Build GRCh38
rs367629466 Chr.19: 4323062-4323062 on Build GRCh38
rs771556879 Chr.19: 4323278-4323278 on Build GRCh38
rs368914993 Chr.19: 4323339-4323339 on Build GRCh38
rs762043518 Chr.19: 4323378-4323378 on Build GRCh38
rs772027664 Chr.19: 4323135-4323135 on Build GRCh38
rs747923647 Chr.19: 4322967-4322967 on Build GRCh38
rs761003361 Chr.19: 4323004-4323004 on Build GRCh38
rs151077874 Chr.19: 4323056-4323056 on Build GRCh38
rs746580293 Chr.19: 4323151-4323151 on Build GRCh38
rs373867402 Chr.19: 4323137-4323137 on Build GRCh38
rs767745067 Chr.19: 4323191-4323191 on Build GRCh38
rs750477041 Chr.19: 4323323-4323323 on Build GRCh38
rs565055426 Chr.19: 4323287-4323287 on Build GRCh38
rs753659338 Chr.19: 4323111-4323111 on Build GRCh38
rs562939184 Chr.19: 4323197-4323197 on Build GRCh38
rs778984896 Chr.19: 4322957-4322957 on Build GRCh38
rs745949639 Chr.19: 4322964-4322964 on Build GRCh38
rs770007894 Chr.19: 4323168-4323168 on Build GRCh38
rs775035314 Chr.19: 4323286-4323286 on Build GRCh38
rs372366315 Chr.19: 4322936-4322936 on Build GRCh38
rs757726414 Chr.19: 4322955-4322955 on Build GRCh38
rs145547298 Chr.19: 4323032-4323032 on Build GRCh38
rs748859636 Chr.19: 4323333-4323334 on Build GRCh38
rs759023466 Chr.19: 4323000-4323000 on Build GRCh38
rs539005775 Chr.19: 4323027-4323027 on Build GRCh38
rs758593665 Chr.19: 4323025-4323025 on Build GRCh38
rs755189864 Chr.19: 4323097-4323097 on Build GRCh38
rs373115179 Chr.19: 4323371-4323371 on Build GRCh38
rs558999778 Chr.19: 4323037-4323037 on Build GRCh38
rs141304180 Chr.19: 4323316-4323316 on Build GRCh38
rs747703427 Chr.19: 4323335-4323335 on Build GRCh38
rs530676411 Chr.19: 4323291-4323291 on Build GRCh38
rs375422446 Chr.19: 4322978-4322978 on Build GRCh38
rs140631506 Chr.19: 4323173-4323173 on Build GRCh38
rs758318107 Chr.19: 4323221-4323221 on Build GRCh38
rs745349961 Chr.19: 4323049-4323049 on Build GRCh38
rs528108937 Chr.19: 4323257-4323257 on Build GRCh38
rs778323509 Chr.19: 4322944-4322944 on Build GRCh38
rs765722428 Chr.19: 4323021-4323021 on Build GRCh38
rs754497632 Chr.19: 4323233-4323233 on Build GRCh38
rs377079081 Chr.19: 4323249-4323249 on Build GRCh38
rs575078874 Chr.19: 4323059-4323059 on Build GRCh38
rs750223108 Chr.19: 4323203-4323203 on Build GRCh38
rs145078106 Chr.19: 4323317-4323317 on Build GRCh38

View Full Product Details