Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4361668-4362162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3GL1
Entrez Gene ID
6455
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
Gene Aliases
CNSA1, EEN, SH3D2B, SH3P8
UniGene
Hs.490415
Gene Chromosome Location
Chr.19: 4360367-4400568 on Build GRCh38
Transcripts
NM_001199943.1 NM_001199944.1 NM_003025.3 XM_011528183.2 XM_017027146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230118 c.1032C>T p.Y344Y Chr.19: 4361675-4361675 on Build GRCh38
1304694 c.993C>T p.D331D Chr.19: 4361714-4361714 on Build GRCh38
216458 c.966C>T p.D322D Chr.19: 4361741-4361741 on Build GRCh38
4925172 c.946G>C p.D316H Chr.19: 4361761-4361761 on Build GRCh38
1717084 c.925C>T p.P309S Chr.19: 4361782-4361782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898810656 Chr.19: 4362127-4362127 on Build GRCh38
rs374770483 Chr.19: 4361805-4361805 on Build GRCh38
rs569020542 Chr.19: 4361897-4361897 on Build GRCh38
rs116801056 Chr.19: 4361952-4361952 on Build GRCh38
rs748966874 Chr.19: 4361771-4361771 on Build GRCh38
rs554608129 Chr.19: 4361924-4361924 on Build GRCh38
rs934822278 Chr.19: 4362058-4362058 on Build GRCh38
rs765497411 Chr.19: 4361803-4361803 on Build GRCh38
rs200643274 Chr.19: 4361834-4361834 on Build GRCh38
rs192508769 Chr.19: 4362077-4362077 on Build GRCh38
rs767470473 Chr.19: 4361808-4361808 on Build GRCh38
rs1018024699 Chr.19: 4361861-4361861 on Build GRCh38
rs576935472 Chr.19: 4361756-4361756 on Build GRCh38
rs548039826 Chr.19: 4361874-4361874 on Build GRCh38
rs550085467 Chr.19: 4361896-4361896 on Build GRCh38
rs947545451 Chr.19: 4361850-4361850 on Build GRCh38
rs994684993 Chr.19: 4361935-4361935 on Build GRCh38
rs541877513 Chr.19: 4362105-4362105 on Build GRCh38
rs560112509 Chr.19: 4361750-4361750 on Build GRCh38
rs370615254 Chr.19: 4362018-4362018 on Build GRCh38
rs752705794 Chr.19: 4361762-4361762 on Build GRCh38
rs753233298 Chr.19: 4361818-4361818 on Build GRCh38
rs371201004 Chr.19: 4361714-4361714 on Build GRCh38
rs577104346 Chr.19: 4361903-4361924 on Build GRCh38
rs756923164 Chr.19: 4361780-4361780 on Build GRCh38
rs934502777 Chr.19: 4361928-4361928 on Build GRCh38
rs115516342 Chr.19: 4361702-4361702 on Build GRCh38
rs1028252201 Chr.19: 4362052-4362052 on Build GRCh38
rs373251765 Chr.19: 4361741-4361741 on Build GRCh38
rs370190895 Chr.19: 4361810-4361810 on Build GRCh38
rs768746858 Chr.19: 4361799-4361799 on Build GRCh38
rs537931060 Chr.19: 4361917-4361917 on Build GRCh38
rs747516715 Chr.19: 4361796-4361796 on Build GRCh38
rs948650692 Chr.19: 4361958-4361958 on Build GRCh38
rs748107859 Chr.19: 4361845-4361845 on Build GRCh38
rs770475747 Chr.19: 4361737-4361737 on Build GRCh38
rs554068094 Chr.19: 4362016-4362016 on Build GRCh38
rs367904112 Chr.19: 4361690-4361690 on Build GRCh38
rs989998492 Chr.19: 4361942-4361942 on Build GRCh38
rs755630348 Chr.19: 4361764-4361764 on Build GRCh38
rs377322414 Chr.19: 4361743-4361743 on Build GRCh38
rs142514939 Chr.19: 4361744-4361744 on Build GRCh38
rs997497497 Chr.19: 4362120-4362120 on Build GRCh38
rs138715687 Chr.19: 4361755-4361755 on Build GRCh38
rs201812287 Chr.19: 4361823-4361823 on Build GRCh38
rs199921584 Chr.19: 4361835-4361835 on Build GRCh38
rs762081145 Chr.19: 4361802-4361802 on Build GRCh38
rs576134997 Chr.19: 4362092-4362092 on Build GRCh38
rs986371533 Chr.19: 4361856-4361856 on Build GRCh38
rs201385705 Chr.19: 4361726-4361726 on Build GRCh38
rs975732901 Chr.19: 4362034-4362034 on Build GRCh38
rs764312178 Chr.19: 4361689-4361689 on Build GRCh38
rs982905358 Chr.19: 4361960-4361960 on Build GRCh38
rs574457005 Chr.19: 4361947-4361947 on Build GRCh38
rs368446362 Chr.19: 4361844-4361844 on Build GRCh38
rs767494738 Chr.19: 4361761-4361761 on Build GRCh38
rs200448085 Chr.19: 4361740-4361740 on Build GRCh38
rs777435580 Chr.19: 4361723-4361723 on Build GRCh38
rs1017941278 Chr.19: 4361914-4361914 on Build GRCh38
rs778499476 Chr.19: 4361824-4361824 on Build GRCh38
rs540060168 Chr.19: 4361883-4361883 on Build GRCh38
rs906903976 Chr.19: 4361990-4361990 on Build GRCh38
rs370642174 Chr.19: 4361768-4361768 on Build GRCh38
rs894677569 Chr.19: 4361773-4361773 on Build GRCh38
rs899056501 Chr.19: 4361915-4361915 on Build GRCh38
rs911605865 Chr.19: 4362091-4362091 on Build GRCh38
rs749757706 Chr.19: 4361887-4361887 on Build GRCh38
rs976125279 Chr.19: 4361795-4361795 on Build GRCh38
rs141228555 Chr.19: 4361792-4361792 on Build GRCh38
rs776637003 Chr.19: 4361800-4361800 on Build GRCh38
rs978971015 Chr.19: 4362043-4362043 on Build GRCh38
rs372737423 Chr.19: 4361828-4361828 on Build GRCh38
rs561704218 Chr.19: 4361841-4361841 on Build GRCh38
rs570970521 Chr.19: 4361882-4361882 on Build GRCh38
rs755032679 Chr.19: 4361713-4361713 on Build GRCh38
rs887918263 Chr.19: 4362059-4362059 on Build GRCh38
rs902953882 Chr.19: 4361868-4361868 on Build GRCh38
rs796982465 Chr.19: 4361996-4361997 on Build GRCh38
rs371855853 Chr.19: 4361848-4361848 on Build GRCh38
rs773571556 Chr.19: 4361804-4361804 on Build GRCh38
rs138078259 Chr.19: 4362040-4362040 on Build GRCh38
rs147712071 Chr.19: 4361717-4361717 on Build GRCh38
rs1017846108 Chr.19: 4362103-4362103 on Build GRCh38
rs61192422 Chr.19: 4361995-4361996 on Build GRCh38
rs373649334 Chr.19: 4361811-4361811 on Build GRCh38
rs778554390 Chr.19: 4361788-4361788 on Build GRCh38
rs779758597 Chr.19: 4361705-4361705 on Build GRCh38
rs747601449 Chr.19: 4361716-4361716 on Build GRCh38
rs775925269 Chr.19: 4361794-4361794 on Build GRCh38
rs928883482 Chr.19: 4361753-4361753 on Build GRCh38
rs963240023 Chr.19: 4361735-4361735 on Build GRCh38
rs745540193 Chr.19: 4361826-4361826 on Build GRCh38
rs751507640 Chr.19: 4361706-4361706 on Build GRCh38
rs1044282933 Chr.19: 4361978-4361978 on Build GRCh38
rs367903090 Chr.19: 4361809-4361809 on Build GRCh38
rs995193332 Chr.19: 4362042-4362042 on Build GRCh38
rs771296697 Chr.19: 4361739-4361739 on Build GRCh38
rs146842065 Chr.19: 4361749-4361749 on Build GRCh38
rs966068326 Chr.19: 4361869-4361869 on Build GRCh38
rs1051457504 Chr.19: 4361871-4361871 on Build GRCh38
rs746167461 Chr.19: 4361791-4361791 on Build GRCh38
rs1031651439 Chr.19: 4362137-4362137 on Build GRCh38
rs756193647 Chr.19: 4362083-4362083 on Build GRCh38

View Full Product Details