Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4537023-4537556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRG1
Entrez Gene ID
116844
Gene Name
leucine rich alpha-2-glycoprotein 1
Gene Aliases
HMFT1766, LRG
UniGene
Hs.655559
Gene Chromosome Location
Chr.19: 4537215-4540024 on Build GRCh38
Transcripts
NM_052972.2

SNP Detail

SNP ID Chromosome Location
rs535212158 Chr.19: 4537506-4537506 on Build GRCh38
rs147062322 Chr.19: 4537422-4537422 on Build GRCh38
rs527386138 Chr.19: 4537371-4537371 on Build GRCh38
rs929587359 Chr.19: 4537339-4537339 on Build GRCh38
rs796826768 Chr.19: 4537472-4537472 on Build GRCh38
rs533222445 Chr.19: 4537137-4537137 on Build GRCh38
rs951034464 Chr.19: 4537082-4537082 on Build GRCh38
rs1022227920 Chr.19: 4537391-4537391 on Build GRCh38
rs866627799 Chr.19: 4537144-4537144 on Build GRCh38
rs140017805 Chr.19: 4537110-4537110 on Build GRCh38
rs563476118 Chr.19: 4537223-4537223 on Build GRCh38
rs145833936 Chr.19: 4537233-4537233 on Build GRCh38
rs989237636 Chr.19: 4537203-4537203 on Build GRCh38
rs549490465 Chr.19: 4537236-4537236 on Build GRCh38
rs763301940 Chr.19: 4537063-4537063 on Build GRCh38
rs552273854 Chr.19: 4537336-4537336 on Build GRCh38
rs979112425 Chr.19: 4537434-4537434 on Build GRCh38
rs967758931 Chr.19: 4537054-4537054 on Build GRCh38
rs927151965 Chr.19: 4537244-4537244 on Build GRCh38
rs975369923 Chr.19: 4537516-4537516 on Build GRCh38
rs912085268 Chr.19: 4537439-4537439 on Build GRCh38
rs767554387 Chr.19: 4537114-4537114 on Build GRCh38
rs937132686 Chr.19: 4537531-4537531 on Build GRCh38
rs968086677 Chr.19: 4537410-4537410 on Build GRCh38
rs954796034 Chr.19: 4537374-4537374 on Build GRCh38
rs886686169 Chr.19: 4537349-4537349 on Build GRCh38
rs377432783 Chr.19: 4537302-4537302 on Build GRCh38
rs149851480 Chr.19: 4537162-4537162 on Build GRCh38
rs752257782 Chr.19: 4537213-4537213 on Build GRCh38
rs111965887 Chr.19: 4537214-4537214 on Build GRCh38
rs1026541788 Chr.19: 4537056-4537056 on Build GRCh38
rs913109048 Chr.19: 4537210-4537210 on Build GRCh38
rs1030276916 Chr.19: 4537121-4537121 on Build GRCh38
rs141706676 Chr.19: 4537361-4537361 on Build GRCh38
rs988797102 Chr.19: 4537380-4537380 on Build GRCh38
rs7246823 Chr.19: 4537412-4537412 on Build GRCh38
rs1016413065 Chr.19: 4537358-4537358 on Build GRCh38
rs1047175485 Chr.19: 4537340-4537340 on Build GRCh38
rs10406111 Chr.19: 4537266-4537266 on Build GRCh38
rs899135874 Chr.19: 4537281-4537281 on Build GRCh38
rs560340753 Chr.19: 4537046-4537046 on Build GRCh38
rs759603554 Chr.19: 4537090-4537090 on Build GRCh38
rs573429906 Chr.19: 4537321-4537322 on Build GRCh38
rs751288362 Chr.19: 4537055-4537055 on Build GRCh38
rs927206073 Chr.19: 4537515-4537515 on Build GRCh38
rs895585513 Chr.19: 4537321-4537321 on Build GRCh38
rs11540023 Chr.19: 4537473-4537473 on Build GRCh38
rs1006922881 Chr.19: 4537355-4537355 on Build GRCh38
rs1016877949 Chr.19: 4537108-4537108 on Build GRCh38
rs550918444 Chr.19: 4537423-4537423 on Build GRCh38
rs996315288 Chr.19: 4537366-4537366 on Build GRCh38
rs1022401978 Chr.19: 4537054-4537056 on Build GRCh38
rs780527648 Chr.19: 4537470-4537470 on Build GRCh38
rs943628957 Chr.19: 4537258-4537258 on Build GRCh38
rs147685460 Chr.19: 4537492-4537492 on Build GRCh38

View Full Product Details