Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6440038-6440583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
UniGene
Hs.356231
Gene Chromosome Location
Chr.19: 6436081-6460221 on Build GRCh38
Transcripts
NM_024103.2 XM_011528274.1 XM_011528275.1 XM_011528276.1 XM_011528277.1 XM_011528278.1 XM_011528279.1 XM_011528280.1 XM_011528281.1 XM_011528282.1 XM_011528283.1 XM_011528284.1 XM_011528285.1 XM_017027285.1 XM_017027286.1 XM_017027287.1 XM_017027288.1 XM_017027289.1 XM_017027290.1 XM_017027291.1 XM_017027292.1

SNP Detail

SNP ID Chromosome Location
rs541710914 Chr.19: 6440134-6440134 on Build GRCh38
rs113007336 Chr.19: 6440354-6440354 on Build GRCh38
rs1047124168 Chr.19: 6440188-6440188 on Build GRCh38
rs997026105 Chr.19: 6440267-6440267 on Build GRCh38
rs528257498 Chr.19: 6440291-6440291 on Build GRCh38
rs543045345 Chr.19: 6440225-6440225 on Build GRCh38
rs1005677721 Chr.19: 6440228-6440228 on Build GRCh38
rs926396001 Chr.19: 6440299-6440299 on Build GRCh38
rs866345596 Chr.19: 6440165-6440165 on Build GRCh38
rs920097894 Chr.19: 6440412-6440412 on Build GRCh38
rs147955952 Chr.19: 6440148-6440148 on Build GRCh38
rs998774095 Chr.19: 6440341-6440341 on Build GRCh38
rs542832278 Chr.19: 6440278-6440278 on Build GRCh38
rs1043454214 Chr.19: 6440146-6440146 on Build GRCh38
rs549868412 Chr.19: 6440364-6440364 on Build GRCh38
rs993795672 Chr.19: 6440180-6440180 on Build GRCh38
rs746805700 Chr.19: 6440352-6440352 on Build GRCh38
rs1030220261 Chr.19: 6440273-6440273 on Build GRCh38
rs886798981 Chr.19: 6440253-6440253 on Build GRCh38
rs183518708 Chr.19: 6440386-6440386 on Build GRCh38
rs1003007950 Chr.19: 6440256-6440256 on Build GRCh38
rs535514084 Chr.19: 6440138-6440138 on Build GRCh38
rs572797452 Chr.19: 6440101-6440101 on Build GRCh38
rs997310821 Chr.19: 6440540-6440540 on Build GRCh38
rs1029858110 Chr.19: 6440541-6440541 on Build GRCh38
rs913610361 Chr.19: 6440279-6440279 on Build GRCh38
rs185773152 Chr.19: 6440166-6440166 on Build GRCh38
rs535550291 Chr.19: 6440482-6440482 on Build GRCh38
rs1038337421 Chr.19: 6440467-6440467 on Build GRCh38
rs1005561634 Chr.19: 6440455-6440455 on Build GRCh38
rs191090210 Chr.19: 6440262-6440262 on Build GRCh38
rs570338530 Chr.19: 6440135-6440135 on Build GRCh38
rs987785236 Chr.19: 6440094-6440094 on Build GRCh38
rs200394701 Chr.19: 6440348-6440349 on Build GRCh38
rs115233280 Chr.19: 6440277-6440277 on Build GRCh38
rs1017021722 Chr.19: 6440241-6440241 on Build GRCh38
rs559090599 Chr.19: 6440281-6440281 on Build GRCh38
rs979033072 Chr.19: 6440284-6440284 on Build GRCh38
rs896423740 Chr.19: 6440167-6440167 on Build GRCh38
rs937737113 Chr.19: 6440320-6440320 on Build GRCh38
rs968587914 Chr.19: 6440318-6440318 on Build GRCh38
rs942868799 Chr.19: 6440186-6440186 on Build GRCh38
rs899932433 Chr.19: 6440471-6440471 on Build GRCh38
rs564835528 Chr.19: 6440349-6440349 on Build GRCh38
rs1021534127 Chr.19: 6440107-6440107 on Build GRCh38
rs551293116 Chr.19: 6440348-6440348 on Build GRCh38

View Full Product Details