Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12351000-12351508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF442
Entrez Gene ID
79973
Gene Name
zinc finger protein 442
Gene Aliases
-
UniGene
Hs.253193
Gene Chromosome Location
Chr.19: 12349371-12372636 on Build GRCh38
Transcripts
NM_030824.2 XM_006722908.3 XM_017027316.1 XM_017027317.1 XM_017027318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854676 c.322G>A p.E108K Chr.19: 12351263-12351263 on Build GRCh38
4743897 c.544G>A p.D182N Chr.19: 12351041-12351041 on Build GRCh38
248573 c.396C>T p.C132C Chr.19: 12351189-12351189 on Build GRCh38
6830268 c.567C>A p.T189T Chr.19: 12351018-12351018 on Build GRCh38
4743898 c.378T>C p.G126G Chr.19: 12351207-12351207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763590374 Chr.19: 12351252-12351254 on Build GRCh38
rs139605905 Chr.19: 12351047-12351047 on Build GRCh38
rs745883416 Chr.19: 12351085-12351085 on Build GRCh38
rs577076651 Chr.19: 12351443-12351443 on Build GRCh38
rs867143320 Chr.19: 12351253-12351253 on Build GRCh38
rs766964704 Chr.19: 12351330-12351330 on Build GRCh38
rs1041745286 Chr.19: 12351210-12351210 on Build GRCh38
rs779226521 Chr.19: 12351187-12351187 on Build GRCh38
rs758970559 Chr.19: 12351250-12351250 on Build GRCh38
rs769610141 Chr.19: 12351215-12351215 on Build GRCh38
rs139470971 Chr.19: 12351167-12351167 on Build GRCh38
rs1030977083 Chr.19: 12351458-12351458 on Build GRCh38
rs148884848 Chr.19: 12351142-12351142 on Build GRCh38
rs200709259 Chr.19: 12351268-12351268 on Build GRCh38
rs777246702 Chr.19: 12351340-12351340 on Build GRCh38
rs149737197 Chr.19: 12351212-12351212 on Build GRCh38
rs140568989 Chr.19: 12351287-12351287 on Build GRCh38
rs559114202 Chr.19: 12351439-12351439 on Build GRCh38
rs370768648 Chr.19: 12351298-12351298 on Build GRCh38
rs768573314 Chr.19: 12351205-12351205 on Build GRCh38
rs112738148 Chr.19: 12351228-12351228 on Build GRCh38
rs148124900 Chr.19: 12351432-12351432 on Build GRCh38
rs80077701 Chr.19: 12351380-12351380 on Build GRCh38
rs774293442 Chr.19: 12351270-12351271 on Build GRCh38
rs774716348 Chr.19: 12351082-12351082 on Build GRCh38
rs778245061 Chr.19: 12351064-12351064 on Build GRCh38
rs748127648 Chr.19: 12351293-12351293 on Build GRCh38
rs924585975 Chr.19: 12351211-12351211 on Build GRCh38
rs933279025 Chr.19: 12351475-12351475 on Build GRCh38
rs372457481 Chr.19: 12351196-12351196 on Build GRCh38
rs143702602 Chr.19: 12351286-12351286 on Build GRCh38
rs558926072 Chr.19: 12351026-12351026 on Build GRCh38
rs1033465834 Chr.19: 12351388-12351388 on Build GRCh38
rs747894889 Chr.19: 12351366-12351371 on Build GRCh38
rs770548492 Chr.19: 12351319-12351319 on Build GRCh38
rs10415207 Chr.19: 12351257-12351257 on Build GRCh38
rs146815090 Chr.19: 12351266-12351266 on Build GRCh38
rs756406764 Chr.19: 12351062-12351062 on Build GRCh38
rs537651121 Chr.19: 12351191-12351191 on Build GRCh38
rs775445705 Chr.19: 12351220-12351221 on Build GRCh38
rs371229430 Chr.19: 12351117-12351117 on Build GRCh38
rs746916128 Chr.19: 12351201-12351201 on Build GRCh38
rs775251243 Chr.19: 12351094-12351094 on Build GRCh38
rs746124728 Chr.19: 12351190-12351190 on Build GRCh38
rs756962508 Chr.19: 12351043-12351043 on Build GRCh38
rs753411248 Chr.19: 12351040-12351040 on Build GRCh38
rs970765213 Chr.19: 12351346-12351346 on Build GRCh38
rs915425689 Chr.19: 12351161-12351161 on Build GRCh38
rs966673144 Chr.19: 12351462-12351462 on Build GRCh38
rs554792146 Chr.19: 12351418-12351418 on Build GRCh38
rs201814204 Chr.19: 12351048-12351048 on Build GRCh38
rs10414971 Chr.19: 12351131-12351131 on Build GRCh38
rs749453596 Chr.19: 12351070-12351070 on Build GRCh38
rs762584941 Chr.19: 12351318-12351318 on Build GRCh38
rs748724137 Chr.19: 12351061-12351061 on Build GRCh38
rs772298832 Chr.19: 12351092-12351092 on Build GRCh38
rs372360440 Chr.19: 12351120-12351120 on Build GRCh38
rs764600310 Chr.19: 12351254-12351254 on Build GRCh38
rs765957509 Chr.19: 12351122-12351122 on Build GRCh38
rs112592794 Chr.19: 12351466-12351466 on Build GRCh38
rs756698924 Chr.19: 12351132-12351132 on Build GRCh38
rs867024420 Chr.19: 12351473-12351473 on Build GRCh38
rs547916207 Chr.19: 12351046-12351046 on Build GRCh38
rs764815715 Chr.19: 12351115-12351115 on Build GRCh38
rs576574825 Chr.19: 12351425-12351425 on Build GRCh38
rs61737005 Chr.19: 12351045-12351045 on Build GRCh38
rs751143115 Chr.19: 12351242-12351242 on Build GRCh38
rs574416295 Chr.19: 12351442-12351442 on Build GRCh38
rs754416665 Chr.19: 12351256-12351256 on Build GRCh38
rs10423273 Chr.19: 12351308-12351308 on Build GRCh38
rs145133796 Chr.19: 12351108-12351108 on Build GRCh38
rs771344116 Chr.19: 12351071-12351071 on Build GRCh38
rs200641863 Chr.19: 12351063-12351063 on Build GRCh38
rs780145845 Chr.19: 12351278-12351278 on Build GRCh38
rs868331830 Chr.19: 12351227-12351227 on Build GRCh38
rs779460436 Chr.19: 12351258-12351258 on Build GRCh38
rs759035581 Chr.19: 12351321-12351321 on Build GRCh38
rs145650732 Chr.19: 12351221-12351221 on Build GRCh38
rs776447371 Chr.19: 12351208-12351208 on Build GRCh38
rs779794694 Chr.19: 12351064-12351064 on Build GRCh38
rs376756721 Chr.19: 12351316-12351316 on Build GRCh38
rs202020261 Chr.19: 12351119-12351119 on Build GRCh38
rs773045427 Chr.19: 12351311-12351311 on Build GRCh38
rs534921580 Chr.19: 12351179-12351179 on Build GRCh38
rs963596241 Chr.19: 12351067-12351067 on Build GRCh38
rs530665196 Chr.19: 12351114-12351114 on Build GRCh38
rs959101074 Chr.19: 12351411-12351411 on Build GRCh38
rs765789723 Chr.19: 12351229-12351229 on Build GRCh38
rs776557727 Chr.19: 12351111-12351111 on Build GRCh38
rs924644194 Chr.19: 12351474-12351474 on Build GRCh38
rs772196592 Chr.19: 12351368-12351369 on Build GRCh38

View Full Product Details