Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12465175-12465683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF709
Entrez Gene ID
163051
Gene Name
zinc finger protein 709
Gene Aliases
-
UniGene
Hs.631623
Gene Chromosome Location
Chr.19: 12461184-12484818 on Build GRCh38
Transcripts
NM_152601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6832594 c.406G>A p.G136R Chr.19: 12465516-12465516 on Build GRCh38
6003814 c.313A>G p.S105G Chr.19: 12465609-12465609 on Build GRCh38
991307 c.719G>A p.R240Q Chr.19: 12465203-12465203 on Build GRCh38
3891559 c.559G>A p.E187K Chr.19: 12465363-12465363 on Build GRCh38
991311 c.559G>T p.E187* Chr.19: 12465363-12465363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777192870 Chr.19: 12465280-12465280 on Build GRCh38
rs371379809 Chr.19: 12465456-12465456 on Build GRCh38
rs376497348 Chr.19: 12465507-12465507 on Build GRCh38
rs930447266 Chr.19: 12465609-12465609 on Build GRCh38
rs752731920 Chr.19: 12465384-12465384 on Build GRCh38
rs756656156 Chr.19: 12465638-12465638 on Build GRCh38
rs771624762 Chr.19: 12465449-12465449 on Build GRCh38
rs779513337 Chr.19: 12465455-12465455 on Build GRCh38
rs770506747 Chr.19: 12465407-12465408 on Build GRCh38
rs753675343 Chr.19: 12465549-12465549 on Build GRCh38
rs779568323 Chr.19: 12465604-12465604 on Build GRCh38
rs770452098 Chr.19: 12465249-12465249 on Build GRCh38
rs763264478 Chr.19: 12465327-12465327 on Build GRCh38
rs947999836 Chr.19: 12465554-12465554 on Build GRCh38
rs753172455 Chr.19: 12465339-12465339 on Build GRCh38
rs911555397 Chr.19: 12465356-12465356 on Build GRCh38
rs770239899 Chr.19: 12465307-12465307 on Build GRCh38
rs759242650 Chr.19: 12465634-12465635 on Build GRCh38
rs781441952 Chr.19: 12465576-12465576 on Build GRCh38
rs7259240 Chr.19: 12465235-12465235 on Build GRCh38
rs747167268 Chr.19: 12465437-12465437 on Build GRCh38
rs537621032 Chr.19: 12465288-12465288 on Build GRCh38
rs369251411 Chr.19: 12465226-12465226 on Build GRCh38
rs562857083 Chr.19: 12465440-12465440 on Build GRCh38
rs753529941 Chr.19: 12465397-12465397 on Build GRCh38
rs749011851 Chr.19: 12465240-12465240 on Build GRCh38
rs749797361 Chr.19: 12465651-12465651 on Build GRCh38
rs369579383 Chr.19: 12465479-12465479 on Build GRCh38
rs774781699 Chr.19: 12465502-12465502 on Build GRCh38
rs750712959 Chr.19: 12465204-12465204 on Build GRCh38
rs756089020 Chr.19: 12465394-12465394 on Build GRCh38
rs552007338 Chr.19: 12465639-12465639 on Build GRCh38
rs944461274 Chr.19: 12465363-12465363 on Build GRCh38
rs751671063 Chr.19: 12465349-12465349 on Build GRCh38
rs770157844 Chr.19: 12465464-12465464 on Build GRCh38
rs970181772 Chr.19: 12465203-12465203 on Build GRCh38
rs374882516 Chr.19: 12465391-12465391 on Build GRCh38
rs376255683 Chr.19: 12465238-12465238 on Build GRCh38
rs767701173 Chr.19: 12465370-12465370 on Build GRCh38
rs778366933 Chr.19: 12465418-12465418 on Build GRCh38
rs775037669 Chr.19: 12465558-12465558 on Build GRCh38
rs567117663 Chr.19: 12465652-12465652 on Build GRCh38
rs776426371 Chr.19: 12465511-12465512 on Build GRCh38
rs781217705 Chr.19: 12465227-12465227 on Build GRCh38
rs763882660 Chr.19: 12465548-12465548 on Build GRCh38
rs778510510 Chr.19: 12465261-12465261 on Build GRCh38
rs757070588 Chr.19: 12465557-12465557 on Build GRCh38
rs762291246 Chr.19: 12465287-12465287 on Build GRCh38
rs760353241 Chr.19: 12465366-12465367 on Build GRCh38
rs987202141 Chr.19: 12465355-12465355 on Build GRCh38
rs769159349 Chr.19: 12465279-12465279 on Build GRCh38
rs765071473 Chr.19: 12465561-12465561 on Build GRCh38
rs371695870 Chr.19: 12465505-12465505 on Build GRCh38
rs899117848 Chr.19: 12465511-12465511 on Build GRCh38
rs758523865 Chr.19: 12465222-12465222 on Build GRCh38
rs878920513 Chr.19: 12465527-12465527 on Build GRCh38
rs921781632 Chr.19: 12465588-12465588 on Build GRCh38
rs746471677 Chr.19: 12465633-12465633 on Build GRCh38
rs771188520 Chr.19: 12465497-12465497 on Build GRCh38
rs772662049 Chr.19: 12465221-12465221 on Build GRCh38
rs759621924 Chr.19: 12465359-12465359 on Build GRCh38
rs1044968609 Chr.19: 12465556-12465556 on Build GRCh38
rs745326480 Chr.19: 12465272-12465272 on Build GRCh38
rs766518380 Chr.19: 12465223-12465223 on Build GRCh38
rs773414903 Chr.19: 12465326-12465326 on Build GRCh38
rs760686703 Chr.19: 12465535-12465535 on Build GRCh38
rs745881161 Chr.19: 12465586-12465586 on Build GRCh38
rs188565094 Chr.19: 12465590-12465590 on Build GRCh38
rs758676680 Chr.19: 12465408-12465408 on Build GRCh38
rs773646499 Chr.19: 12465468-12465468 on Build GRCh38
rs376934566 Chr.19: 12465412-12465412 on Build GRCh38
rs777657184 Chr.19: 12465395-12465395 on Build GRCh38
rs550923838 Chr.19: 12465345-12465345 on Build GRCh38
rs752544330 Chr.19: 12465234-12465234 on Build GRCh38
rs374009759 Chr.19: 12465265-12465265 on Build GRCh38
rs752785395 Chr.19: 12465523-12465523 on Build GRCh38

View Full Product Details