Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12899000-12899494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCDH
Entrez Gene ID
2639
Gene Name
glutaryl-CoA dehydrogenase
Gene Aliases
ACAD5, GCD
UniGene
Hs.532699
Gene Chromosome Location
Chr.19: 12891129-12915345 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1 NM_001105578.1 XM_005259848.4 XM_011527882.2 XM_011527883.2

Gene Symbol
SYCE2
Entrez Gene ID
256126
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
CESC1
UniGene
Hs.655173
Gene Chromosome Location
Chr.19: 12898229-12919674 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1 NM_001105578.1 XM_005259848.4 XM_011527882.2 XM_011527883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446322 c.1128C>T p.H376H Chr.19: 12899484-12899484 on Build GRCh38
4455074 c.657A>G p.*219W Chr.19: 12899341-12899341 on Build GRCh38
6446321 c.1260C>T p.H420H Chr.19: 12899484-12899484 on Build GRCh38
4462857 c.1131C>T p.A377A Chr.19: 12899487-12899487 on Build GRCh38
4462856 c.1263C>T p.A421A Chr.19: 12899487-12899487 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770370531 Chr.19: 12899396-12899396 on Build GRCh38
rs532398592 Chr.19: 12899260-12899260 on Build GRCh38
rs1019680244 Chr.19: 12899309-12899309 on Build GRCh38
rs751543103 Chr.19: 12899366-12899366 on Build GRCh38
rs926799623 Chr.19: 12899043-12899043 on Build GRCh38
rs945534924 Chr.19: 12899141-12899141 on Build GRCh38
rs553191132 Chr.19: 12899214-12899214 on Build GRCh38
rs201407990 Chr.19: 12899453-12899453 on Build GRCh38
rs747302250 Chr.19: 12899348-12899353 on Build GRCh38
rs952621636 Chr.19: 12899039-12899040 on Build GRCh38
rs371447029 Chr.19: 12899445-12899445 on Build GRCh38
rs559779791 Chr.19: 12899359-12899359 on Build GRCh38
rs780617569 Chr.19: 12899303-12899303 on Build GRCh38
rs765367111 Chr.19: 12899353-12899353 on Build GRCh38
rs770133763 Chr.19: 12899440-12899440 on Build GRCh38
rs117984891 Chr.19: 12899161-12899161 on Build GRCh38
rs901120308 Chr.19: 12899135-12899135 on Build GRCh38
rs879066665 Chr.19: 12899246-12899246 on Build GRCh38
rs979742155 Chr.19: 12899071-12899071 on Build GRCh38
rs756872756 Chr.19: 12899147-12899147 on Build GRCh38
rs199999619 Chr.19: 12899466-12899466 on Build GRCh38
rs912994327 Chr.19: 12899120-12899120 on Build GRCh38
rs368707026 Chr.19: 12899347-12899347 on Build GRCh38
rs970957171 Chr.19: 12899022-12899022 on Build GRCh38
rs757434577 Chr.19: 12899406-12899407 on Build GRCh38
rs1008684561 Chr.19: 12899290-12899291 on Build GRCh38
rs982302178 Chr.19: 12899033-12899033 on Build GRCh38
rs970696781 Chr.19: 12899345-12899345 on Build GRCh38
rs747924530 Chr.19: 12899379-12899379 on Build GRCh38
rs777197667 Chr.19: 12899424-12899424 on Build GRCh38
rs1053547842 Chr.19: 12899058-12899058 on Build GRCh38
rs952572363 Chr.19: 12899384-12899384 on Build GRCh38
rs121434368 Chr.19: 12899471-12899471 on Build GRCh38
rs751631637 Chr.19: 12899444-12899444 on Build GRCh38
rs184847392 Chr.19: 12899442-12899442 on Build GRCh38
rs938379681 Chr.19: 12899116-12899116 on Build GRCh38
rs993539907 Chr.19: 12899323-12899323 on Build GRCh38
rs769283497 Chr.19: 12899423-12899423 on Build GRCh38
rs938134014 Chr.19: 12899047-12899047 on Build GRCh38
rs754877145 Chr.19: 12899367-12899367 on Build GRCh38
rs777025420 Chr.19: 12899419-12899419 on Build GRCh38
rs547797520 Chr.19: 12899301-12899301 on Build GRCh38
rs945102219 Chr.19: 12899074-12899074 on Build GRCh38
rs776172011 Chr.19: 12899321-12899321 on Build GRCh38
rs375825710 Chr.19: 12899070-12899070 on Build GRCh38
rs781028895 Chr.19: 12899378-12899378 on Build GRCh38
rs767494242 Chr.19: 12899451-12899451 on Build GRCh38
rs778174422 Chr.19: 12899299-12899301 on Build GRCh38
rs774546166 Chr.19: 12899464-12899464 on Build GRCh38
rs766394111 Chr.19: 12899365-12899365 on Build GRCh38
rs747562138 Chr.19: 12899418-12899418 on Build GRCh38
rs761215274 Chr.19: 12899322-12899322 on Build GRCh38
rs933926888 Chr.19: 12899148-12899148 on Build GRCh38
rs745657713 Chr.19: 12899450-12899451 on Build GRCh38
rs749703088 Chr.19: 12899304-12899304 on Build GRCh38
rs982376975 Chr.19: 12899388-12899388 on Build GRCh38
rs200750801 Chr.19: 12899333-12899333 on Build GRCh38
rs371416403 Chr.19: 12899332-12899332 on Build GRCh38
rs745373181 Chr.19: 12899170-12899170 on Build GRCh38
rs143166329 Chr.19: 12899117-12899117 on Build GRCh38
rs778544038 Chr.19: 12899415-12899415 on Build GRCh38
rs777295436 Chr.19: 12899390-12899390 on Build GRCh38
rs750605333 Chr.19: 12899355-12899355 on Build GRCh38
rs755878919 Chr.19: 12899387-12899387 on Build GRCh38
rs1001073929 Chr.19: 12899198-12899198 on Build GRCh38
rs773341347 Chr.19: 12899441-12899441 on Build GRCh38
rs563496118 Chr.19: 12899091-12899091 on Build GRCh38
rs561275591 Chr.19: 12899311-12899311 on Build GRCh38
rs771517320 Chr.19: 12899349-12899350 on Build GRCh38
rs748930169 Chr.19: 12899395-12899395 on Build GRCh38
rs762100038 Chr.19: 12899427-12899427 on Build GRCh38

View Full Product Details