Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44700785-44701293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf228
Entrez Gene ID
339541
Gene Name
chromosome 1 open reading frame 228
Gene Aliases
NCRNA00082, p40
UniGene
Hs.173679
Gene Chromosome Location
Chr.1: 44674696-44725591 on Build GRCh38
Transcripts
NM_001145636.1 XM_006710603.2 XM_006710604.2 XM_011541340.1 XM_011541341.1 XM_011541343.1 XM_011541345.1 XM_011541349.1 XM_011541354.1 XM_011541355.1 XM_011541356.2 XM_017001140.1 XM_017001141.1 XM_017001142.1 XM_017001143.1 XM_017001144.1 XM_017001145.1 XM_017001146.1 XM_017001147.1 XM_017001148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6578832 c.484G>C p.E162Q Chr.1: 44700964-44700964 on Build GRCh38
3419264 c.530A>G p.H177R Chr.1: 44701010-44701010 on Build GRCh38
7428178 c.504G>A p.V168V Chr.1: 44700984-44700984 on Build GRCh38
5780782 c.516G>A p.L172L Chr.1: 44700996-44700996 on Build GRCh38
1660870 c.565G>A p.G189S Chr.1: 44701045-44701045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933568614 Chr.1: 44701214-44701214 on Build GRCh38
rs1014296825 Chr.1: 44701010-44701010 on Build GRCh38
rs774401018 Chr.1: 44701099-44701099 on Build GRCh38
rs865964741 Chr.1: 44701013-44701013 on Build GRCh38
rs2355710 Chr.1: 44700841-44700841 on Build GRCh38
rs997809773 Chr.1: 44700828-44700828 on Build GRCh38
rs559763206 Chr.1: 44700928-44700928 on Build GRCh38
rs975760904 Chr.1: 44701160-44701160 on Build GRCh38
rs775159506 Chr.1: 44701012-44701012 on Build GRCh38
rs889377488 Chr.1: 44701225-44701225 on Build GRCh38
rs372099401 Chr.1: 44700972-44700972 on Build GRCh38
rs4322271 Chr.1: 44701045-44701045 on Build GRCh38
rs763933805 Chr.1: 44701069-44701069 on Build GRCh38
rs750034772 Chr.1: 44701254-44701254 on Build GRCh38
rs935278340 Chr.1: 44700976-44700976 on Build GRCh38
rs370463783 Chr.1: 44701088-44701088 on Build GRCh38
rs952287585 Chr.1: 44701062-44701062 on Build GRCh38
rs773547144 Chr.1: 44700866-44700866 on Build GRCh38
rs889164293 Chr.1: 44700872-44700873 on Build GRCh38
rs561930475 Chr.1: 44701074-44701074 on Build GRCh38
rs200737089 Chr.1: 44700872-44700872 on Build GRCh38
rs967259153 Chr.1: 44701027-44701027 on Build GRCh38
rs114369447 Chr.1: 44701224-44701224 on Build GRCh38
rs761287308 Chr.1: 44701205-44701205 on Build GRCh38
rs896431655 Chr.1: 44701229-44701229 on Build GRCh38
rs768474560 Chr.1: 44700856-44700856 on Build GRCh38
rs543698474 Chr.1: 44700811-44700811 on Build GRCh38
rs908939914 Chr.1: 44701130-44701130 on Build GRCh38
rs979124761 Chr.1: 44701202-44701202 on Build GRCh38
rs762709558 Chr.1: 44701068-44701068 on Build GRCh38
rs764867154 Chr.1: 44701127-44701127 on Build GRCh38
rs530966313 Chr.1: 44701108-44701108 on Build GRCh38
rs78729868 Chr.1: 44701184-44701184 on Build GRCh38
rs550874286 Chr.1: 44701118-44701118 on Build GRCh38
rs1027837227 Chr.1: 44701052-44701052 on Build GRCh38
rs1045768849 Chr.1: 44701237-44701237 on Build GRCh38
rs958132183 Chr.1: 44700941-44700941 on Build GRCh38
rs143884786 Chr.1: 44701213-44701213 on Build GRCh38
rs533128830 Chr.1: 44701199-44701199 on Build GRCh38
rs896387943 Chr.1: 44701005-44701005 on Build GRCh38
rs931731807 Chr.1: 44700906-44700906 on Build GRCh38
rs575641425 Chr.1: 44701075-44701075 on Build GRCh38
rs185823968 Chr.1: 44700922-44700922 on Build GRCh38
rs76132381 Chr.1: 44701011-44701011 on Build GRCh38
rs192194233 Chr.1: 44700824-44700824 on Build GRCh38
rs747276218 Chr.1: 44700918-44700918 on Build GRCh38
rs903653146 Chr.1: 44700929-44700929 on Build GRCh38
rs921564177 Chr.1: 44701196-44701196 on Build GRCh38
rs949242896 Chr.1: 44701230-44701230 on Build GRCh38
rs921720458 Chr.1: 44700861-44700861 on Build GRCh38
rs1052803292 Chr.1: 44701003-44701003 on Build GRCh38
rs1015621803 Chr.1: 44701112-44701112 on Build GRCh38
rs966312595 Chr.1: 44701146-44701146 on Build GRCh38

View Full Product Details