Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44801544-44802056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK3
Entrez Gene ID
1263
Gene Name
polo like kinase 3
Gene Aliases
CNK, FNK, PLK-3, PRK
UniGene
Hs.632415
Gene Chromosome Location
Chr.1: 44799952-44805995 on Build GRCh38
Transcripts
NM_004073.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910068 c.534G>T p.K178N Chr.1: 44801930-44801930 on Build GRCh38
910067 c.383G>A p.R128Q Chr.1: 44801686-44801686 on Build GRCh38
6750230 c.454T>G p.F152V Chr.1: 44801850-44801850 on Build GRCh38
4830355 c.395C>T p.S132F Chr.1: 44801698-44801698 on Build GRCh38
6750190 c.491A>C p.Y164S Chr.1: 44801677-44801677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769591382 Chr.1: 44801785-44801788 on Build GRCh38
rs144145740 Chr.1: 44801978-44801978 on Build GRCh38
rs759648683 Chr.1: 44801987-44801987 on Build GRCh38
rs201696175 Chr.1: 44801868-44801868 on Build GRCh38
rs753006230 Chr.1: 44801647-44801647 on Build GRCh38
rs761184514 Chr.1: 44801829-44801829 on Build GRCh38
rs373585256 Chr.1: 44801826-44801826 on Build GRCh38
rs765341257 Chr.1: 44801779-44801779 on Build GRCh38
rs780845966 Chr.1: 44801783-44801784 on Build GRCh38
rs774543553 Chr.1: 44801949-44801949 on Build GRCh38
rs771899950 Chr.1: 44801595-44801595 on Build GRCh38
rs866781128 Chr.1: 44801739-44801739 on Build GRCh38
rs747813654 Chr.1: 44801793-44801793 on Build GRCh38
rs779848699 Chr.1: 44801785-44801785 on Build GRCh38
rs779797606 Chr.1: 44801685-44801685 on Build GRCh38
rs758319360 Chr.1: 44801782-44801782 on Build GRCh38
rs759667934 Chr.1: 44801981-44801982 on Build GRCh38
rs372124188 Chr.1: 44801897-44801897 on Build GRCh38
rs544406648 Chr.1: 44801722-44801722 on Build GRCh38
rs772646740 Chr.1: 44801825-44801825 on Build GRCh38
rs770721843 Chr.1: 44801578-44801578 on Build GRCh38
rs554442652 Chr.1: 44801995-44801995 on Build GRCh38
rs751494257 Chr.1: 44801881-44801881 on Build GRCh38
rs749267376 Chr.1: 44801576-44801576 on Build GRCh38
rs544668289 Chr.1: 44801570-44801570 on Build GRCh38
rs754630035 Chr.1: 44801788-44801788 on Build GRCh38
rs762068566 Chr.1: 44801834-44801834 on Build GRCh38
rs770887437 Chr.1: 44801934-44801940 on Build GRCh38
rs17885796 Chr.1: 44801794-44801794 on Build GRCh38
rs370642204 Chr.1: 44801841-44801841 on Build GRCh38
rs760988578 Chr.1: 44801754-44801754 on Build GRCh38
rs766738837 Chr.1: 44801636-44801636 on Build GRCh38
rs754816024 Chr.1: 44801882-44801882 on Build GRCh38
rs777158496 Chr.1: 44801900-44801900 on Build GRCh38
rs757343328 Chr.1: 44801777-44801777 on Build GRCh38
rs985432770 Chr.1: 44801698-44801698 on Build GRCh38
rs376275667 Chr.1: 44801684-44801684 on Build GRCh38
rs147843910 Chr.1: 44801736-44801736 on Build GRCh38
rs770002752 Chr.1: 44801948-44801948 on Build GRCh38
rs144798633 Chr.1: 44801646-44801646 on Build GRCh38
rs745577907 Chr.1: 44801588-44801588 on Build GRCh38
rs745691603 Chr.1: 44801918-44801918 on Build GRCh38
rs192834831 Chr.1: 44801907-44801907 on Build GRCh38
rs750369907 Chr.1: 44801781-44801781 on Build GRCh38
rs756362842 Chr.1: 44801654-44801654 on Build GRCh38
rs142872152 Chr.1: 44801674-44801674 on Build GRCh38
rs140199593 Chr.1: 44801630-44801630 on Build GRCh38
rs372956515 Chr.1: 44801747-44801747 on Build GRCh38
rs746750110 Chr.1: 44801786-44801786 on Build GRCh38
rs747873557 Chr.1: 44801785-44801792 on Build GRCh38
rs989061985 Chr.1: 44801724-44801724 on Build GRCh38
rs775945433 Chr.1: 44801748-44801748 on Build GRCh38
rs55786996 Chr.1: 44801913-44801913 on Build GRCh38
rs954177670 Chr.1: 44801678-44801678 on Build GRCh38
rs773086172 Chr.1: 44801784-44801785 on Build GRCh38
rs368138414 Chr.1: 44801830-44801830 on Build GRCh38
rs950253386 Chr.1: 44801795-44801795 on Build GRCh38
rs546252951 Chr.1: 44802028-44802028 on Build GRCh38
rs761966974 Chr.1: 44801943-44801943 on Build GRCh38
rs746609304 Chr.1: 44801590-44801591 on Build GRCh38
rs187943956 Chr.1: 44801721-44801721 on Build GRCh38
rs3795716 Chr.1: 44802026-44802026 on Build GRCh38
rs745809141 Chr.1: 44801677-44801677 on Build GRCh38
rs775305276 Chr.1: 44801983-44801983 on Build GRCh38
rs778018679 Chr.1: 44801664-44801664 on Build GRCh38
rs776080894 Chr.1: 44801618-44801618 on Build GRCh38
rs780761434 Chr.1: 44801906-44801906 on Build GRCh38
rs754105582 Chr.1: 44801767-44801767 on Build GRCh38
rs769703531 Chr.1: 44801980-44801980 on Build GRCh38
rs749257106 Chr.1: 44801808-44801810 on Build GRCh38
rs777016586 Chr.1: 44801941-44801941 on Build GRCh38
rs780934582 Chr.1: 44801885-44801885 on Build GRCh38
rs770557218 Chr.1: 44801904-44801904 on Build GRCh38
rs17886752 Chr.1: 44801998-44801998 on Build GRCh38
rs373341543 Chr.1: 44801673-44801673 on Build GRCh38
rs775184931 Chr.1: 44801597-44801597 on Build GRCh38
rs548445780 Chr.1: 44801971-44801971 on Build GRCh38
rs17880195 Chr.1: 44801938-44801944 on Build GRCh38
rs747786002 Chr.1: 44801891-44801891 on Build GRCh38
rs746698594 Chr.1: 44801702-44801702 on Build GRCh38
rs367678628 Chr.1: 44801836-44801836 on Build GRCh38
rs748762833 Chr.1: 44801901-44801901 on Build GRCh38
rs764341842 Chr.1: 44801760-44801760 on Build GRCh38
rs368367546 Chr.1: 44801965-44801965 on Build GRCh38
rs768197931 Chr.1: 44801607-44801607 on Build GRCh38
rs937120397 Chr.1: 44801879-44801879 on Build GRCh38
rs1014670596 Chr.1: 44801622-44801622 on Build GRCh38
rs968028040 Chr.1: 44802019-44802019 on Build GRCh38
rs767843437 Chr.1: 44801984-44801987 on Build GRCh38
rs956047695 Chr.1: 44801727-44801727 on Build GRCh38
rs17880618 Chr.1: 44801586-44801586 on Build GRCh38
rs1049270033 Chr.1: 44801849-44801849 on Build GRCh38
rs767802375 Chr.1: 44801737-44801737 on Build GRCh38
rs997738705 Chr.1: 44801992-44801992 on Build GRCh38
rs751958856 Chr.1: 44801643-44801643 on Build GRCh38
rs780889952 Chr.1: 44801789-44801789 on Build GRCh38
rs746382553 Chr.1: 44801803-44801803 on Build GRCh38
rs142337193 Chr.1: 44801723-44801723 on Build GRCh38
rs754707242 Chr.1: 44801592-44801593 on Build GRCh38
rs760302006 Chr.1: 44801603-44801603 on Build GRCh38
rs758282351 Chr.1: 44801856-44801856 on Build GRCh38
rs375166158 Chr.1: 44801910-44801910 on Build GRCh38

View Full Product Details