Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13889874-13890367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 XM_017027301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003516 c.776G>T p.R259M Chr.19: 13890080-13890080 on Build GRCh38
7262982 c.757delC p.Q253fs*90 Chr.19: 13890099-13890099 on Build GRCh38
7211093 c.652G>A p.D218N Chr.19: 13890204-13890204 on Build GRCh38
401807 c.531G>T p.Q177H Chr.19: 13890325-13890325 on Build GRCh38
5748016 c.853A>G p.T285A Chr.19: 13890003-13890003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768983179 Chr.19: 13889930-13889930 on Build GRCh38
rs779495894 Chr.19: 13890006-13890006 on Build GRCh38
rs757389709 Chr.19: 13890344-13890344 on Build GRCh38
rs376578744 Chr.19: 13890213-13890213 on Build GRCh38
rs372991147 Chr.19: 13890165-13890165 on Build GRCh38
rs761978855 Chr.19: 13889954-13889954 on Build GRCh38
rs555578448 Chr.19: 13890147-13890147 on Build GRCh38
rs145123144 Chr.19: 13889953-13889953 on Build GRCh38
rs144892455 Chr.19: 13890098-13890098 on Build GRCh38
rs567787641 Chr.19: 13890162-13890162 on Build GRCh38
rs771219621 Chr.19: 13890190-13890190 on Build GRCh38
rs765592852 Chr.19: 13890088-13890088 on Build GRCh38
rs747014631 Chr.19: 13890278-13890278 on Build GRCh38
rs772439235 Chr.19: 13890266-13890267 on Build GRCh38
rs780700795 Chr.19: 13890025-13890025 on Build GRCh38
rs149879449 Chr.19: 13889950-13889950 on Build GRCh38
rs761733743 Chr.19: 13890049-13890049 on Build GRCh38
rs748176154 Chr.19: 13890144-13890144 on Build GRCh38
rs750663995 Chr.19: 13889973-13889973 on Build GRCh38
rs77941658 Chr.19: 13890139-13890139 on Build GRCh38
rs939620339 Chr.19: 13890178-13890178 on Build GRCh38
rs1004827399 Chr.19: 13890132-13890132 on Build GRCh38
rs753470005 Chr.19: 13890312-13890312 on Build GRCh38
rs765935675 Chr.19: 13890311-13890311 on Build GRCh38
rs777463792 Chr.19: 13889915-13889915 on Build GRCh38
rs747000381 Chr.19: 13890340-13890340 on Build GRCh38
rs780642908 Chr.19: 13889899-13889899 on Build GRCh38
rs753558315 Chr.19: 13890252-13890252 on Build GRCh38
rs752198106 Chr.19: 13890326-13890327 on Build GRCh38
rs766910345 Chr.19: 13889993-13889993 on Build GRCh38
rs752065500 Chr.19: 13890307-13890307 on Build GRCh38
rs756704932 Chr.19: 13890092-13890092 on Build GRCh38
rs370920646 Chr.19: 13890183-13890183 on Build GRCh38
rs569548881 Chr.19: 13890023-13890023 on Build GRCh38
rs749028986 Chr.19: 13890121-13890121 on Build GRCh38
rs148701997 Chr.19: 13890276-13890276 on Build GRCh38
rs370180523 Chr.19: 13890223-13890223 on Build GRCh38
rs994665667 Chr.19: 13890020-13890020 on Build GRCh38
rs201032125 Chr.19: 13890033-13890033 on Build GRCh38
rs114838605 Chr.19: 13890109-13890109 on Build GRCh38
rs142863596 Chr.19: 13890280-13890280 on Build GRCh38
rs774655091 Chr.19: 13890098-13890098 on Build GRCh38
rs774358429 Chr.19: 13890039-13890039 on Build GRCh38
rs755726277 Chr.19: 13890112-13890112 on Build GRCh38
rs763250600 Chr.19: 13890303-13890303 on Build GRCh38
rs201485058 Chr.19: 13890265-13890265 on Build GRCh38
rs757958519 Chr.19: 13889906-13889906 on Build GRCh38
rs766737643 Chr.19: 13890061-13890061 on Build GRCh38
rs762085330 Chr.19: 13890256-13890263 on Build GRCh38
rs755634391 Chr.19: 13890238-13890238 on Build GRCh38
rs761222402 Chr.19: 13890293-13890325 on Build GRCh38
rs749626528 Chr.19: 13890001-13890001 on Build GRCh38
rs752475723 Chr.19: 13890321-13890321 on Build GRCh38
rs527298477 Chr.19: 13889941-13889941 on Build GRCh38
rs529452628 Chr.19: 13889952-13889952 on Build GRCh38
rs767577795 Chr.19: 13889963-13889963 on Build GRCh38
rs773613379 Chr.19: 13890285-13890300 on Build GRCh38
rs759850697 Chr.19: 13890287-13890287 on Build GRCh38
rs771993838 Chr.19: 13890262-13890262 on Build GRCh38
rs886249763 Chr.19: 13890267-13890267 on Build GRCh38
rs773294466 Chr.19: 13890053-13890053 on Build GRCh38
rs775885327 Chr.19: 13890298-13890298 on Build GRCh38
rs764604383 Chr.19: 13890304-13890304 on Build GRCh38
rs912893251 Chr.19: 13890282-13890282 on Build GRCh38
rs369871268 Chr.19: 13890003-13890003 on Build GRCh38
rs200089068 Chr.19: 13890205-13890205 on Build GRCh38
rs555971035 Chr.19: 13890214-13890214 on Build GRCh38
rs753309834 Chr.19: 13890089-13890089 on Build GRCh38
rs144556741 Chr.19: 13890345-13890345 on Build GRCh38
rs533639778 Chr.19: 13890051-13890051 on Build GRCh38
rs2305775 Chr.19: 13890057-13890057 on Build GRCh38
rs768503544 Chr.19: 13890124-13890124 on Build GRCh38
rs749591307 Chr.19: 13889906-13889916 on Build GRCh38
rs754718658 Chr.19: 13890256-13890256 on Build GRCh38
rs147137336 Chr.19: 13890260-13890260 on Build GRCh38
rs764541473 Chr.19: 13890220-13890220 on Build GRCh38
rs374898954 Chr.19: 13890250-13890250 on Build GRCh38
rs771553371 Chr.19: 13890159-13890159 on Build GRCh38
rs371277207 Chr.19: 13890319-13890319 on Build GRCh38
rs141210941 Chr.19: 13890028-13890028 on Build GRCh38
rs759971791 Chr.19: 13890204-13890204 on Build GRCh38
rs201941868 Chr.19: 13890259-13890259 on Build GRCh38
rs760177608 Chr.19: 13890310-13890310 on Build GRCh38
rs776997563 Chr.19: 13890196-13890196 on Build GRCh38
rs754205833 Chr.19: 13890062-13890062 on Build GRCh38
rs963405881 Chr.19: 13890166-13890166 on Build GRCh38
rs1022357095 Chr.19: 13890305-13890305 on Build GRCh38
rs754702885 Chr.19: 13890314-13890314 on Build GRCh38
rs756544648 Chr.19: 13889984-13889984 on Build GRCh38
rs764766078 Chr.19: 13890309-13890311 on Build GRCh38
rs755406846 Chr.19: 13890071-13890071 on Build GRCh38
rs770020516 Chr.19: 13890296-13890296 on Build GRCh38
rs749115967 Chr.19: 13890034-13890034 on Build GRCh38
rs779707758 Chr.19: 13890116-13890116 on Build GRCh38
rs758128875 Chr.19: 13890334-13890334 on Build GRCh38
rs773301362 Chr.19: 13890177-13890177 on Build GRCh38
rs746559438 Chr.19: 13889938-13889938 on Build GRCh38
rs745579606 Chr.19: 13889942-13889942 on Build GRCh38
rs139071536 Chr.19: 13889944-13889944 on Build GRCh38
rs746441946 Chr.19: 13890008-13890008 on Build GRCh38
rs1030598281 Chr.19: 13890099-13890099 on Build GRCh38
rs974861111 Chr.19: 13890063-13890063 on Build GRCh38
rs369138973 Chr.19: 13889900-13889900 on Build GRCh38

View Full Product Details