Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16750383-16750900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD1
Entrez Gene ID
284434
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
-
UniGene
Hs.406014
Gene Chromosome Location
Chr.19: 16719526-16817963 on Build GRCh38
Transcripts
NM_001007525.3 NM_001290355.1 XM_011527929.1 XM_011527930.2 XM_011527931.2 XM_011527934.1 XM_011527935.1 XM_017026669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385738 c.1361C>A p.S454Y Chr.19: 16750408-16750408 on Build GRCh38
4708816 c.1363C>T p.R455* Chr.19: 16750410-16750410 on Build GRCh38
271865 c.1744G>A p.V582M Chr.19: 16750386-16750386 on Build GRCh38
1630749 c.1761G>A p.V587V Chr.19: 16750403-16750403 on Build GRCh38
3530108 c.1767C>T p.S589S Chr.19: 16750409-16750409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755638592 Chr.19: 16750433-16750433 on Build GRCh38
rs34231229 Chr.19: 16750627-16750627 on Build GRCh38
rs879492115 Chr.19: 16750697-16750697 on Build GRCh38
rs1026725089 Chr.19: 16750481-16750481 on Build GRCh38
rs865915852 Chr.19: 16750408-16750408 on Build GRCh38
rs879391924 Chr.19: 16750696-16750696 on Build GRCh38
rs922822218 Chr.19: 16750849-16750849 on Build GRCh38
rs1035297383 Chr.19: 16750875-16750875 on Build GRCh38
rs932651238 Chr.19: 16750670-16750670 on Build GRCh38
rs961648338 Chr.19: 16750837-16750837 on Build GRCh38
rs567512263 Chr.19: 16750667-16750667 on Build GRCh38
rs1009436283 Chr.19: 16750629-16750634 on Build GRCh38
rs753056090 Chr.19: 16750415-16750415 on Build GRCh38
rs933087315 Chr.19: 16750587-16750587 on Build GRCh38
rs35901830 Chr.19: 16750544-16750545 on Build GRCh38
rs1044233664 Chr.19: 16750603-16750603 on Build GRCh38
rs745620126 Chr.19: 16750478-16750478 on Build GRCh38
rs551104474 Chr.19: 16750487-16750487 on Build GRCh38
rs780018372 Chr.19: 16750451-16750451 on Build GRCh38
rs919044059 Chr.19: 16750418-16750418 on Build GRCh38
rs531304336 Chr.19: 16750463-16750463 on Build GRCh38
rs940219170 Chr.19: 16750531-16750531 on Build GRCh38
rs979753883 Chr.19: 16750512-16750512 on Build GRCh38
rs570037005 Chr.19: 16750736-16750736 on Build GRCh38
rs75813323 Chr.19: 16750864-16750864 on Build GRCh38
rs972969082 Chr.19: 16750844-16750844 on Build GRCh38
rs910136765 Chr.19: 16750489-16750489 on Build GRCh38
rs969801856 Chr.19: 16750828-16750828 on Build GRCh38
rs1007396792 Chr.19: 16750612-16750612 on Build GRCh38
rs1003924421 Chr.19: 16750751-16750751 on Build GRCh38
rs984310623 Chr.19: 16750812-16750812 on Build GRCh38
rs547516223 Chr.19: 16750594-16750594 on Build GRCh38
rs1049720590 Chr.19: 16750678-16750678 on Build GRCh38
rs775472332 Chr.19: 16750761-16750761 on Build GRCh38
rs959679858 Chr.19: 16750780-16750780 on Build GRCh38
rs201110661 Chr.19: 16750819-16750819 on Build GRCh38
rs780830235 Chr.19: 16750420-16750420 on Build GRCh38
rs35363823 Chr.19: 16750729-16750730 on Build GRCh38
rs115698605 Chr.19: 16750564-16750564 on Build GRCh38
rs992306951 Chr.19: 16750576-16750576 on Build GRCh38
rs945536350 Chr.19: 16750595-16750595 on Build GRCh38
rs970825122 Chr.19: 16750405-16750405 on Build GRCh38
rs893758407 Chr.19: 16750626-16750626 on Build GRCh38
rs556418892 Chr.19: 16750699-16750699 on Build GRCh38
rs776843065 Chr.19: 16750475-16750475 on Build GRCh38
rs756874132 Chr.19: 16750417-16750417 on Build GRCh38
rs35810105 Chr.19: 16750782-16750783 on Build GRCh38
rs1008169438 Chr.19: 16750732-16750732 on Build GRCh38
rs745437156 Chr.19: 16750430-16750430 on Build GRCh38
rs980190424 Chr.19: 16750868-16750868 on Build GRCh38
rs888840822 Chr.19: 16750608-16750608 on Build GRCh38
rs914096068 Chr.19: 16750581-16750581 on Build GRCh38
rs754371654 Chr.19: 16750410-16750410 on Build GRCh38
rs1027154459 Chr.19: 16750778-16750778 on Build GRCh38
rs1053996773 Chr.19: 16750623-16750623 on Build GRCh38
rs562796972 Chr.19: 16750411-16750411 on Build GRCh38
rs906530005 Chr.19: 16750737-16750737 on Build GRCh38
rs769597727 Chr.19: 16750483-16750483 on Build GRCh38
rs897022585 Chr.19: 16750760-16750760 on Build GRCh38
rs527397943 Chr.19: 16750567-16750567 on Build GRCh38
rs536484005 Chr.19: 16750668-16750668 on Build GRCh38
rs777660512 Chr.19: 16750429-16750429 on Build GRCh38
rs768734526 Chr.19: 16750435-16750436 on Build GRCh38

View Full Product Details