Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17943169-17943665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC124
Entrez Gene ID
115098
Gene Name
coiled-coil domain containing 124
Gene Aliases
-
UniGene
Hs.100043
Gene Chromosome Location
Chr.19: 17933015-17943985 on Build GRCh38
Transcripts
NM_001136203.1 NM_138442.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188256 c.521C>G p.T174R Chr.19: 17943564-17943564 on Build GRCh38
5972792 c.547C>T p.R183W Chr.19: 17943590-17943590 on Build GRCh38
5855033 c.508C>T p.R170W Chr.19: 17943551-17943551 on Build GRCh38
6451456 c.617G>A p.R206H Chr.19: 17943660-17943660 on Build GRCh38
4669708 c.521delC p.T174fs*23 Chr.19: 17943564-17943564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019967325 Chr.19: 17943219-17943219 on Build GRCh38
rs201072306 Chr.19: 17943531-17943531 on Build GRCh38
rs780880387 Chr.19: 17943259-17943259 on Build GRCh38
rs750682028 Chr.19: 17943381-17943423 on Build GRCh38
rs778429795 Chr.19: 17943396-17943396 on Build GRCh38
rs767162137 Chr.19: 17943315-17943315 on Build GRCh38
rs770342715 Chr.19: 17943558-17943558 on Build GRCh38
rs962840092 Chr.19: 17943243-17943243 on Build GRCh38
rs11555958 Chr.19: 17943301-17943301 on Build GRCh38
rs764078802 Chr.19: 17943469-17943469 on Build GRCh38
rs1020451476 Chr.19: 17943509-17943509 on Build GRCh38
rs372798916 Chr.19: 17943526-17943526 on Build GRCh38
rs112420170 Chr.19: 17943212-17943212 on Build GRCh38
rs545432433 Chr.19: 17943623-17943623 on Build GRCh38
rs753234796 Chr.19: 17943467-17943467 on Build GRCh38
rs868029099 Chr.19: 17943635-17943635 on Build GRCh38
rs754830225 Chr.19: 17943525-17943525 on Build GRCh38
rs183174989 Chr.19: 17943487-17943487 on Build GRCh38
rs767576336 Chr.19: 17943589-17943589 on Build GRCh38
rs777067126 Chr.19: 17943461-17943461 on Build GRCh38
rs564155220 Chr.19: 17943468-17943468 on Build GRCh38
rs149667982 Chr.19: 17943333-17943333 on Build GRCh38
rs778858953 Chr.19: 17943481-17943481 on Build GRCh38
rs562040077 Chr.19: 17943418-17943418 on Build GRCh38
rs373972823 Chr.19: 17943420-17943420 on Build GRCh38
rs866892969 Chr.19: 17943305-17943305 on Build GRCh38
rs779814220 Chr.19: 17943374-17943374 on Build GRCh38
rs370379719 Chr.19: 17943482-17943482 on Build GRCh38
rs755173481 Chr.19: 17943251-17943251 on Build GRCh38
rs749240321 Chr.19: 17943381-17943381 on Build GRCh38
rs758381298 Chr.19: 17943617-17943617 on Build GRCh38
rs781033204 Chr.19: 17943546-17943546 on Build GRCh38
rs994267868 Chr.19: 17943228-17943228 on Build GRCh38
rs779832360 Chr.19: 17943475-17943475 on Build GRCh38
rs142729821 Chr.19: 17943580-17943580 on Build GRCh38
rs769006893 Chr.19: 17943566-17943566 on Build GRCh38
rs760827266 Chr.19: 17943602-17943602 on Build GRCh38
rs373694474 Chr.19: 17943494-17943494 on Build GRCh38
rs780837327 Chr.19: 17943643-17943643 on Build GRCh38
rs756896160 Chr.19: 17943632-17943632 on Build GRCh38
rs772981762 Chr.19: 17943421-17943421 on Build GRCh38
rs774230647 Chr.19: 17943514-17943514 on Build GRCh38
rs776132627 Chr.19: 17943497-17943497 on Build GRCh38
rs2302174 Chr.19: 17943460-17943460 on Build GRCh38
rs967908564 Chr.19: 17943521-17943521 on Build GRCh38
rs781381727 Chr.19: 17943315-17943318 on Build GRCh38
rs148486654 Chr.19: 17943522-17943522 on Build GRCh38
rs1027723934 Chr.19: 17943590-17943590 on Build GRCh38
rs944801090 Chr.19: 17943595-17943595 on Build GRCh38
rs760151822 Chr.19: 17943463-17943463 on Build GRCh38
rs752783480 Chr.19: 17943345-17943345 on Build GRCh38
rs761933328 Chr.19: 17943582-17943582 on Build GRCh38
rs751482394 Chr.19: 17943629-17943629 on Build GRCh38
rs890515042 Chr.19: 17943366-17943366 on Build GRCh38
rs776481895 Chr.19: 17943242-17943243 on Build GRCh38
rs768517635 Chr.19: 17943385-17943385 on Build GRCh38
rs144428317 Chr.19: 17943353-17943353 on Build GRCh38
rs756005273 Chr.19: 17943371-17943371 on Build GRCh38
rs779122056 Chr.19: 17943254-17943254 on Build GRCh38
rs1026643129 Chr.19: 17943241-17943241 on Build GRCh38
rs368704121 Chr.19: 17943519-17943519 on Build GRCh38
rs925956474 Chr.19: 17943346-17943346 on Build GRCh38
rs1027799343 Chr.19: 17943368-17943368 on Build GRCh38
rs765410443 Chr.19: 17943340-17943340 on Build GRCh38
rs751296538 Chr.19: 17943358-17943358 on Build GRCh38
rs776602665 Chr.19: 17943533-17943533 on Build GRCh38
rs775448977 Chr.19: 17943278-17943278 on Build GRCh38
rs558099896 Chr.19: 17943214-17943214 on Build GRCh38
rs78604062 Chr.19: 17943570-17943570 on Build GRCh38
rs758572228 Chr.19: 17943352-17943352 on Build GRCh38
rs748392497 Chr.19: 17943255-17943255 on Build GRCh38
rs758202188 Chr.19: 17943529-17943529 on Build GRCh38
rs986074982 Chr.19: 17943574-17943574 on Build GRCh38
rs368177324 Chr.19: 17943464-17943464 on Build GRCh38
rs946315036 Chr.19: 17943332-17943332 on Build GRCh38
rs745627421 Chr.19: 17943260-17943260 on Build GRCh38
rs764007565 Chr.19: 17943462-17943462 on Build GRCh38
rs952318278 Chr.19: 17943287-17943287 on Build GRCh38
rs368235415 Chr.19: 17943564-17943564 on Build GRCh38
rs754935353 Chr.19: 17943480-17943480 on Build GRCh38
rs377012919 Chr.19: 17943527-17943527 on Build GRCh38
rs201845785 Chr.19: 17943504-17943504 on Build GRCh38
rs1034073037 Chr.19: 17943304-17943304 on Build GRCh38
rs201001069 Chr.19: 17943552-17943552 on Build GRCh38
rs905868003 Chr.19: 17943575-17943575 on Build GRCh38
rs751361182 Chr.19: 17943251-17943252 on Build GRCh38
rs749220204 Chr.19: 17943292-17943292 on Build GRCh38
rs746677658 Chr.19: 17943490-17943490 on Build GRCh38
rs777086914 Chr.19: 17943489-17943489 on Build GRCh38
rs992994146 Chr.19: 17943316-17943316 on Build GRCh38
rs750735928 Chr.19: 17943224-17943224 on Build GRCh38
rs542569449 Chr.19: 17943380-17943380 on Build GRCh38
rs751867293 Chr.19: 17943472-17943472 on Build GRCh38
rs764845963 Chr.19: 17943243-17943244 on Build GRCh38
rs770252773 Chr.19: 17943496-17943496 on Build GRCh38
rs760061738 Chr.19: 17943337-17943337 on Build GRCh38
rs1008030842 Chr.19: 17943205-17943205 on Build GRCh38
rs759097688 Chr.19: 17943244-17943246 on Build GRCh38
rs199982966 Chr.19: 17943321-17943321 on Build GRCh38
rs769454504 Chr.19: 17943264-17943264 on Build GRCh38
rs756106175 Chr.19: 17943644-17943646 on Build GRCh38
rs775660804 Chr.19: 17943563-17943563 on Build GRCh38
rs754363342 Chr.19: 17943247-17943247 on Build GRCh38
rs753595110 Chr.19: 17943477-17943477 on Build GRCh38
rs757111998 Chr.19: 17943259-17943260 on Build GRCh38
rs761462851 Chr.19: 17943313-17943313 on Build GRCh38
rs8104153 Chr.19: 17943323-17943323 on Build GRCh38
rs752393387 Chr.19: 17943616-17943616 on Build GRCh38
rs547133856 Chr.19: 17943200-17943200 on Build GRCh38
rs766954050 Chr.19: 17943244-17943244 on Build GRCh38
rs773477203 Chr.19: 17943596-17943596 on Build GRCh38
rs756804466 Chr.19: 17943538-17943538 on Build GRCh38
rs200440400 Chr.19: 17943587-17943587 on Build GRCh38
rs868684376 Chr.19: 17943536-17943536 on Build GRCh38
rs773795946 Chr.19: 17943311-17943311 on Build GRCh38
rs758579902 Chr.19: 17943258-17943258 on Build GRCh38
rs546726017 Chr.19: 17943502-17943502 on Build GRCh38
rs745511293 Chr.19: 17943547-17943547 on Build GRCh38
rs146510396 Chr.19: 17943613-17943613 on Build GRCh38
rs756598683 Chr.19: 17943226-17943226 on Build GRCh38

View Full Product Details