Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18194699-18195195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPV17L2
Entrez Gene ID
84769
Gene Name
MPV17 mitochondrial inner membrane protein like 2
Gene Aliases
FKSG24
UniGene
Hs.515254
Gene Chromosome Location
Chr.19: 18193230-18196742 on Build GRCh38
Transcripts
NM_032683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085310 c.414G>T p.E138D Chr.19: 18194832-18194832 on Build GRCh38
5516831 c.488C>T p.P163L Chr.19: 18195010-18195010 on Build GRCh38
7082171 c.500G>C p.R167P Chr.19: 18195022-18195022 on Build GRCh38
5144792 c.499C>T p.R167* Chr.19: 18195021-18195021 on Build GRCh38
6714945 c.547T>C p.S183P Chr.19: 18195069-18195069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199504179 Chr.19: 18195062-18195062 on Build GRCh38
rs776205205 Chr.19: 18195063-18195064 on Build GRCh38
rs550700618 Chr.19: 18195053-18195053 on Build GRCh38
rs748126818 Chr.19: 18195027-18195027 on Build GRCh38
rs544730014 Chr.19: 18194921-18194921 on Build GRCh38
rs573212590 Chr.19: 18194732-18194732 on Build GRCh38
rs779433771 Chr.19: 18194895-18194895 on Build GRCh38
rs762999197 Chr.19: 18194862-18194862 on Build GRCh38
rs946145819 Chr.19: 18194723-18194723 on Build GRCh38
rs755345253 Chr.19: 18194804-18194804 on Build GRCh38
rs746106099 Chr.19: 18195102-18195102 on Build GRCh38
rs200247021 Chr.19: 18194990-18194990 on Build GRCh38
rs370688867 Chr.19: 18195091-18195091 on Build GRCh38
rs775952160 Chr.19: 18194757-18194757 on Build GRCh38
rs759004552 Chr.19: 18195071-18195071 on Build GRCh38
rs751069693 Chr.19: 18194873-18194873 on Build GRCh38
rs888153783 Chr.19: 18195175-18195175 on Build GRCh38
rs182504662 Chr.19: 18194960-18194960 on Build GRCh38
rs774368275 Chr.19: 18194942-18194942 on Build GRCh38
rs932022207 Chr.19: 18195067-18195067 on Build GRCh38
rs201082077 Chr.19: 18194943-18194943 on Build GRCh38
rs766888670 Chr.19: 18194876-18194876 on Build GRCh38
rs761950788 Chr.19: 18194760-18194760 on Build GRCh38
rs747239343 Chr.19: 18194918-18194918 on Build GRCh38
rs749743038 Chr.19: 18194758-18194758 on Build GRCh38
rs772228188 Chr.19: 18194846-18194846 on Build GRCh38
rs1014210673 Chr.19: 18194925-18194925 on Build GRCh38
rs775485291 Chr.19: 18194932-18194932 on Build GRCh38
rs372763146 Chr.19: 18194785-18194785 on Build GRCh38
rs16982263 Chr.19: 18194768-18194768 on Build GRCh38
rs770702263 Chr.19: 18195009-18195009 on Build GRCh38
rs770463658 Chr.19: 18195138-18195138 on Build GRCh38
rs201505480 Chr.19: 18195111-18195111 on Build GRCh38
rs746672563 Chr.19: 18194726-18194726 on Build GRCh38
rs772218899 Chr.19: 18194912-18194912 on Build GRCh38
rs764087873 Chr.19: 18194865-18194865 on Build GRCh38
rs780354194 Chr.19: 18194786-18194786 on Build GRCh38
rs752705625 Chr.19: 18194770-18194770 on Build GRCh38
rs754185842 Chr.19: 18194883-18194883 on Build GRCh38
rs772040049 Chr.19: 18195123-18195123 on Build GRCh38
rs371336268 Chr.19: 18195126-18195126 on Build GRCh38
rs775360671 Chr.19: 18194854-18194854 on Build GRCh38
rs758764734 Chr.19: 18194984-18194984 on Build GRCh38
rs1019373336 Chr.19: 18194863-18194863 on Build GRCh38
rs777960452 Chr.19: 18194914-18194914 on Build GRCh38
rs749143286 Chr.19: 18195143-18195143 on Build GRCh38
rs904752117 Chr.19: 18195147-18195147 on Build GRCh38
rs868251510 Chr.19: 18194959-18194959 on Build GRCh38
rs369530601 Chr.19: 18194878-18194878 on Build GRCh38
rs746238567 Chr.19: 18194738-18194738 on Build GRCh38
rs757433018 Chr.19: 18195094-18195094 on Build GRCh38
rs746279528 Chr.19: 18194783-18194783 on Build GRCh38
rs377732895 Chr.19: 18194980-18194980 on Build GRCh38
rs780866737 Chr.19: 18194779-18194779 on Build GRCh38
rs890301876 Chr.19: 18194934-18194947 on Build GRCh38
rs755506759 Chr.19: 18194897-18194897 on Build GRCh38
rs533417217 Chr.19: 18194951-18194951 on Build GRCh38
rs200529266 Chr.19: 18195119-18195119 on Build GRCh38
rs374882868 Chr.19: 18194849-18194849 on Build GRCh38
rs746591385 Chr.19: 18195008-18195009 on Build GRCh38
rs927530781 Chr.19: 18194955-18194955 on Build GRCh38
rs908626098 Chr.19: 18195115-18195115 on Build GRCh38
rs779446782 Chr.19: 18194903-18194903 on Build GRCh38
rs756925685 Chr.19: 18194777-18194777 on Build GRCh38
rs753122252 Chr.19: 18194728-18194728 on Build GRCh38
rs772772653 Chr.19: 18195039-18195039 on Build GRCh38
rs374286427 Chr.19: 18194979-18194979 on Build GRCh38
rs566108531 Chr.19: 18195139-18195139 on Build GRCh38
rs760932530 Chr.19: 18194851-18194851 on Build GRCh38
rs760150755 Chr.19: 18195046-18195046 on Build GRCh38
rs755352078 Chr.19: 18194970-18194970 on Build GRCh38
rs368055313 Chr.19: 18194737-18194737 on Build GRCh38
rs969988790 Chr.19: 18195037-18195037 on Build GRCh38
rs749786948 Chr.19: 18194737-18194737 on Build GRCh38
rs376802431 Chr.19: 18195087-18195087 on Build GRCh38
rs1024166228 Chr.19: 18194938-18194938 on Build GRCh38
rs768526306 Chr.19: 18195015-18195015 on Build GRCh38
rs373165616 Chr.19: 18194840-18194840 on Build GRCh38
rs199816189 Chr.19: 18194813-18194813 on Build GRCh38
rs769742915 Chr.19: 18194853-18194853 on Build GRCh38
rs770323063 Chr.19: 18194741-18194741 on Build GRCh38
rs778921059 Chr.19: 18194827-18194827 on Build GRCh38
rs768522639 Chr.19: 18194939-18194939 on Build GRCh38
rs371492409 Chr.19: 18194991-18194991 on Build GRCh38
rs202082460 Chr.19: 18195085-18195085 on Build GRCh38
rs1022745648 Chr.19: 18195021-18195021 on Build GRCh38
rs367837296 Chr.19: 18194773-18194773 on Build GRCh38
rs749875208 Chr.19: 18194953-18194953 on Build GRCh38
rs757592714 Chr.19: 18195002-18195002 on Build GRCh38
rs765640001 Chr.19: 18195068-18195068 on Build GRCh38
rs538268363 Chr.19: 18195013-18195013 on Build GRCh38
rs761363562 Chr.19: 18194875-18194875 on Build GRCh38
rs748230066 Chr.19: 18194907-18194907 on Build GRCh38
rs780407494 Chr.19: 18194882-18194882 on Build GRCh38
rs755693250 Chr.19: 18194880-18194880 on Build GRCh38
rs535777068 Chr.19: 18195167-18195167 on Build GRCh38
rs771887246 Chr.19: 18195028-18195028 on Build GRCh38
rs372953081 Chr.19: 18194892-18194892 on Build GRCh38
rs746903444 Chr.19: 18195133-18195133 on Build GRCh38
rs375792025 Chr.19: 18194771-18194771 on Build GRCh38
rs961187872 Chr.19: 18194869-18194869 on Build GRCh38
rs768752488 Chr.19: 18194759-18194759 on Build GRCh38
rs752947286 Chr.19: 18195070-18195070 on Build GRCh38
rs781380868 Chr.19: 18195096-18195096 on Build GRCh38
rs201845134 Chr.19: 18195017-18195017 on Build GRCh38
rs754243377 Chr.19: 18194969-18194969 on Build GRCh38
rs747888293 Chr.19: 18195120-18195120 on Build GRCh38
rs767769579 Chr.19: 18194763-18194763 on Build GRCh38
rs762809917 Chr.19: 18194937-18194937 on Build GRCh38
rs913439901 Chr.19: 18194978-18194978 on Build GRCh38
rs776407108 Chr.19: 18195110-18195111 on Build GRCh38
rs776361240 Chr.19: 18194764-18194764 on Build GRCh38
rs369388426 Chr.19: 18194816-18194816 on Build GRCh38
rs776292304 Chr.19: 18195055-18195055 on Build GRCh38
rs2271881 Chr.19: 18195014-18195014 on Build GRCh38

View Full Product Details