Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19540270-19540809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP2
Entrez Gene ID
148113
Gene Name
cartilage intermediate layer protein 2
Gene Aliases
CLIP-2
UniGene
Hs.279574
Gene Chromosome Location
Chr.19: 19538265-19546659 on Build GRCh38
Transcripts
NM_153221.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750752 c.240C>T p.I80I Chr.19: 19540280-19540280 on Build GRCh38
4672450 c.241C>T p.R81C Chr.19: 19540281-19540281 on Build GRCh38
7298449 c.429C>T p.C143C Chr.19: 19540469-19540469 on Build GRCh38
6500839 c.376C>T p.R126C Chr.19: 19540416-19540416 on Build GRCh38
4566093 c.303_304GG>AA p.D102N Chr.19: 19540343-19540344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973402733 Chr.19: 19540465-19540465 on Build GRCh38
rs972901618 Chr.19: 19540675-19540675 on Build GRCh38
rs1015879240 Chr.19: 19540637-19540637 on Build GRCh38
rs553783271 Chr.19: 19540416-19540416 on Build GRCh38
rs901682790 Chr.19: 19540575-19540575 on Build GRCh38
rs955497886 Chr.19: 19540622-19540622 on Build GRCh38
rs899846804 Chr.19: 19540497-19540497 on Build GRCh38
rs12981405 Chr.19: 19540768-19540768 on Build GRCh38
rs746193083 Chr.19: 19540382-19540382 on Build GRCh38
rs1045351297 Chr.19: 19540392-19540392 on Build GRCh38
rs747574429 Chr.19: 19540365-19540365 on Build GRCh38
rs774402520 Chr.19: 19540429-19540429 on Build GRCh38
rs750400983 Chr.19: 19540319-19540319 on Build GRCh38
rs753667666 Chr.19: 19540464-19540464 on Build GRCh38
rs964305563 Chr.19: 19540742-19540742 on Build GRCh38
rs770076426 Chr.19: 19540393-19540393 on Build GRCh38
rs542470578 Chr.19: 19540553-19540553 on Build GRCh38
rs962490528 Chr.19: 19540648-19540648 on Build GRCh38
rs1052607181 Chr.19: 19540749-19540749 on Build GRCh38
rs752851855 Chr.19: 19540309-19540309 on Build GRCh38
rs200837636 Chr.19: 19540442-19540442 on Build GRCh38
rs908867792 Chr.19: 19540528-19540528 on Build GRCh38
rs1026215200 Chr.19: 19540699-19540699 on Build GRCh38
rs1042718192 Chr.19: 19540593-19540593 on Build GRCh38
rs562735212 Chr.19: 19540560-19540560 on Build GRCh38
rs939551863 Chr.19: 19540425-19540425 on Build GRCh38
rs112118952 Chr.19: 19540653-19540653 on Build GRCh38
rs753576788 Chr.19: 19540324-19540324 on Build GRCh38
rs914094858 Chr.19: 19540764-19540764 on Build GRCh38
rs1021801814 Chr.19: 19540559-19540559 on Build GRCh38
rs747811647 Chr.19: 19540353-19540353 on Build GRCh38
rs1011296302 Chr.19: 19540554-19540554 on Build GRCh38
rs200679414 Chr.19: 19540374-19540374 on Build GRCh38
rs761229399 Chr.19: 19540293-19540293 on Build GRCh38
rs953917929 Chr.19: 19540500-19540515 on Build GRCh38
rs369666713 Chr.19: 19540689-19540689 on Build GRCh38
rs756041640 Chr.19: 19540320-19540320 on Build GRCh38
rs765307900 Chr.19: 19540308-19540308 on Build GRCh38
rs915548944 Chr.19: 19540664-19540664 on Build GRCh38
rs777328980 Chr.19: 19540358-19540358 on Build GRCh38
rs1004038174 Chr.19: 19540630-19540630 on Build GRCh38
rs777852229 Chr.19: 19540437-19540437 on Build GRCh38
rs751053548 Chr.19: 19540484-19540485 on Build GRCh38
rs867004106 Chr.19: 19540398-19540398 on Build GRCh38
rs756155955 Chr.19: 19540426-19540426 on Build GRCh38
rs939626319 Chr.19: 19540741-19540741 on Build GRCh38
rs902454344 Chr.19: 19540613-19540613 on Build GRCh38
rs933141934 Chr.19: 19540583-19540583 on Build GRCh38
rs752356605 Chr.19: 19540480-19540480 on Build GRCh38
rs796344460 Chr.19: 19540503-19540503 on Build GRCh38
rs539835682 Chr.19: 19540397-19540397 on Build GRCh38
rs746068017 Chr.19: 19540705-19540705 on Build GRCh38
rs952992412 Chr.19: 19540704-19540704 on Build GRCh38
rs531383058 Chr.19: 19540605-19540605 on Build GRCh38
rs763206432 Chr.19: 19540417-19540417 on Build GRCh38
rs758051793 Chr.19: 19540514-19540514 on Build GRCh38
rs376021345 Chr.19: 19540364-19540364 on Build GRCh38
rs928217527 Chr.19: 19540679-19540679 on Build GRCh38
rs867408738 Chr.19: 19540411-19540411 on Build GRCh38
rs4808970 Chr.19: 19540331-19540331 on Build GRCh38
rs764213884 Chr.19: 19540313-19540313 on Build GRCh38
rs943154688 Chr.19: 19540536-19540536 on Build GRCh38
rs974913015 Chr.19: 19540762-19540762 on Build GRCh38
rs754683675 Chr.19: 19540467-19540467 on Build GRCh38
rs771505963 Chr.19: 19540371-19540371 on Build GRCh38
rs771485418 Chr.19: 19540295-19540295 on Build GRCh38
rs1011440846 Chr.19: 19540463-19540463 on Build GRCh38
rs969721655 Chr.19: 19540673-19540673 on Build GRCh38
rs773637814 Chr.19: 19540292-19540292 on Build GRCh38
rs779885757 Chr.19: 19540321-19540321 on Build GRCh38
rs777122149 Chr.19: 19540298-19540298 on Build GRCh38
rs778393507 Chr.19: 19540338-19540338 on Build GRCh38
rs1012341607 Chr.19: 19540650-19540650 on Build GRCh38
rs928992115 Chr.19: 19540486-19540486 on Build GRCh38
rs996691148 Chr.19: 19540500-19540500 on Build GRCh38
rs1032378827 Chr.19: 19540634-19540634 on Build GRCh38
rs765048575 Chr.19: 19540472-19540472 on Build GRCh38
rs757883801 Chr.19: 19540357-19540357 on Build GRCh38
rs201714553 Chr.19: 19540306-19540306 on Build GRCh38
rs980994399 Chr.19: 19540674-19540674 on Build GRCh38
rs762894785 Chr.19: 19540312-19540312 on Build GRCh38

View Full Product Details