Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32353889-32354394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF507
Entrez Gene ID
22847
Gene Name
zinc finger protein 507
Gene Aliases
-
UniGene
Hs.205392
Gene Chromosome Location
Chr.19: 32345608-32387667 on Build GRCh38
Transcripts
NM_001136156.1 NM_014910.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630845 c.1097C>T p.S366F Chr.19: 32353927-32353927 on Build GRCh38
5667099 c.1198G>A p.E400K Chr.19: 32354028-32354028 on Build GRCh38
4744055 c.1162A>C p.S388R Chr.19: 32353992-32353992 on Build GRCh38
5818752 c.1310G>A p.G437E Chr.19: 32354140-32354140 on Build GRCh38
7284847 c.1084C>T p.L362L Chr.19: 32353914-32353914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763527137 Chr.19: 32354347-32354347 on Build GRCh38
rs953034089 Chr.19: 32354346-32354346 on Build GRCh38
rs753257091 Chr.19: 32354226-32354226 on Build GRCh38
rs935148687 Chr.19: 32354093-32354093 on Build GRCh38
rs112907634 Chr.19: 32354207-32354207 on Build GRCh38
rs776282884 Chr.19: 32354107-32354107 on Build GRCh38
rs147974433 Chr.19: 32354318-32354318 on Build GRCh38
rs746197758 Chr.19: 32353947-32353947 on Build GRCh38
rs541497402 Chr.19: 32354152-32354152 on Build GRCh38
rs759737843 Chr.19: 32354336-32354336 on Build GRCh38
rs758823849 Chr.19: 32354228-32354228 on Build GRCh38
rs766773762 Chr.19: 32354134-32354134 on Build GRCh38
rs955842641 Chr.19: 32354117-32354117 on Build GRCh38
rs371157788 Chr.19: 32354032-32354032 on Build GRCh38
rs1035828670 Chr.19: 32353939-32353939 on Build GRCh38
rs767720397 Chr.19: 32354010-32354010 on Build GRCh38
rs747018400 Chr.19: 32354307-32354307 on Build GRCh38
rs776888042 Chr.19: 32354330-32354330 on Build GRCh38
rs746739319 Chr.19: 32354157-32354157 on Build GRCh38
rs747153604 Chr.19: 32354217-32354217 on Build GRCh38
rs1002029020 Chr.19: 32354155-32354155 on Build GRCh38
rs769684706 Chr.19: 32354175-32354175 on Build GRCh38
rs765653413 Chr.19: 32354342-32354342 on Build GRCh38
rs769948845 Chr.19: 32354261-32354261 on Build GRCh38
rs755409752 Chr.19: 32354067-32354067 on Build GRCh38
rs987202825 Chr.19: 32354073-32354073 on Build GRCh38
rs772903575 Chr.19: 32353999-32353999 on Build GRCh38
rs899358896 Chr.19: 32353933-32353933 on Build GRCh38
rs201011234 Chr.19: 32354031-32354031 on Build GRCh38
rs763662013 Chr.19: 32354236-32354236 on Build GRCh38
rs539162666 Chr.19: 32354112-32354112 on Build GRCh38
rs140268134 Chr.19: 32354251-32354251 on Build GRCh38
rs368028755 Chr.19: 32354194-32354194 on Build GRCh38
rs756030890 Chr.19: 32354272-32354272 on Build GRCh38
rs145399600 Chr.19: 32353986-32353986 on Build GRCh38
rs781427190 Chr.19: 32353940-32353940 on Build GRCh38
rs145107585 Chr.19: 32354089-32354089 on Build GRCh38
rs770421341 Chr.19: 32354158-32354158 on Build GRCh38
rs375485281 Chr.19: 32354133-32354133 on Build GRCh38
rs868660122 Chr.19: 32354269-32354269 on Build GRCh38
rs749737590 Chr.19: 32353960-32353960 on Build GRCh38
rs751885894 Chr.19: 32353931-32353931 on Build GRCh38
rs779992375 Chr.19: 32354275-32354275 on Build GRCh38
rs572778593 Chr.19: 32354286-32354286 on Build GRCh38
rs750987529 Chr.19: 32354242-32354242 on Build GRCh38
rs771653957 Chr.19: 32354076-32354076 on Build GRCh38
rs760963664 Chr.19: 32354199-32354199 on Build GRCh38
rs962753042 Chr.19: 32354220-32354220 on Build GRCh38
rs905901493 Chr.19: 32354069-32354069 on Build GRCh38
rs768422293 Chr.19: 32354294-32354294 on Build GRCh38
rs201809940 Chr.19: 32354168-32354168 on Build GRCh38
rs747608161 Chr.19: 32354075-32354075 on Build GRCh38
rs756615149 Chr.19: 32353948-32353948 on Build GRCh38
rs775618211 Chr.19: 32354179-32354179 on Build GRCh38
rs376942615 Chr.19: 32354068-32354068 on Build GRCh38
rs199735563 Chr.19: 32354123-32354123 on Build GRCh38
rs762970740 Chr.19: 32354188-32354188 on Build GRCh38
rs769808418 Chr.19: 32354115-32354115 on Build GRCh38
rs150068736 Chr.19: 32353984-32353984 on Build GRCh38
rs777988726 Chr.19: 32354117-32354119 on Build GRCh38
rs773101494 Chr.19: 32354196-32354196 on Build GRCh38
rs774608116 Chr.19: 32354121-32354121 on Build GRCh38
rs753171160 Chr.19: 32354146-32354146 on Build GRCh38
rs777010759 Chr.19: 32354206-32354206 on Build GRCh38
rs780651957 Chr.19: 32353951-32353951 on Build GRCh38
rs779352140 Chr.19: 32354142-32354142 on Build GRCh38
rs777518106 Chr.19: 32354084-32354084 on Build GRCh38
rs576068133 Chr.19: 32354135-32354135 on Build GRCh38
rs773362265 Chr.19: 32354303-32354303 on Build GRCh38
rs764276656 Chr.19: 32354021-32354021 on Build GRCh38
rs760478438 Chr.19: 32354008-32354008 on Build GRCh38
rs112069020 Chr.19: 32354048-32354048 on Build GRCh38
rs766053307 Chr.19: 32354011-32354011 on Build GRCh38
rs768214523 Chr.19: 32353965-32353965 on Build GRCh38
rs755497497 Chr.19: 32354136-32354136 on Build GRCh38
rs12327571 Chr.19: 32354126-32354126 on Build GRCh38
rs950330640 Chr.19: 32354353-32354353 on Build GRCh38
rs775762068 Chr.19: 32354343-32354343 on Build GRCh38
rs1038607673 Chr.19: 32354129-32354129 on Build GRCh38
rs896023652 Chr.19: 32354195-32354195 on Build GRCh38
rs745621748 Chr.19: 32354171-32354171 on Build GRCh38
rs201657508 Chr.19: 32354096-32354096 on Build GRCh38
rs759934117 Chr.19: 32354221-32354221 on Build GRCh38
rs773849111 Chr.19: 32353967-32353967 on Build GRCh38
rs754410990 Chr.19: 32354056-32354056 on Build GRCh38
rs61758385 Chr.19: 32353917-32353917 on Build GRCh38
rs780269657 Chr.19: 32354055-32354055 on Build GRCh38
rs766462052 Chr.19: 32354205-32354205 on Build GRCh38
rs973807748 Chr.19: 32354222-32354222 on Build GRCh38
rs767742977 Chr.19: 32354036-32354036 on Build GRCh38
rs200009091 Chr.19: 32354225-32354225 on Build GRCh38
rs866977942 Chr.19: 32353982-32353982 on Build GRCh38
rs750579963 Chr.19: 32354045-32354045 on Build GRCh38
rs764469911 Chr.19: 32354365-32354365 on Build GRCh38
rs201355263 Chr.19: 32354149-32354149 on Build GRCh38
rs149202856 Chr.19: 32354054-32354054 on Build GRCh38
rs762028788 Chr.19: 32354125-32354125 on Build GRCh38
rs139421096 Chr.19: 32353963-32353963 on Build GRCh38
rs987257402 Chr.19: 32354363-32354363 on Build GRCh38
rs199526138 Chr.19: 32354018-32354018 on Build GRCh38
rs915148556 Chr.19: 32354120-32354120 on Build GRCh38
rs757490118 Chr.19: 32353937-32353937 on Build GRCh38
rs781159611 Chr.19: 32354170-32354170 on Build GRCh38

View Full Product Details