Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44176618-44177132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF226
Entrez Gene ID
7769
Gene Name
zinc finger protein 226
Gene Aliases
-
UniGene
Hs.145956
Gene Chromosome Location
Chr.19: 44165062-44177685 on Build GRCh38
Transcripts
NM_001032372.1 NM_001032373.1 NM_001319088.1 NM_001319089.1 NM_001319090.1 NM_016444.2 XM_005259227.2 XM_006723367.3 XM_006723368.3 XM_006723369.2 XM_017027262.1 XM_017027263.1 XM_017027264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526778 c.1419G>A p.K473K Chr.19: 44176681-44176681 on Build GRCh38
3535476 c.1825C>T p.H609Y Chr.19: 44177087-44177087 on Build GRCh38
6290951 c.1815C>T p.H605H Chr.19: 44177077-44177077 on Build GRCh38
6303692 c.1847A>G p.N616S Chr.19: 44177109-44177109 on Build GRCh38
5698337 c.1473A>G p.Q491Q Chr.19: 44176735-44176735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202111408 Chr.19: 44176690-44176690 on Build GRCh38
rs985877854 Chr.19: 44177017-44177017 on Build GRCh38
rs756404205 Chr.19: 44176720-44176720 on Build GRCh38
rs766842293 Chr.19: 44176644-44176644 on Build GRCh38
rs370984107 Chr.19: 44177074-44177074 on Build GRCh38
rs758212263 Chr.19: 44176743-44176743 on Build GRCh38
rs575965116 Chr.19: 44177055-44177055 on Build GRCh38
rs773511574 Chr.19: 44177098-44177098 on Build GRCh38
rs747582087 Chr.19: 44176750-44176750 on Build GRCh38
rs767593738 Chr.19: 44176708-44176708 on Build GRCh38
rs760663528 Chr.19: 44176807-44176807 on Build GRCh38
rs758629812 Chr.19: 44177004-44177004 on Build GRCh38
rs767274447 Chr.19: 44176743-44176744 on Build GRCh38
rs996348629 Chr.19: 44177057-44177057 on Build GRCh38
rs748898280 Chr.19: 44176880-44176880 on Build GRCh38
rs745632015 Chr.19: 44176676-44176676 on Build GRCh38
rs773608113 Chr.19: 44176694-44176694 on Build GRCh38
rs777393073 Chr.19: 44176762-44176762 on Build GRCh38
rs376102633 Chr.19: 44177081-44177081 on Build GRCh38
rs376553934 Chr.19: 44176980-44176980 on Build GRCh38
rs751439312 Chr.19: 44176948-44176948 on Build GRCh38
rs145998371 Chr.19: 44177014-44177014 on Build GRCh38
rs1046773862 Chr.19: 44176669-44176669 on Build GRCh38
rs771217901 Chr.19: 44176777-44176777 on Build GRCh38
rs764427753 Chr.19: 44176810-44176810 on Build GRCh38
rs377614893 Chr.19: 44177054-44177054 on Build GRCh38
rs764376913 Chr.19: 44176735-44176735 on Build GRCh38
rs774780465 Chr.19: 44176699-44176699 on Build GRCh38
rs752880087 Chr.19: 44176716-44176716 on Build GRCh38
rs61742481 Chr.19: 44176674-44176674 on Build GRCh38
rs116372639 Chr.19: 44176977-44176977 on Build GRCh38
rs767645265 Chr.19: 44176664-44176664 on Build GRCh38
rs377205758 Chr.19: 44176749-44176749 on Build GRCh38
rs766231791 Chr.19: 44176840-44176840 on Build GRCh38
rs778797352 Chr.19: 44176675-44176675 on Build GRCh38
rs370423984 Chr.19: 44177037-44177037 on Build GRCh38
rs770201923 Chr.19: 44177020-44177020 on Build GRCh38
rs755070113 Chr.19: 44176653-44176653 on Build GRCh38
rs755016120 Chr.19: 44176941-44176953 on Build GRCh38
rs1056674018 Chr.19: 44177095-44177095 on Build GRCh38
rs752311991 Chr.19: 44176862-44176862 on Build GRCh38
rs114353503 Chr.19: 44176837-44176837 on Build GRCh38
rs781638325 Chr.19: 44176683-44176686 on Build GRCh38
rs866792431 Chr.19: 44176854-44176854 on Build GRCh38
rs763305694 Chr.19: 44176695-44176695 on Build GRCh38
rs760651440 Chr.19: 44177034-44177034 on Build GRCh38
rs967865655 Chr.19: 44176861-44176861 on Build GRCh38
rs772104222 Chr.19: 44176855-44176855 on Build GRCh38
rs200493278 Chr.19: 44177099-44177099 on Build GRCh38
rs767505724 Chr.19: 44176959-44176959 on Build GRCh38
rs968032171 Chr.19: 44176916-44176916 on Build GRCh38
rs372362519 Chr.19: 44176779-44176779 on Build GRCh38
rs536017980 Chr.19: 44177007-44177007 on Build GRCh38
rs751707034 Chr.19: 44176645-44176645 on Build GRCh38
rs199870248 Chr.19: 44177032-44177032 on Build GRCh38
rs61742482 Chr.19: 44176682-44176682 on Build GRCh38
rs763987545 Chr.19: 44177036-44177036 on Build GRCh38
rs780988862 Chr.19: 44176850-44176850 on Build GRCh38
rs759377322 Chr.19: 44177024-44177024 on Build GRCh38
rs760039084 Chr.19: 44176795-44176795 on Build GRCh38
rs768819526 Chr.19: 44176921-44176921 on Build GRCh38
rs777405832 Chr.19: 44176871-44176871 on Build GRCh38
rs369575092 Chr.19: 44176934-44176934 on Build GRCh38
rs752507988 Chr.19: 44176963-44176963 on Build GRCh38
rs750307681 Chr.19: 44176740-44176740 on Build GRCh38
rs764954266 Chr.19: 44177041-44177041 on Build GRCh38
rs772432212 Chr.19: 44176798-44176798 on Build GRCh38
rs755650710 Chr.19: 44176968-44176968 on Build GRCh38
rs908844625 Chr.19: 44177018-44177018 on Build GRCh38
rs201119111 Chr.19: 44176691-44176691 on Build GRCh38
rs754050408 Chr.19: 44176834-44176834 on Build GRCh38
rs753367409 Chr.19: 44176840-44176840 on Build GRCh38
rs771307811 Chr.19: 44176889-44176889 on Build GRCh38
rs10413887 Chr.19: 44176843-44176843 on Build GRCh38
rs760816615 Chr.19: 44176913-44176913 on Build GRCh38
rs746299849 Chr.19: 44177000-44177000 on Build GRCh38
rs765525957 Chr.19: 44176944-44176944 on Build GRCh38
rs200558002 Chr.19: 44177008-44177008 on Build GRCh38
rs759539042 Chr.19: 44176707-44176707 on Build GRCh38
rs773252890 Chr.19: 44177023-44177023 on Build GRCh38
rs780129731 Chr.19: 44176679-44176679 on Build GRCh38
rs771159613 Chr.19: 44177106-44177106 on Build GRCh38
rs751478221 Chr.19: 44176747-44176747 on Build GRCh38
rs755562353 Chr.19: 44176867-44176867 on Build GRCh38
rs1030938826 Chr.19: 44176975-44176975 on Build GRCh38
rs368637456 Chr.19: 44177094-44177094 on Build GRCh38
rs756745955 Chr.19: 44177038-44177038 on Build GRCh38
rs747987463 Chr.19: 44177069-44177069 on Build GRCh38
rs775844795 Chr.19: 44176909-44176909 on Build GRCh38
rs772700276 Chr.19: 44176894-44176894 on Build GRCh38
rs902043343 Chr.19: 44176728-44176728 on Build GRCh38
rs755612852 Chr.19: 44177066-44177066 on Build GRCh38
rs981577820 Chr.19: 44176962-44176962 on Build GRCh38
rs368921321 Chr.19: 44176988-44176988 on Build GRCh38
rs749664996 Chr.19: 44176775-44176775 on Build GRCh38
rs779380157 Chr.19: 44176893-44176893 on Build GRCh38
rs759240596 Chr.19: 44176958-44176958 on Build GRCh38
rs754984072 Chr.19: 44176849-44176849 on Build GRCh38
rs778944458 Chr.19: 44177035-44177036 on Build GRCh38
rs777019835 Chr.19: 44176922-44176922 on Build GRCh38
rs747347532 Chr.19: 44177061-44177061 on Build GRCh38
rs749988341 Chr.19: 44176659-44176660 on Build GRCh38
rs187243325 Chr.19: 44176677-44176677 on Build GRCh38
rs769558345 Chr.19: 44176754-44176754 on Build GRCh38
rs544911012 Chr.19: 44177091-44177091 on Build GRCh38
rs374827802 Chr.19: 44177046-44177046 on Build GRCh38
rs753319006 Chr.19: 44176979-44176979 on Build GRCh38
rs1000710744 Chr.19: 44176926-44176926 on Build GRCh38
rs746534261 Chr.19: 44176688-44176688 on Build GRCh38
rs762179935 Chr.19: 44176929-44176929 on Build GRCh38
rs752896775 Chr.19: 44176672-44176672 on Build GRCh38
rs187489302 Chr.19: 44176661-44176661 on Build GRCh38
rs770380297 Chr.19: 44177028-44177028 on Build GRCh38
rs766928159 Chr.19: 44176697-44176697 on Build GRCh38

View Full Product Details