Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46605675-46606169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM3
Entrez Gene ID
808
Gene Name
calmodulin 3
Gene Aliases
HEL-S-72, PHKD, PHKD3
UniGene
Hs.515487
Gene Chromosome Location
Chr.19: 46601255-46610782 on Build GRCh38
Transcripts
NM_005184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
459921 c.27G>C p.Q9H Chr.19: 46605850-46605850 on Build GRCh38
5718213 c.30T>G p.I10M Chr.19: 46605853-46605853 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs529690199 Chr.19: 46606039-46606039 on Build GRCh38
rs941832333 Chr.19: 46606017-46606017 on Build GRCh38
rs1015780810 Chr.19: 46606135-46606135 on Build GRCh38
rs919089898 Chr.19: 46606131-46606140 on Build GRCh38
rs765275273 Chr.19: 46605881-46605881 on Build GRCh38
rs772361208 Chr.19: 46605777-46605777 on Build GRCh38
rs917862214 Chr.19: 46605947-46605947 on Build GRCh38
rs1033021816 Chr.19: 46605764-46605764 on Build GRCh38
rs1042190380 Chr.19: 46605714-46605714 on Build GRCh38
rs949277469 Chr.19: 46605986-46605986 on Build GRCh38
rs752528019 Chr.19: 46605802-46605802 on Build GRCh38
rs192607690 Chr.19: 46605912-46605912 on Build GRCh38
rs773339957 Chr.19: 46605886-46605886 on Build GRCh38
rs556811467 Chr.19: 46605780-46605780 on Build GRCh38
rs1000985622 Chr.19: 46606143-46606143 on Build GRCh38
rs764310978 Chr.19: 46605782-46605782 on Build GRCh38
rs903290860 Chr.19: 46605753-46605753 on Build GRCh38
rs1028296904 Chr.19: 46606100-46606100 on Build GRCh38
rs768965117 Chr.19: 46605870-46605870 on Build GRCh38
rs777256869 Chr.19: 46605706-46605706 on Build GRCh38
rs762004724 Chr.19: 46605875-46605875 on Build GRCh38
rs970900898 Chr.19: 46605976-46605976 on Build GRCh38
rs1054986629 Chr.19: 46606040-46606040 on Build GRCh38
rs756875833 Chr.19: 46605824-46605824 on Build GRCh38
rs573935530 Chr.19: 46605749-46605749 on Build GRCh38
rs3729510 Chr.19: 46605754-46605754 on Build GRCh38
rs893201674 Chr.19: 46606051-46606051 on Build GRCh38
rs748909624 Chr.19: 46605815-46605815 on Build GRCh38
rs776863863 Chr.19: 46605873-46605873 on Build GRCh38
rs549848361 Chr.19: 46606142-46606142 on Build GRCh38
rs778455345 Chr.19: 46605830-46605830 on Build GRCh38
rs760941952 Chr.19: 46605781-46605781 on Build GRCh38
rs541090118 Chr.19: 46605933-46605933 on Build GRCh38
rs1042221298 Chr.19: 46606004-46606004 on Build GRCh38
rs919680694 Chr.19: 46605902-46605902 on Build GRCh38
rs752615570 Chr.19: 46605891-46605891 on Build GRCh38
rs370743010 Chr.19: 46606007-46606007 on Build GRCh38
rs3729511 Chr.19: 46605894-46605894 on Build GRCh38
rs761908881 Chr.19: 46605789-46605789 on Build GRCh38
rs572190013 Chr.19: 46605917-46605917 on Build GRCh38
rs371227609 Chr.19: 46605919-46605919 on Build GRCh38
rs112165289 Chr.19: 46605712-46605712 on Build GRCh38
rs747374745 Chr.19: 46605869-46605869 on Build GRCh38
rs776763027 Chr.19: 46605786-46605786 on Build GRCh38
rs775799940 Chr.19: 46605860-46605860 on Build GRCh38
rs767285476 Chr.19: 46605798-46605798 on Build GRCh38
rs188865816 Chr.19: 46605790-46605790 on Build GRCh38
rs759451253 Chr.19: 46605887-46605887 on Build GRCh38
rs990428032 Chr.19: 46605925-46605925 on Build GRCh38
rs769659778 Chr.19: 46606062-46606062 on Build GRCh38
rs1037658738 Chr.19: 46606033-46606033 on Build GRCh38
rs936509014 Chr.19: 46606018-46606018 on Build GRCh38
rs370333318 Chr.19: 46605810-46605810 on Build GRCh38
rs961879840 Chr.19: 46606141-46606141 on Build GRCh38
rs754915710 Chr.19: 46605792-46605792 on Build GRCh38
rs375340678 Chr.19: 46606138-46606138 on Build GRCh38
rs1001080500 Chr.19: 46605847-46605847 on Build GRCh38
rs996844874 Chr.19: 46605772-46605772 on Build GRCh38
rs746427314 Chr.19: 46605835-46605835 on Build GRCh38
rs1008444147 Chr.19: 46606124-46606124 on Build GRCh38
rs983182164 Chr.19: 46605981-46605981 on Build GRCh38
rs756111967 Chr.19: 46605776-46605776 on Build GRCh38
rs891400838 Chr.19: 46606101-46606101 on Build GRCh38
rs553765441 Chr.19: 46605707-46605707 on Build GRCh38
rs1037588711 Chr.19: 46605740-46605740 on Build GRCh38
rs772618542 Chr.19: 46605852-46605852 on Build GRCh38
rs767505624 Chr.19: 46605890-46605890 on Build GRCh38
rs997282949 Chr.19: 46606055-46606055 on Build GRCh38

View Full Product Details