Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46620804-46621296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGIR
Entrez Gene ID
5739
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
Gene Aliases
IP, PRIPR
UniGene
Hs.458324
Gene Chromosome Location
Chr.19: 46610040-46625118 on Build GRCh38
Transcripts
NM_000960.3 XM_005259093.2 XM_005259095.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4978475 c.1160G>C p.*387S Chr.19: 46621281-46621281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900732966 Chr.19: 46621113-46621113 on Build GRCh38
rs983585479 Chr.19: 46620877-46620877 on Build GRCh38
rs201926050 Chr.19: 46620914-46620914 on Build GRCh38
rs758348089 Chr.19: 46621263-46621263 on Build GRCh38
rs201166212 Chr.19: 46620853-46620853 on Build GRCh38
rs201416097 Chr.19: 46621265-46621265 on Build GRCh38
rs760404101 Chr.19: 46620893-46620893 on Build GRCh38
rs937175268 Chr.19: 46620986-46620986 on Build GRCh38
rs373443618 Chr.19: 46621142-46621142 on Build GRCh38
rs769627504 Chr.19: 46620913-46620913 on Build GRCh38
rs200293051 Chr.19: 46621251-46621251 on Build GRCh38
rs375214182 Chr.19: 46621266-46621266 on Build GRCh38
rs936172709 Chr.19: 46620999-46620999 on Build GRCh38
rs977903067 Chr.19: 46620990-46620990 on Build GRCh38
rs916174476 Chr.19: 46621009-46621009 on Build GRCh38
rs1032576158 Chr.19: 46621104-46621104 on Build GRCh38
rs1056995016 Chr.19: 46621037-46621037 on Build GRCh38
rs546717509 Chr.19: 46621199-46621199 on Build GRCh38
rs926026520 Chr.19: 46620998-46620998 on Build GRCh38
rs774774728 Chr.19: 46621231-46621231 on Build GRCh38
rs184926891 Chr.19: 46621008-46621008 on Build GRCh38
rs199887129 Chr.19: 46621172-46621172 on Build GRCh38
rs754755781 Chr.19: 46621272-46621272 on Build GRCh38
rs957446189 Chr.19: 46621240-46621240 on Build GRCh38
rs201005621 Chr.19: 46621091-46621091 on Build GRCh38
rs145573729 Chr.19: 46621241-46621241 on Build GRCh38
rs199842130 Chr.19: 46621045-46621045 on Build GRCh38
rs368348673 Chr.19: 46621268-46621268 on Build GRCh38
rs996816582 Chr.19: 46621118-46621118 on Build GRCh38
rs200548857 Chr.19: 46620922-46620922 on Build GRCh38
rs1016807013 Chr.19: 46620859-46620859 on Build GRCh38
rs923024484 Chr.19: 46620844-46620844 on Build GRCh38
rs201083449 Chr.19: 46621238-46621238 on Build GRCh38
rs112295993 Chr.19: 46621153-46621153 on Build GRCh38
rs199560174 Chr.19: 46621086-46621086 on Build GRCh38
rs36029279 Chr.19: 46621138-46621138 on Build GRCh38
rs201506935 Chr.19: 46621084-46621084 on Build GRCh38
rs926933951 Chr.19: 46621213-46621213 on Build GRCh38
rs201276283 Chr.19: 46621044-46621044 on Build GRCh38
rs141412300 Chr.19: 46620981-46620981 on Build GRCh38
rs144126196 Chr.19: 46620857-46620857 on Build GRCh38
rs1055946431 Chr.19: 46621001-46621001 on Build GRCh38
rs201617405 Chr.19: 46621140-46621140 on Build GRCh38
rs993151773 Chr.19: 46621222-46621222 on Build GRCh38
rs201806865 Chr.19: 46621100-46621100 on Build GRCh38
rs973323692 Chr.19: 46620929-46620929 on Build GRCh38
rs201045216 Chr.19: 46621155-46621155 on Build GRCh38
rs202126416 Chr.19: 46620894-46620894 on Build GRCh38
rs745492771 Chr.19: 46620903-46620903 on Build GRCh38
rs201523536 Chr.19: 46621246-46621246 on Build GRCh38
rs578102191 Chr.19: 46620939-46620939 on Build GRCh38
rs200210797 Chr.19: 46621005-46621005 on Build GRCh38
rs200095263 Chr.19: 46621143-46621143 on Build GRCh38
rs199947202 Chr.19: 46621129-46621129 on Build GRCh38
rs200594965 Chr.19: 46621131-46621131 on Build GRCh38
rs4987264 Chr.19: 46620833-46620834 on Build GRCh38
rs201505166 Chr.19: 46620944-46620944 on Build GRCh38
rs955246213 Chr.19: 46620915-46620915 on Build GRCh38
rs200303055 Chr.19: 46621218-46621218 on Build GRCh38
rs200589666 Chr.19: 46621069-46621069 on Build GRCh38
rs200975566 Chr.19: 46620901-46620901 on Build GRCh38
rs199984361 Chr.19: 46620895-46620895 on Build GRCh38
rs576291772 Chr.19: 46621137-46621137 on Build GRCh38
rs10422782 Chr.19: 46621217-46621217 on Build GRCh38
rs1003999021 Chr.19: 46621130-46621130 on Build GRCh38
rs73940735 Chr.19: 46621200-46621200 on Build GRCh38
rs200395250 Chr.19: 46621243-46621243 on Build GRCh38
rs775649266 Chr.19: 46621242-46621242 on Build GRCh38
rs199619505 Chr.19: 46621248-46621248 on Build GRCh38
rs987497976 Chr.19: 46620948-46620948 on Build GRCh38
rs202199705 Chr.19: 46621165-46621165 on Build GRCh38
rs887153996 Chr.19: 46620864-46620864 on Build GRCh38
rs1000784657 Chr.19: 46621092-46621092 on Build GRCh38
rs926051506 Chr.19: 46621239-46621239 on Build GRCh38

View Full Product Details