Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48498036-48498527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMTK3
Entrez Gene ID
114783
Gene Name
lemur tyrosine kinase 3
Gene Aliases
LMR3, PPP1R101, TYKLM3
UniGene
Hs.207426
Gene Chromosome Location
Chr.19: 48485271-48513926 on Build GRCh38
Transcripts
NM_001080434.1 XM_011526411.2 XM_011526412.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999000 c.2664G>A p.T888T Chr.19: 48498492-48498492 on Build GRCh38
4540008 c.2771G>A p.G924E Chr.19: 48498385-48498385 on Build GRCh38
1564332 c.2916G>A p.A972A Chr.19: 48498240-48498240 on Build GRCh38
998999 c.3037G>A p.A1013T Chr.19: 48498119-48498119 on Build GRCh38
3893002 c.2883G>A p.G961G Chr.19: 48498273-48498273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370333957 Chr.19: 48498357-48498357 on Build GRCh38
rs374685950 Chr.19: 48498366-48498366 on Build GRCh38
rs776969963 Chr.19: 48498332-48498332 on Build GRCh38
rs768491589 Chr.19: 48498364-48498364 on Build GRCh38
rs759608746 Chr.19: 48498090-48498090 on Build GRCh38
rs35275345 Chr.19: 48498431-48498431 on Build GRCh38
rs752185124 Chr.19: 48498424-48498426 on Build GRCh38
rs1006876729 Chr.19: 48498446-48498446 on Build GRCh38
rs370560246 Chr.19: 48498187-48498187 on Build GRCh38
rs746156771 Chr.19: 48498184-48498184 on Build GRCh38
rs767476481 Chr.19: 48498360-48498360 on Build GRCh38
rs919457074 Chr.19: 48498283-48498283 on Build GRCh38
rs201586464 Chr.19: 48498199-48498199 on Build GRCh38
rs892309802 Chr.19: 48498367-48498367 on Build GRCh38
rs753390537 Chr.19: 48498113-48498113 on Build GRCh38
rs779113531 Chr.19: 48498285-48498285 on Build GRCh38
rs747172279 Chr.19: 48498321-48498321 on Build GRCh38
rs370902571 Chr.19: 48498449-48498449 on Build GRCh38
rs201444727 Chr.19: 48498174-48498174 on Build GRCh38
rs757975747 Chr.19: 48498151-48498151 on Build GRCh38
rs748440723 Chr.19: 48498421-48498421 on Build GRCh38
rs61743326 Chr.19: 48498288-48498288 on Build GRCh38
rs767935576 Chr.19: 48498092-48498092 on Build GRCh38
rs923126427 Chr.19: 48498342-48498342 on Build GRCh38
rs777131416 Chr.19: 48498220-48498220 on Build GRCh38
rs755191134 Chr.19: 48498391-48498391 on Build GRCh38
rs775701669 Chr.19: 48498107-48498107 on Build GRCh38
rs562441963 Chr.19: 48498309-48498309 on Build GRCh38
rs781066186 Chr.19: 48498259-48498259 on Build GRCh38
rs1799285 Chr.19: 48498270-48498270 on Build GRCh38
rs866593260 Chr.19: 48498115-48498115 on Build GRCh38
rs377699902 Chr.19: 48498351-48498351 on Build GRCh38
rs952680573 Chr.19: 48498268-48498268 on Build GRCh38
rs764833476 Chr.19: 48498156-48498158 on Build GRCh38
rs754851655 Chr.19: 48498249-48498249 on Build GRCh38
rs373942106 Chr.19: 48498493-48498493 on Build GRCh38
rs201904181 Chr.19: 48498116-48498116 on Build GRCh38
rs1057278322 Chr.19: 48498410-48498410 on Build GRCh38
rs867245025 Chr.19: 48498263-48498263 on Build GRCh38
rs781578205 Chr.19: 48498413-48498413 on Build GRCh38
rs780055149 Chr.19: 48498386-48498386 on Build GRCh38
rs750107495 Chr.19: 48498150-48498150 on Build GRCh38
rs763906029 Chr.19: 48498476-48498476 on Build GRCh38
rs374953584 Chr.19: 48498221-48498221 on Build GRCh38
rs749898421 Chr.19: 48498237-48498237 on Build GRCh38
rs755244371 Chr.19: 48498500-48498500 on Build GRCh38
rs773467354 Chr.19: 48498063-48498063 on Build GRCh38
rs756795214 Chr.19: 48498114-48498114 on Build GRCh38
rs781416673 Chr.19: 48498506-48498506 on Build GRCh38
rs759327551 Chr.19: 48498430-48498430 on Build GRCh38
rs753737623 Chr.19: 48498477-48498477 on Build GRCh38
rs751260723 Chr.19: 48498354-48498354 on Build GRCh38
rs866613109 Chr.19: 48498167-48498167 on Build GRCh38
rs751370624 Chr.19: 48498161-48498161 on Build GRCh38
rs781301076 Chr.19: 48498171-48498171 on Build GRCh38
rs576612975 Chr.19: 48498284-48498284 on Build GRCh38
rs1643478 Chr.19: 48498371-48498371 on Build GRCh38
rs748362287 Chr.19: 48498507-48498507 on Build GRCh38
rs572849420 Chr.19: 48498135-48498135 on Build GRCh38
rs762708061 Chr.19: 48498238-48498238 on Build GRCh38
rs61745738 Chr.19: 48498343-48498343 on Build GRCh38
rs764550106 Chr.19: 48498111-48498111 on Build GRCh38
rs747292842 Chr.19: 48498387-48498387 on Build GRCh38
rs368345403 Chr.19: 48498083-48498083 on Build GRCh38
rs774664240 Chr.19: 48498072-48498072 on Build GRCh38
rs759205143 Chr.19: 48498356-48498356 on Build GRCh38
rs773623410 Chr.19: 48498424-48498424 on Build GRCh38
rs770867017 Chr.19: 48498071-48498071 on Build GRCh38
rs748057251 Chr.19: 48498173-48498173 on Build GRCh38
rs372400709 Chr.19: 48498219-48498219 on Build GRCh38
rs772177146 Chr.19: 48498198-48498198 on Build GRCh38
rs200616585 Chr.19: 48498422-48498422 on Build GRCh38
rs905648442 Chr.19: 48498120-48498120 on Build GRCh38
rs760405879 Chr.19: 48498469-48498469 on Build GRCh38
rs762452338 Chr.19: 48498333-48498333 on Build GRCh38
rs761001211 Chr.19: 48498212-48498212 on Build GRCh38
rs764862461 Chr.19: 48498225-48498225 on Build GRCh38
rs749357838 Chr.19: 48498066-48498066 on Build GRCh38
rs965802553 Chr.19: 48498235-48498235 on Build GRCh38
rs375332014 Chr.19: 48498177-48498177 on Build GRCh38
rs866156335 Chr.19: 48498257-48498257 on Build GRCh38
rs374555928 Chr.19: 48498348-48498348 on Build GRCh38
rs749736250 Chr.19: 48498425-48498425 on Build GRCh38
rs868535969 Chr.19: 48498337-48498337 on Build GRCh38
rs755016125 Chr.19: 48498163-48498163 on Build GRCh38
rs1022607132 Chr.19: 48498102-48498102 on Build GRCh38
rs769026208 Chr.19: 48498326-48498326 on Build GRCh38
rs200612607 Chr.19: 48498429-48498429 on Build GRCh38
rs757177302 Chr.19: 48498492-48498492 on Build GRCh38
rs762173176 Chr.19: 48498222-48498222 on Build GRCh38
rs762392357 Chr.19: 48498349-48498349 on Build GRCh38
rs201144422 Chr.19: 48498297-48498297 on Build GRCh38
rs775400823 Chr.19: 48498465-48498465 on Build GRCh38
rs916108288 Chr.19: 48498311-48498311 on Build GRCh38
rs765926338 Chr.19: 48498241-48498241 on Build GRCh38
rs779968314 Chr.19: 48498159-48498159 on Build GRCh38
rs377391186 Chr.19: 48498336-48498336 on Build GRCh38
rs751209205 Chr.19: 48498248-48498248 on Build GRCh38

View Full Product Details