Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48635451-48635995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBP
Entrez Gene ID
1628
Gene Name
D-box binding PAR bZIP transcription factor
Gene Aliases
DABP
UniGene
Hs.414480
Gene Chromosome Location
Chr.19: 48630560-48637550 on Build GRCh38
Transcripts
NM_001352.4 XM_017026388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408743 c.511G>A p.A171T Chr.19: 48635619-48635619 on Build GRCh38
6402283 c.448C>T p.P150S Chr.19: 48635682-48635682 on Build GRCh38
5620623 c.475T>C p.S159P Chr.19: 48635655-48635655 on Build GRCh38
6401051 c.349G>A p.V117I Chr.19: 48635781-48635781 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202083967 Chr.19: 48635918-48635918 on Build GRCh38
rs1037720762 Chr.19: 48635674-48635674 on Build GRCh38
rs537199001 Chr.19: 48635619-48635619 on Build GRCh38
rs1005040641 Chr.19: 48635483-48635483 on Build GRCh38
rs1037036818 Chr.19: 48635565-48635565 on Build GRCh38
rs975437284 Chr.19: 48635903-48635903 on Build GRCh38
rs774216657 Chr.19: 48635559-48635559 on Build GRCh38
rs780543551 Chr.19: 48635935-48635935 on Build GRCh38
rs1037481506 Chr.19: 48635689-48635689 on Build GRCh38
rs915541410 Chr.19: 48635851-48635851 on Build GRCh38
rs1049761952 Chr.19: 48635953-48635953 on Build GRCh38
rs1000184091 Chr.19: 48635832-48635832 on Build GRCh38
rs898529606 Chr.19: 48635592-48635592 on Build GRCh38
rs1015997218 Chr.19: 48635490-48635490 on Build GRCh38
rs907477620 Chr.19: 48635549-48635549 on Build GRCh38
rs1049732523 Chr.19: 48635717-48635717 on Build GRCh38
rs780706474 Chr.19: 48635900-48635900 on Build GRCh38
rs1033098129 Chr.19: 48635853-48635853 on Build GRCh38
rs1055964875 Chr.19: 48635698-48635698 on Build GRCh38
rs977124329 Chr.19: 48635506-48635506 on Build GRCh38
rs1018756855 Chr.19: 48635616-48635620 on Build GRCh38
rs758773876 Chr.19: 48635899-48635899 on Build GRCh38
rs895951153 Chr.19: 48635705-48635705 on Build GRCh38
rs965904291 Chr.19: 48635800-48635800 on Build GRCh38
rs990236284 Chr.19: 48635848-48635848 on Build GRCh38
rs201353940 Chr.19: 48635971-48635971 on Build GRCh38
rs747873014 Chr.19: 48635937-48635937 on Build GRCh38
rs972906148 Chr.19: 48635929-48635929 on Build GRCh38
rs899342651 Chr.19: 48635528-48635528 on Build GRCh38
rs1045881899 Chr.19: 48635659-48635659 on Build GRCh38
rs967382623 Chr.19: 48635569-48635569 on Build GRCh38
rs889883821 Chr.19: 48635725-48635725 on Build GRCh38
rs764122816 Chr.19: 48635564-48635564 on Build GRCh38
rs977783244 Chr.19: 48635628-48635628 on Build GRCh38
rs1051649627 Chr.19: 48635662-48635662 on Build GRCh38
rs1028852911 Chr.19: 48635599-48635599 on Build GRCh38
rs749095357 Chr.19: 48635963-48635963 on Build GRCh38
rs866905766 Chr.19: 48635930-48635930 on Build GRCh38
rs777681993 Chr.19: 48635958-48635958 on Build GRCh38
rs924017539 Chr.19: 48635555-48635555 on Build GRCh38
rs754557287 Chr.19: 48635931-48635931 on Build GRCh38
rs981195309 Chr.19: 48635869-48635869 on Build GRCh38
rs570153198 Chr.19: 48635598-48635598 on Build GRCh38
rs567853417 Chr.19: 48635666-48635666 on Build GRCh38
rs934030111 Chr.19: 48635642-48635642 on Build GRCh38
rs528598284 Chr.19: 48635952-48635952 on Build GRCh38
rs889832010 Chr.19: 48635954-48635954 on Build GRCh38
rs1001687773 Chr.19: 48635533-48635533 on Build GRCh38
rs940338572 Chr.19: 48635686-48635686 on Build GRCh38
rs998686655 Chr.19: 48635814-48635814 on Build GRCh38
rs957288095 Chr.19: 48635844-48635844 on Build GRCh38
rs944014826 Chr.19: 48635962-48635962 on Build GRCh38
rs1034624054 Chr.19: 48635544-48635544 on Build GRCh38
rs1054647793 Chr.19: 48635524-48635524 on Build GRCh38
rs926026724 Chr.19: 48635604-48635604 on Build GRCh38
rs760752026 Chr.19: 48635561-48635561 on Build GRCh38
rs921773516 Chr.19: 48635948-48635948 on Build GRCh38
rs1014289799 Chr.19: 48635556-48635556 on Build GRCh38
rs924546235 Chr.19: 48635639-48635639 on Build GRCh38
rs954543216 Chr.19: 48635961-48635961 on Build GRCh38
rs898895323 Chr.19: 48635692-48635692 on Build GRCh38
rs754115103 Chr.19: 48635600-48635600 on Build GRCh38
rs1051028143 Chr.19: 48635648-48635648 on Build GRCh38
rs770650040 Chr.19: 48635970-48635970 on Build GRCh38
rs1014923201 Chr.19: 48635730-48635730 on Build GRCh38
rs924234645 Chr.19: 48635512-48635512 on Build GRCh38
rs960276232 Chr.19: 48635553-48635553 on Build GRCh38
rs957889583 Chr.19: 48635611-48635611 on Build GRCh38
rs769328103 Chr.19: 48635943-48635943 on Build GRCh38
rs867470520 Chr.19: 48635934-48635934 on Build GRCh38
rs915649185 Chr.19: 48635530-48635530 on Build GRCh38
rs535385622 Chr.19: 48635733-48635733 on Build GRCh38
rs921890064 Chr.19: 48635647-48635647 on Build GRCh38
rs928380809 Chr.19: 48635873-48635873 on Build GRCh38
rs935680669 Chr.19: 48635521-48635521 on Build GRCh38
rs752612550 Chr.19: 48635537-48635538 on Build GRCh38
rs1001741857 Chr.19: 48635693-48635693 on Build GRCh38
rs979269508 Chr.19: 48635576-48635576 on Build GRCh38
rs995509059 Chr.19: 48635595-48635595 on Build GRCh38
rs954090605 Chr.19: 48635607-48635607 on Build GRCh38
rs948445742 Chr.19: 48635658-48635658 on Build GRCh38
rs1020506906 Chr.19: 48635734-48635734 on Build GRCh38
rs752072096 Chr.19: 48635921-48635921 on Build GRCh38
rs750736355 Chr.19: 48635801-48635801 on Build GRCh38
rs940210015 Chr.19: 48635919-48635919 on Build GRCh38
rs933243111 Chr.19: 48635653-48635653 on Build GRCh38
rs770842215 Chr.19: 48635480-48635480 on Build GRCh38
rs112887941 Chr.19: 48635740-48635740 on Build GRCh38
rs555937572 Chr.19: 48635643-48635643 on Build GRCh38
rs967813313 Chr.19: 48635746-48635746 on Build GRCh38

View Full Product Details