Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49556437-49556929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOSIP
Entrez Gene ID
51070
Gene Name
nitric oxide synthase interacting protein
Gene Aliases
CGI-25
UniGene
Hs.7236
Gene Chromosome Location
Chr.19: 49555468-49580599 on Build GRCh38
Transcripts
NM_001270960.1 NM_015953.4 XM_005258964.4 XM_006723235.3 XM_011527015.2 XM_011527017.2 XM_017026851.1 XM_017026852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110814 c.633C>T p.R211R Chr.19: 49556641-49556641 on Build GRCh38
5703190 c.713delT p.V238fs*17 Chr.19: 49556561-49556561 on Build GRCh38
7262733 c.708C>T p.C236C Chr.19: 49556566-49556566 on Build GRCh38
5614695 c.538-1G>T p.? Chr.19: 49556737-49556737 on Build GRCh38
1681215 c.650G>A p.R217H Chr.19: 49556624-49556624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781734698 Chr.19: 49556857-49556857 on Build GRCh38
rs760457647 Chr.19: 49556784-49556784 on Build GRCh38
rs376400410 Chr.19: 49556729-49556729 on Build GRCh38
rs758374592 Chr.19: 49556539-49556539 on Build GRCh38
rs779180325 Chr.19: 49556828-49556835 on Build GRCh38
rs971092767 Chr.19: 49556681-49556681 on Build GRCh38
rs772418014 Chr.19: 49556508-49556508 on Build GRCh38
rs551642728 Chr.19: 49556817-49556817 on Build GRCh38
rs745886518 Chr.19: 49556578-49556578 on Build GRCh38
rs957992254 Chr.19: 49556888-49556888 on Build GRCh38
rs898831268 Chr.19: 49556751-49556751 on Build GRCh38
rs865863056 Chr.19: 49556663-49556663 on Build GRCh38
rs775855155 Chr.19: 49556588-49556588 on Build GRCh38
rs756619279 Chr.19: 49556538-49556539 on Build GRCh38
rs370045117 Chr.19: 49556865-49556865 on Build GRCh38
rs781185665 Chr.19: 49556504-49556505 on Build GRCh38
rs924569928 Chr.19: 49556694-49556694 on Build GRCh38
rs748461813 Chr.19: 49556717-49556717 on Build GRCh38
rs781518095 Chr.19: 49556555-49556555 on Build GRCh38
rs747462873 Chr.19: 49556592-49556592 on Build GRCh38
rs774388893 Chr.19: 49556756-49556756 on Build GRCh38
rs755552712 Chr.19: 49556855-49556855 on Build GRCh38
rs759426411 Chr.19: 49556619-49556619 on Build GRCh38
rs578126932 Chr.19: 49556493-49556493 on Build GRCh38
rs1028904359 Chr.19: 49556479-49556479 on Build GRCh38
rs368759917 Chr.19: 49556514-49556514 on Build GRCh38
rs758542969 Chr.19: 49556844-49556844 on Build GRCh38
rs769578558 Chr.19: 49556723-49556723 on Build GRCh38
rs761155161 Chr.19: 49556820-49556820 on Build GRCh38
rs369839191 Chr.19: 49556594-49556594 on Build GRCh38
rs751378014 Chr.19: 49556548-49556548 on Build GRCh38
rs770273311 Chr.19: 49556867-49556867 on Build GRCh38
rs1015190269 Chr.19: 49556566-49556566 on Build GRCh38
rs773758100 Chr.19: 49556881-49556881 on Build GRCh38
rs770579527 Chr.19: 49556875-49556875 on Build GRCh38
rs1009253752 Chr.19: 49556834-49556834 on Build GRCh38
rs751605503 Chr.19: 49556853-49556853 on Build GRCh38
rs767271460 Chr.19: 49556625-49556625 on Build GRCh38
rs769242681 Chr.19: 49556702-49556702 on Build GRCh38
rs6509437 Chr.19: 49556461-49556461 on Build GRCh38
rs772637224 Chr.19: 49556579-49556579 on Build GRCh38
rs765551872 Chr.19: 49556824-49556824 on Build GRCh38
rs372106102 Chr.19: 49556704-49556704 on Build GRCh38
rs890413166 Chr.19: 49556743-49556743 on Build GRCh38
rs752451382 Chr.19: 49556627-49556627 on Build GRCh38
rs141077974 Chr.19: 49556714-49556714 on Build GRCh38
rs544641789 Chr.19: 49556700-49556700 on Build GRCh38
rs747441040 Chr.19: 49556690-49556690 on Build GRCh38
rs780658210 Chr.19: 49556677-49556677 on Build GRCh38
rs574062883 Chr.19: 49556460-49556460 on Build GRCh38
rs748863883 Chr.19: 49556874-49556874 on Build GRCh38
rs1050277018 Chr.19: 49556810-49556812 on Build GRCh38
rs780026225 Chr.19: 49556846-49556846 on Build GRCh38
rs746981590 Chr.19: 49556878-49556878 on Build GRCh38
rs766173976 Chr.19: 49556544-49556544 on Build GRCh38
rs1020103040 Chr.19: 49556843-49556843 on Build GRCh38
rs749742910 Chr.19: 49556748-49556751 on Build GRCh38
rs752502677 Chr.19: 49556780-49556780 on Build GRCh38
rs774886527 Chr.19: 49556499-49556499 on Build GRCh38
rs764812182 Chr.19: 49556532-49556532 on Build GRCh38
rs890474969 Chr.19: 49556482-49556482 on Build GRCh38
rs776411101 Chr.19: 49556521-49556521 on Build GRCh38
rs779040890 Chr.19: 49556647-49556647 on Build GRCh38
rs750587774 Chr.19: 49556830-49556830 on Build GRCh38
rs528844653 Chr.19: 49556664-49556664 on Build GRCh38
rs545540288 Chr.19: 49556495-49556495 on Build GRCh38
rs200454878 Chr.19: 49556557-49556557 on Build GRCh38
rs376440318 Chr.19: 49556662-49556662 on Build GRCh38
rs201833397 Chr.19: 49556572-49556572 on Build GRCh38
rs763906860 Chr.19: 49556520-49556520 on Build GRCh38
rs569864356 Chr.19: 49556845-49556845 on Build GRCh38
rs1027799623 Chr.19: 49556829-49556829 on Build GRCh38
rs770327606 Chr.19: 49556732-49556732 on Build GRCh38
rs761460943 Chr.19: 49556523-49556523 on Build GRCh38
rs769018498 Chr.19: 49556748-49556749 on Build GRCh38
rs866283704 Chr.19: 49556596-49556596 on Build GRCh38
rs757369043 Chr.19: 49556571-49556571 on Build GRCh38
rs60259512 Chr.19: 49556872-49556872 on Build GRCh38
rs754728597 Chr.19: 49556553-49556553 on Build GRCh38
rs1008745775 Chr.19: 49556565-49556565 on Build GRCh38
rs534514457 Chr.19: 49556808-49556809 on Build GRCh38
rs376009859 Chr.19: 49556476-49556476 on Build GRCh38
rs757493593 Chr.19: 49556634-49556634 on Build GRCh38
rs750493022 Chr.19: 49556525-49556526 on Build GRCh38
rs759502889 Chr.19: 49556774-49556774 on Build GRCh38
rs757495143 Chr.19: 49556499-49556500 on Build GRCh38
rs749909012 Chr.19: 49556535-49556535 on Build GRCh38
rs1036610890 Chr.19: 49556810-49556810 on Build GRCh38
rs764930641 Chr.19: 49556597-49556597 on Build GRCh38
rs991270581 Chr.19: 49556656-49556656 on Build GRCh38
rs147853428 Chr.19: 49556602-49556602 on Build GRCh38
rs772788289 Chr.19: 49556601-49556601 on Build GRCh38
rs4594362 Chr.19: 49556725-49556725 on Build GRCh38
rs17850728 Chr.19: 49556909-49556909 on Build GRCh38
rs186237221 Chr.19: 49556465-49556465 on Build GRCh38
rs780566627 Chr.19: 49556539-49556540 on Build GRCh38
rs966801837 Chr.19: 49556581-49556581 on Build GRCh38
rs751441811 Chr.19: 49556610-49556610 on Build GRCh38
rs757747143 Chr.19: 49556676-49556676 on Build GRCh38
rs767394385 Chr.19: 49556779-49556779 on Build GRCh38
rs34789987 Chr.19: 49556689-49556690 on Build GRCh38
rs746353105 Chr.19: 49556506-49556506 on Build GRCh38
rs776460161 Chr.19: 49556593-49556593 on Build GRCh38
rs958721605 Chr.19: 49556611-49556611 on Build GRCh38
rs751845796 Chr.19: 49556471-49556471 on Build GRCh38
rs749491556 Chr.19: 49556570-49556570 on Build GRCh38
rs989970099 Chr.19: 49556708-49556708 on Build GRCh38

View Full Product Details