Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50418840-50419338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIB
Entrez Gene ID
6689
Gene Name
Spi-B transcription factor
Gene Aliases
SPI-B
UniGene
Hs.437905
Gene Chromosome Location
Chr.19: 50418938-50431053 on Build GRCh38
Transcripts
NM_001243998.1 NM_001243999.1 NM_001244000.1 NM_003121.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6247729 c.7G>T p.A3S Chr.19: 50418969-50418969 on Build GRCh38
999717 c.23_23+3delAGTA p.? Chr.19: 50418985-50418988 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761958052 Chr.19: 50419008-50419008 on Build GRCh38
rs890800285 Chr.19: 50419227-50419227 on Build GRCh38
rs1032135300 Chr.19: 50419115-50419115 on Build GRCh38
rs550062106 Chr.19: 50418887-50418887 on Build GRCh38
rs1054186186 Chr.19: 50419124-50419124 on Build GRCh38
rs971434929 Chr.19: 50419304-50419304 on Build GRCh38
rs966560350 Chr.19: 50419009-50419009 on Build GRCh38
rs769164655 Chr.19: 50418949-50418949 on Build GRCh38
rs981729344 Chr.19: 50419172-50419172 on Build GRCh38
rs565413138 Chr.19: 50418872-50418872 on Build GRCh38
rs568327106 Chr.19: 50418924-50418924 on Build GRCh38
rs185436158 Chr.19: 50419019-50419019 on Build GRCh38
rs373382659 Chr.19: 50419014-50419014 on Build GRCh38
rs746964256 Chr.19: 50418937-50418937 on Build GRCh38
rs764093238 Chr.19: 50418920-50418920 on Build GRCh38
rs765181026 Chr.19: 50419015-50419015 on Build GRCh38
rs770546659 Chr.19: 50419257-50419260 on Build GRCh38
rs755290861 Chr.19: 50418983-50418983 on Build GRCh38
rs779231259 Chr.19: 50418932-50418932 on Build GRCh38
rs958719144 Chr.19: 50419080-50419080 on Build GRCh38
rs977343916 Chr.19: 50419106-50419106 on Build GRCh38
rs1008216763 Chr.19: 50419098-50419098 on Build GRCh38
rs949965564 Chr.19: 50419094-50419094 on Build GRCh38
rs917181780 Chr.19: 50418878-50418878 on Build GRCh38
rs1037178736 Chr.19: 50419081-50419081 on Build GRCh38
rs970390333 Chr.19: 50419171-50419171 on Build GRCh38
rs1019574356 Chr.19: 50419101-50419101 on Build GRCh38
rs990893319 Chr.19: 50419030-50419030 on Build GRCh38
rs1041612458 Chr.19: 50418889-50418889 on Build GRCh38
rs889831898 Chr.19: 50419096-50419096 on Build GRCh38
rs771506184 Chr.19: 50419228-50419228 on Build GRCh38
rs761106907 Chr.19: 50418997-50418997 on Build GRCh38
rs949047702 Chr.19: 50419037-50419037 on Build GRCh38
rs533155148 Chr.19: 50419223-50419223 on Build GRCh38
rs948932390 Chr.19: 50418888-50418888 on Build GRCh38
rs527847312 Chr.19: 50418877-50418877 on Build GRCh38
rs559363950 Chr.19: 50419181-50419181 on Build GRCh38
rs762655908 Chr.19: 50418968-50418968 on Build GRCh38
rs903018514 Chr.19: 50418890-50418890 on Build GRCh38
rs879232857 Chr.19: 50419112-50419113 on Build GRCh38
rs757489574 Chr.19: 50419239-50419240 on Build GRCh38
rs545489456 Chr.19: 50419250-50419250 on Build GRCh38
rs1027729255 Chr.19: 50419272-50419272 on Build GRCh38
rs80162089 Chr.19: 50419293-50419293 on Build GRCh38
rs550672091 Chr.19: 50419285-50419285 on Build GRCh38
rs999414398 Chr.19: 50419312-50419312 on Build GRCh38
rs780991062 Chr.19: 50418940-50418940 on Build GRCh38
rs940238135 Chr.19: 50419211-50419211 on Build GRCh38
rs1009067506 Chr.19: 50418971-50418971 on Build GRCh38
rs994991939 Chr.19: 50419251-50419251 on Build GRCh38
rs916312623 Chr.19: 50419167-50419167 on Build GRCh38
rs928910018 Chr.19: 50419185-50419185 on Build GRCh38
rs1015655239 Chr.19: 50418897-50418897 on Build GRCh38
rs780243736 Chr.19: 50419035-50419035 on Build GRCh38
rs1054767263 Chr.19: 50418870-50418870 on Build GRCh38
rs369962569 Chr.19: 50418925-50418925 on Build GRCh38
rs1019541745 Chr.19: 50418991-50418991 on Build GRCh38
rs902432202 Chr.19: 50419302-50419302 on Build GRCh38
rs758209463 Chr.19: 50418934-50418934 on Build GRCh38
rs749286436 Chr.19: 50418974-50418974 on Build GRCh38
rs547994702 Chr.19: 50419085-50419085 on Build GRCh38
rs916212870 Chr.19: 50419036-50419037 on Build GRCh38
rs551577246 Chr.19: 50418871-50418871 on Build GRCh38
rs549253784 Chr.19: 50418954-50418954 on Build GRCh38
rs777412692 Chr.19: 50419013-50419013 on Build GRCh38
rs1009938588 Chr.19: 50419110-50419110 on Build GRCh38
rs999910331 Chr.19: 50418911-50418911 on Build GRCh38
rs753712086 Chr.19: 50419028-50419028 on Build GRCh38
rs553832216 Chr.19: 50418969-50418969 on Build GRCh38
rs936360987 Chr.19: 50418863-50418864 on Build GRCh38
rs557521292 Chr.19: 50419084-50419084 on Build GRCh38
rs772940778 Chr.19: 50418953-50418953 on Build GRCh38
rs192191813 Chr.19: 50419156-50419156 on Build GRCh38
rs139027495 Chr.19: 50419275-50419275 on Build GRCh38
rs535464804 Chr.19: 50418938-50418938 on Build GRCh38
rs747738249 Chr.19: 50418945-50418945 on Build GRCh38
rs750585778 Chr.19: 50418933-50418933 on Build GRCh38
rs928872589 Chr.19: 50419055-50419055 on Build GRCh38
rs750673739 Chr.19: 50419017-50419017 on Build GRCh38
rs2463237 Chr.19: 50419112-50419112 on Build GRCh38
rs35278617 Chr.19: 50418996-50418996 on Build GRCh38
rs779682541 Chr.19: 50419244-50419244 on Build GRCh38
rs2244095 Chr.19: 50419113-50419113 on Build GRCh38
rs1004335157 Chr.19: 50418893-50418893 on Build GRCh38
rs539566390 Chr.19: 50419054-50419054 on Build GRCh38
rs779781694 Chr.19: 50418936-50418936 on Build GRCh38
rs1008247852 Chr.19: 50419292-50419292 on Build GRCh38
rs543615089 Chr.19: 50419088-50419088 on Build GRCh38
rs959080486 Chr.19: 50419284-50419284 on Build GRCh38
rs751663142 Chr.19: 50418975-50418975 on Build GRCh38
rs757079169 Chr.19: 50419046-50419046 on Build GRCh38
rs761195055 Chr.19: 50418912-50418912 on Build GRCh38
rs549682298 Chr.19: 50419287-50419287 on Build GRCh38
rs768642065 Chr.19: 50418939-50418939 on Build GRCh38
rs767692009 Chr.19: 50418923-50418928 on Build GRCh38
rs745822224 Chr.19: 50419155-50419155 on Build GRCh38

View Full Product Details