Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56811126-56811647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEG3
Entrez Gene ID
5178
Gene Name
paternally expressed 3
Gene Aliases
PW1, ZKSCAN22, ZNF904, ZSCAN24
UniGene
Hs.731875
Gene Chromosome Location
Chr.19: 56810077-56840726 on Build GRCh38
Transcripts
NM_001146184.1 NM_001146185.1 NM_001146186.1 NM_001146187.1 NM_006210.2 NM_001146326.1 NM_001146327.1 NM_015363.4

Gene Symbol
ZIM2
Entrez Gene ID
23619
Gene Name
zinc finger imprinted 2
Gene Aliases
ZNF656
UniGene
Hs.731875
Gene Chromosome Location
Chr.19: 56774547-56840729 on Build GRCh38
Transcripts
NM_001146184.1 NM_001146185.1 NM_001146186.1 NM_001146187.1 NM_006210.2 NM_001146326.1 NM_001146327.1 NM_015363.4

SNP Detail

SNP ID Chromosome Location
rs1041380382 Chr.19: 56811556-56811556 on Build GRCh38
rs1030256740 Chr.19: 56811510-56811511 on Build GRCh38
rs899774102 Chr.19: 56811578-56811578 on Build GRCh38
rs571048482 Chr.19: 56811251-56811251 on Build GRCh38
rs181845486 Chr.19: 56811514-56811514 on Build GRCh38
rs908557950 Chr.19: 56811520-56811520 on Build GRCh38
rs893590878 Chr.19: 56811381-56811381 on Build GRCh38
rs943961653 Chr.19: 56811319-56811319 on Build GRCh38
rs1046654030 Chr.19: 56811346-56811346 on Build GRCh38
rs991700611 Chr.19: 56811433-56811433 on Build GRCh38
rs906491635 Chr.19: 56811364-56811364 on Build GRCh38
rs944015664 Chr.19: 56811544-56811544 on Build GRCh38
rs920045936 Chr.19: 56811212-56811212 on Build GRCh38
rs964595278 Chr.19: 56811457-56811457 on Build GRCh38
rs754198858 Chr.19: 56811339-56811339 on Build GRCh38
rs996467880 Chr.19: 56811492-56811492 on Build GRCh38
rs144123248 Chr.19: 56811259-56811259 on Build GRCh38
rs907611421 Chr.19: 56811542-56811542 on Build GRCh38
rs371223004 Chr.19: 56811250-56811252 on Build GRCh38
rs748632223 Chr.19: 56811277-56811277 on Build GRCh38
rs1155461 Chr.19: 56811206-56811206 on Build GRCh38
rs1035923243 Chr.19: 56811426-56811426 on Build GRCh38
rs1037901776 Chr.19: 56811260-56811262 on Build GRCh38
rs11666110 Chr.19: 56811387-56811387 on Build GRCh38
rs374631594 Chr.19: 56811512-56811514 on Build GRCh38
rs963146405 Chr.19: 56811555-56811555 on Build GRCh38
rs1052472280 Chr.19: 56811390-56811390 on Build GRCh38
rs553752940 Chr.19: 56811620-56811620 on Build GRCh38
rs574029043 Chr.19: 56811510-56811512 on Build GRCh38
rs1032573450 Chr.19: 56811344-56811344 on Build GRCh38
rs931437470 Chr.19: 56811438-56811438 on Build GRCh38
rs765218502 Chr.19: 56811451-56811452 on Build GRCh38
rs1011352891 Chr.19: 56811423-56811423 on Build GRCh38
rs973166072 Chr.19: 56811575-56811575 on Build GRCh38
rs751235428 Chr.19: 56811242-56811242 on Build GRCh38
rs796914535 Chr.19: 56811432-56811432 on Build GRCh38
rs778047889 Chr.19: 56811327-56811327 on Build GRCh38
rs899680417 Chr.19: 56811335-56811335 on Build GRCh38
rs78174116 Chr.19: 56811428-56811428 on Build GRCh38
rs1009316236 Chr.19: 56811384-56811384 on Build GRCh38
rs12980845 Chr.19: 56811481-56811481 on Build GRCh38
rs983149038 Chr.19: 56811529-56811529 on Build GRCh38
rs569805695 Chr.19: 56811334-56811334 on Build GRCh38
rs970345229 Chr.19: 56811388-56811388 on Build GRCh38
rs982791417 Chr.19: 56811482-56811482 on Build GRCh38
rs982111692 Chr.19: 56811415-56811415 on Build GRCh38
rs920082175 Chr.19: 56811434-56811434 on Build GRCh38
rs534107764 Chr.19: 56811609-56811609 on Build GRCh38
rs146733496 Chr.19: 56811311-56811311 on Build GRCh38

View Full Product Details