Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57443945-57444446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF749
Entrez Gene ID
388567
Gene Name
zinc finger protein 749
Gene Aliases
-
UniGene
Hs.467358
Gene Chromosome Location
Chr.19: 57428565-57447097 on Build GRCh38
Transcripts
NM_001023561.3 NM_001321952.1 NM_001321953.1 NM_001321954.1 XM_011526956.2 XM_017026802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397024 c.835C>A p.L279I Chr.19: 57443983-57443983 on Build GRCh38
4756039 c.662T>A p.L221H Chr.19: 57444071-57444071 on Build GRCh38
3540413 c.840A>G p.S280S Chr.19: 57443988-57443988 on Build GRCh38
3540417 c.1117C>T p.Q373* Chr.19: 57444265-57444265 on Build GRCh38
5730704 c.746T>C p.M249T Chr.19: 57444155-57444155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139410369 Chr.19: 57444104-57444104 on Build GRCh38
rs1047489847 Chr.19: 57444040-57444040 on Build GRCh38
rs770213081 Chr.19: 57444060-57444060 on Build GRCh38
rs770583440 Chr.19: 57444045-57444045 on Build GRCh38
rs996500681 Chr.19: 57444126-57444126 on Build GRCh38
rs373764319 Chr.19: 57444092-57444092 on Build GRCh38
rs918889511 Chr.19: 57444304-57444304 on Build GRCh38
rs988870435 Chr.19: 57444174-57444174 on Build GRCh38
rs752509148 Chr.19: 57444217-57444217 on Build GRCh38
rs760997579 Chr.19: 57444302-57444302 on Build GRCh38
rs758799359 Chr.19: 57444356-57444356 on Build GRCh38
rs374973355 Chr.19: 57444328-57444328 on Build GRCh38
rs761488390 Chr.19: 57444068-57444070 on Build GRCh38
rs368423173 Chr.19: 57444169-57444169 on Build GRCh38
rs756487522 Chr.19: 57444162-57444162 on Build GRCh38
rs755704601 Chr.19: 57444254-57444254 on Build GRCh38
rs371817415 Chr.19: 57444399-57444399 on Build GRCh38
rs2240038 Chr.19: 57444361-57444361 on Build GRCh38
rs767101501 Chr.19: 57444420-57444422 on Build GRCh38
rs753114536 Chr.19: 57443978-57443978 on Build GRCh38
rs144605370 Chr.19: 57444332-57444332 on Build GRCh38
rs780836369 Chr.19: 57444158-57444158 on Build GRCh38
rs769210270 Chr.19: 57444264-57444264 on Build GRCh38
rs148951080 Chr.19: 57444019-57444019 on Build GRCh38
rs753636501 Chr.19: 57444117-57444117 on Build GRCh38
rs755270639 Chr.19: 57444348-57444348 on Build GRCh38
rs748737966 Chr.19: 57444352-57444352 on Build GRCh38
rs756717240 Chr.19: 57444238-57444238 on Build GRCh38
rs755832300 Chr.19: 57444336-57444336 on Build GRCh38
rs765172042 Chr.19: 57444211-57444211 on Build GRCh38
rs771272981 Chr.19: 57444305-57444305 on Build GRCh38
rs141529170 Chr.19: 57444066-57444066 on Build GRCh38
rs764307048 Chr.19: 57443988-57443988 on Build GRCh38
rs147745291 Chr.19: 57444202-57444202 on Build GRCh38
rs986312399 Chr.19: 57444185-57444185 on Build GRCh38
rs776923369 Chr.19: 57444048-57444048 on Build GRCh38
rs752631847 Chr.19: 57444134-57444134 on Build GRCh38
rs774396329 Chr.19: 57444082-57444082 on Build GRCh38
rs374326804 Chr.19: 57444074-57444074 on Build GRCh38
rs202176595 Chr.19: 57444300-57444300 on Build GRCh38
rs145614824 Chr.19: 57444196-57444196 on Build GRCh38
rs751125524 Chr.19: 57444237-57444237 on Build GRCh38
rs376445073 Chr.19: 57444038-57444038 on Build GRCh38
rs555328607 Chr.19: 57444363-57444363 on Build GRCh38
rs760120463 Chr.19: 57444400-57444400 on Build GRCh38
rs746707877 Chr.19: 57444042-57444042 on Build GRCh38
rs761941846 Chr.19: 57444181-57444181 on Build GRCh38
rs778445937 Chr.19: 57444186-57444186 on Build GRCh38
rs750046224 Chr.19: 57444249-57444249 on Build GRCh38
rs775626887 Chr.19: 57444394-57444394 on Build GRCh38
rs777465429 Chr.19: 57444140-57444140 on Build GRCh38
rs779337654 Chr.19: 57444265-57444265 on Build GRCh38
rs150869866 Chr.19: 57444079-57444079 on Build GRCh38
rs763046448 Chr.19: 57444231-57444231 on Build GRCh38
rs749846283 Chr.19: 57444176-57444176 on Build GRCh38
rs753637754 Chr.19: 57444013-57444013 on Build GRCh38
rs775479559 Chr.19: 57444319-57444319 on Build GRCh38
rs754216639 Chr.19: 57444344-57444344 on Build GRCh38
rs776923926 Chr.19: 57444375-57444375 on Build GRCh38
rs751107017 Chr.19: 57444141-57444141 on Build GRCh38
rs61997241 Chr.19: 57444256-57444256 on Build GRCh38
rs756913366 Chr.19: 57444144-57444144 on Build GRCh38
rs370732413 Chr.19: 57444321-57444321 on Build GRCh38
rs768525102 Chr.19: 57444385-57444385 on Build GRCh38
rs754745939 Chr.19: 57444121-57444121 on Build GRCh38
rs778898753 Chr.19: 57444026-57444026 on Build GRCh38
rs751745714 Chr.19: 57443994-57443994 on Build GRCh38
rs895981997 Chr.19: 57444000-57444000 on Build GRCh38
rs762063833 Chr.19: 57444087-57444087 on Build GRCh38
rs772140852 Chr.19: 57444293-57444293 on Build GRCh38
rs747664633 Chr.19: 57444031-57444031 on Build GRCh38
rs368025048 Chr.19: 57444388-57444388 on Build GRCh38
rs546700168 Chr.19: 57444206-57444206 on Build GRCh38
rs757940280 Chr.19: 57444035-57444035 on Build GRCh38
rs761113951 Chr.19: 57444210-57444210 on Build GRCh38
rs746228215 Chr.19: 57444050-57444050 on Build GRCh38
rs566824590 Chr.19: 57444255-57444255 on Build GRCh38
rs749123236 Chr.19: 57444377-57444377 on Build GRCh38
rs761255497 Chr.19: 57444329-57444329 on Build GRCh38
rs769122264 Chr.19: 57444178-57444178 on Build GRCh38
rs1054954988 Chr.19: 57444337-57444337 on Build GRCh38
rs2240037 Chr.19: 57444351-57444351 on Build GRCh38
rs766003239 Chr.19: 57444341-57444341 on Build GRCh38
rs780741761 Chr.19: 57444242-57444242 on Build GRCh38
rs1055409247 Chr.19: 57444048-57444052 on Build GRCh38
rs748183100 Chr.19: 57444268-57444268 on Build GRCh38
rs192115663 Chr.19: 57444002-57444002 on Build GRCh38
rs771288971 Chr.19: 57444369-57444369 on Build GRCh38
rs770158353 Chr.19: 57444167-57444167 on Build GRCh38
rs764129578 Chr.19: 57444326-57444326 on Build GRCh38
rs1042287491 Chr.19: 57444383-57444383 on Build GRCh38
rs766137333 Chr.19: 57443998-57443998 on Build GRCh38
rs765117537 Chr.19: 57444125-57444125 on Build GRCh38
rs372485774 Chr.19: 57443980-57443980 on Build GRCh38
rs370949608 Chr.19: 57444041-57444041 on Build GRCh38
rs758388230 Chr.19: 57444138-57444138 on Build GRCh38
rs773997616 Chr.19: 57444387-57444387 on Build GRCh38
rs767356757 Chr.19: 57444396-57444396 on Build GRCh38
rs764105149 Chr.19: 57444233-57444233 on Build GRCh38
rs896389917 Chr.19: 57444055-57444055 on Build GRCh38

View Full Product Details