Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 247460-247964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3YL1
Entrez Gene ID
26751
Gene Name
SH3 and SYLF domain containing 1
Gene Aliases
RAY
UniGene
Hs.515951
Gene Chromosome Location
Chr.2: 218136-264866 on Build GRCh38
Transcripts
NM_001159597.2 NM_001282682.1 NM_001282687.1 NM_015677.3 NR_104223.1 NR_104224.1 NR_104225.1 NR_104226.1 NR_104227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140994 c.231G>T p.W77C Chr.2: 247598-247598 on Build GRCh38
5416229 c.227-5A>T p.? Chr.2: 247607-247607 on Build GRCh38
5465509 c.281T>C p.I94T Chr.2: 247548-247548 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048544417 Chr.2: 247757-247757 on Build GRCh38
rs1030797250 Chr.2: 247850-247850 on Build GRCh38
rs75813741 Chr.2: 247563-247563 on Build GRCh38
rs921468530 Chr.2: 247615-247615 on Build GRCh38
rs542455313 Chr.2: 247842-247842 on Build GRCh38
rs938653525 Chr.2: 247624-247624 on Build GRCh38
rs896455877 Chr.2: 247861-247861 on Build GRCh38
rs541176946 Chr.2: 247729-247729 on Build GRCh38
rs890059781 Chr.2: 247787-247787 on Build GRCh38
rs377373324 Chr.2: 247661-247661 on Build GRCh38
rs905108265 Chr.2: 247514-247514 on Build GRCh38
rs1002101614 Chr.2: 247744-247744 on Build GRCh38
rs1030723741 Chr.2: 247566-247566 on Build GRCh38
rs930420131 Chr.2: 247637-247637 on Build GRCh38
rs900847802 Chr.2: 247835-247835 on Build GRCh38
rs1007163086 Chr.2: 247820-247820 on Build GRCh38
rs186470308 Chr.2: 247635-247635 on Build GRCh38
rs533327033 Chr.2: 247780-247780 on Build GRCh38
rs1009989227 Chr.2: 247862-247862 on Build GRCh38
rs769225120 Chr.2: 247511-247511 on Build GRCh38
rs531521664 Chr.2: 247931-247931 on Build GRCh38
rs766977501 Chr.2: 247520-247520 on Build GRCh38
rs774447403 Chr.2: 247601-247601 on Build GRCh38
rs1021468341 Chr.2: 247577-247577 on Build GRCh38
rs774452617 Chr.2: 247866-247866 on Build GRCh38
rs931517440 Chr.2: 247693-247693 on Build GRCh38
rs375677617 Chr.2: 247521-247521 on Build GRCh38
rs770126945 Chr.2: 247537-247537 on Build GRCh38
rs181819043 Chr.2: 247620-247620 on Build GRCh38
rs560736415 Chr.2: 247912-247912 on Build GRCh38
rs1037083723 Chr.2: 247628-247628 on Build GRCh38
rs770934186 Chr.2: 247593-247593 on Build GRCh38
rs748511295 Chr.2: 247522-247522 on Build GRCh38
rs1020996233 Chr.2: 247874-247874 on Build GRCh38
rs1011183921 Chr.2: 247908-247908 on Build GRCh38
rs1041665244 Chr.2: 247827-247827 on Build GRCh38
rs191784539 Chr.2: 247603-247603 on Build GRCh38
rs1048917351 Chr.2: 247648-247648 on Build GRCh38
rs752230859 Chr.2: 247619-247619 on Build GRCh38
rs763180397 Chr.2: 247590-247590 on Build GRCh38
rs745973000 Chr.2: 247649-247649 on Build GRCh38
rs989325085 Chr.2: 247592-247592 on Build GRCh38
rs150398427 Chr.2: 247699-247699 on Build GRCh38
rs17850312 Chr.2: 247550-247550 on Build GRCh38
rs961014158 Chr.2: 247526-247526 on Build GRCh38
rs979570908 Chr.2: 247611-247611 on Build GRCh38
rs574837027 Chr.2: 247832-247832 on Build GRCh38
rs768597665 Chr.2: 247805-247805 on Build GRCh38
rs957861365 Chr.2: 247581-247581 on Build GRCh38
rs761638535 Chr.2: 247898-247898 on Build GRCh38
rs376311077 Chr.2: 247623-247623 on Build GRCh38
rs969601588 Chr.2: 247936-247936 on Build GRCh38

View Full Product Details