Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26127559-26128065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB10
Entrez Gene ID
10890
Gene Name
RAB10, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.467960
Gene Chromosome Location
Chr.2: 26033860-26137454 on Build GRCh38
Transcripts
NM_016131.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751510 c.490C>A p.L164I Chr.2: 26127922-26127922 on Build GRCh38
6467129 c.486G>T p.A162A Chr.2: 26127918-26127918 on Build GRCh38
3962702 c.433G>A p.G145S Chr.2: 26127865-26127865 on Build GRCh38
3839198 c.486G>A p.A162A Chr.2: 26127918-26127918 on Build GRCh38
1614681 c.469A>G p.I157V Chr.2: 26127901-26127901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011634374 Chr.2: 26127982-26127982 on Build GRCh38
rs369611867 Chr.2: 26127954-26127954 on Build GRCh38
rs577859999 Chr.2: 26127660-26127660 on Build GRCh38
rs538867013 Chr.2: 26127667-26127667 on Build GRCh38
rs865948955 Chr.2: 26128015-26128015 on Build GRCh38
rs1047877885 Chr.2: 26127619-26127619 on Build GRCh38
rs1007441177 Chr.2: 26127736-26127736 on Build GRCh38
rs768813611 Chr.2: 26127799-26127799 on Build GRCh38
rs777136174 Chr.2: 26127926-26127926 on Build GRCh38
rs763363831 Chr.2: 26127826-26127826 on Build GRCh38
rs748354286 Chr.2: 26127972-26127972 on Build GRCh38
rs147862296 Chr.2: 26127685-26127685 on Build GRCh38
rs1023457982 Chr.2: 26128012-26128012 on Build GRCh38
rs779869413 Chr.2: 26127959-26127959 on Build GRCh38
rs960433325 Chr.2: 26128034-26128034 on Build GRCh38
rs1038544574 Chr.2: 26127721-26127721 on Build GRCh38
rs970191192 Chr.2: 26128027-26128027 on Build GRCh38
rs769840237 Chr.2: 26127824-26127824 on Build GRCh38
rs61754189 Chr.2: 26127918-26127918 on Build GRCh38
rs745834317 Chr.2: 26127728-26127729 on Build GRCh38
rs753457021 Chr.2: 26127927-26127927 on Build GRCh38
rs139641380 Chr.2: 26127888-26127888 on Build GRCh38
rs201936806 Chr.2: 26127909-26127909 on Build GRCh38
rs145263164 Chr.2: 26127976-26127976 on Build GRCh38
rs987798805 Chr.2: 26128036-26128036 on Build GRCh38
rs377113866 Chr.2: 26127902-26127902 on Build GRCh38
rs996499508 Chr.2: 26127852-26127852 on Build GRCh38
rs755881426 Chr.2: 26127924-26127924 on Build GRCh38
rs773097477 Chr.2: 26127978-26127978 on Build GRCh38
rs372986064 Chr.2: 26127806-26127806 on Build GRCh38
rs745606073 Chr.2: 26127955-26127955 on Build GRCh38
rs980854616 Chr.2: 26127654-26127654 on Build GRCh38
rs929599360 Chr.2: 26127618-26127618 on Build GRCh38
rs939119501 Chr.2: 26127668-26127668 on Build GRCh38
rs757078844 Chr.2: 26127947-26127947 on Build GRCh38
rs763106399 Chr.2: 26127983-26127983 on Build GRCh38
rs773396994 Chr.2: 26127825-26127825 on Build GRCh38
rs771819117 Chr.2: 26127958-26127958 on Build GRCh38
rs981025663 Chr.2: 26128026-26128026 on Build GRCh38
rs776865713 Chr.2: 26127802-26127802 on Build GRCh38
rs901150438 Chr.2: 26127847-26127847 on Build GRCh38
rs946567483 Chr.2: 26127688-26127688 on Build GRCh38
rs889038079 Chr.2: 26127728-26127728 on Build GRCh38
rs771292406 Chr.2: 26127997-26127997 on Build GRCh38
rs556503872 Chr.2: 26127629-26127629 on Build GRCh38
rs572135425 Chr.2: 26127759-26127759 on Build GRCh38

View Full Product Details