Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26793003-26793549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPA
Entrez Gene ID
1058
Gene Name
centromere protein A
Gene Aliases
CENP-A, CenH3
UniGene
Hs.1594
Gene Chromosome Location
Chr.2: 26786014-26794589 on Build GRCh38
Transcripts
NM_001042426.1 NM_001809.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7118952 c.339C>T p.T113T Chr.2: 26793195-26793195 on Build GRCh38
6626477 c.352C>T p.R118* Chr.2: 26793208-26793208 on Build GRCh38
3972188 c.389G>A p.R130Q Chr.2: 26793245-26793245 on Build GRCh38
7144835 c.392G>T p.R131M Chr.2: 26793248-26793248 on Build GRCh38
6091037 c.398G>T p.R133L Chr.2: 26793254-26793254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973050688 Chr.2: 26793368-26793368 on Build GRCh38
rs753653385 Chr.2: 26793114-26793114 on Build GRCh38
rs913180997 Chr.2: 26793369-26793369 on Build GRCh38
rs779132456 Chr.2: 26793201-26793201 on Build GRCh38
rs749684289 Chr.2: 26793192-26793192 on Build GRCh38
rs11692834 Chr.2: 26793483-26793483 on Build GRCh38
rs35243998 Chr.2: 26793189-26793189 on Build GRCh38
rs1043788994 Chr.2: 26793499-26793499 on Build GRCh38
rs920348069 Chr.2: 26793062-26793062 on Build GRCh38
rs771592458 Chr.2: 26793231-26793231 on Build GRCh38
rs757683161 Chr.2: 26793327-26793327 on Build GRCh38
rs771392905 Chr.2: 26793195-26793195 on Build GRCh38
rs750165360 Chr.2: 26793312-26793312 on Build GRCh38
rs182327504 Chr.2: 26793460-26793460 on Build GRCh38
rs986272101 Chr.2: 26793406-26793406 on Build GRCh38
rs763975420 Chr.2: 26793111-26793111 on Build GRCh38
rs961895145 Chr.2: 26793355-26793355 on Build GRCh38
rs549747003 Chr.2: 26793051-26793051 on Build GRCh38
rs372857657 Chr.2: 26793100-26793100 on Build GRCh38
rs774838491 Chr.2: 26793102-26793102 on Build GRCh38
rs765141033 Chr.2: 26793303-26793303 on Build GRCh38
rs1034439725 Chr.2: 26793509-26793509 on Build GRCh38
rs769706177 Chr.2: 26793323-26793323 on Build GRCh38
rs190445966 Chr.2: 26793430-26793430 on Build GRCh38
rs938496437 Chr.2: 26793332-26793332 on Build GRCh38
rs1057087714 Chr.2: 26793063-26793063 on Build GRCh38
rs953170544 Chr.2: 26793398-26793398 on Build GRCh38
rs369238071 Chr.2: 26793180-26793180 on Build GRCh38
rs570315056 Chr.2: 26793323-26793323 on Build GRCh38
rs938136493 Chr.2: 26793514-26793514 on Build GRCh38
rs1056962907 Chr.2: 26793522-26793522 on Build GRCh38
rs745582099 Chr.2: 26793208-26793208 on Build GRCh38
rs772798880 Chr.2: 26793380-26793380 on Build GRCh38
rs57358285 Chr.2: 26793120-26793120 on Build GRCh38
rs911618746 Chr.2: 26793415-26793415 on Build GRCh38
rs558557805 Chr.2: 26793409-26793409 on Build GRCh38
rs1039446598 Chr.2: 26793404-26793404 on Build GRCh38
rs11687642 Chr.2: 26793391-26793391 on Build GRCh38
rs2289018 Chr.2: 26793101-26793101 on Build GRCh38
rs548629129 Chr.2: 26793273-26793273 on Build GRCh38
rs1009912223 Chr.2: 26793092-26793092 on Build GRCh38
rs372773478 Chr.2: 26793298-26793298 on Build GRCh38
rs761656052 Chr.2: 26793295-26793295 on Build GRCh38
rs1053130868 Chr.2: 26793104-26793104 on Build GRCh38
rs1042753191 Chr.2: 26793397-26793397 on Build GRCh38
rs397748149 Chr.2: 26793436-26793437 on Build GRCh38
rs781011731 Chr.2: 26793126-26793131 on Build GRCh38
rs748101574 Chr.2: 26793132-26793132 on Build GRCh38
rs931697735 Chr.2: 26793085-26793085 on Build GRCh38
rs201617693 Chr.2: 26793177-26793177 on Build GRCh38
rs150615508 Chr.2: 26793324-26793324 on Build GRCh38
rs756983399 Chr.2: 26793116-26793116 on Build GRCh38
rs369888622 Chr.2: 26793105-26793105 on Build GRCh38
rs71441045 Chr.2: 26793504-26793504 on Build GRCh38
rs1005276394 Chr.2: 26793451-26793451 on Build GRCh38
rs1050649683 Chr.2: 26793096-26793096 on Build GRCh38
rs775009264 Chr.2: 26793244-26793244 on Build GRCh38
rs776480635 Chr.2: 26793282-26793282 on Build GRCh38
rs992707750 Chr.2: 26793367-26793367 on Build GRCh38
rs377719985 Chr.2: 26793326-26793326 on Build GRCh38
rs988288472 Chr.2: 26793438-26793438 on Build GRCh38
rs2289017 Chr.2: 26793035-26793035 on Build GRCh38
rs755146520 Chr.2: 26793121-26793121 on Build GRCh38
rs769959915 Chr.2: 26793516-26793516 on Build GRCh38
rs914167068 Chr.2: 26793474-26793474 on Build GRCh38
rs577015377 Chr.2: 26793427-26793427 on Build GRCh38
rs960129387 Chr.2: 26793316-26793316 on Build GRCh38
rs766511602 Chr.2: 26793117-26793117 on Build GRCh38
rs997700814 Chr.2: 26793130-26793133 on Build GRCh38
rs566184611 Chr.2: 26793142-26793142 on Build GRCh38
rs35428352 Chr.2: 26793434-26793435 on Build GRCh38
rs201337992 Chr.2: 26793099-26793099 on Build GRCh38
rs905314868 Chr.2: 26793253-26793253 on Build GRCh38
rs768590880 Chr.2: 26793274-26793274 on Build GRCh38
rs749632833 Chr.2: 26793095-26793095 on Build GRCh38
rs756721845 Chr.2: 26793181-26793181 on Build GRCh38

View Full Product Details