Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27199500-27200010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
UniGene
Hs.435735
Gene Chromosome Location
Chr.2: 27199587-27212307 on Build GRCh38
Transcripts
NM_021095.2 NR_028323.1 XM_006712128.1 XM_006712129.1 XM_006712130.1 XM_011533146.2 XM_017005216.1

SNP Detail

SNP ID Chromosome Location
rs570178151 Chr.2: 27199665-27199665 on Build GRCh38
rs879391993 Chr.2: 27199774-27199774 on Build GRCh38
rs898087047 Chr.2: 27199701-27199701 on Build GRCh38
rs766974679 Chr.2: 27199829-27199829 on Build GRCh38
rs1003339188 Chr.2: 27199796-27199816 on Build GRCh38
rs996912217 Chr.2: 27199728-27199728 on Build GRCh38
rs79373807 Chr.2: 27199786-27199786 on Build GRCh38
rs763727985 Chr.2: 27199626-27199626 on Build GRCh38
rs989977278 Chr.2: 27199896-27199896 on Build GRCh38
rs756735809 Chr.2: 27199753-27199753 on Build GRCh38
rs887384381 Chr.2: 27199678-27199678 on Build GRCh38
rs927709076 Chr.2: 27199931-27199931 on Build GRCh38
rs989795852 Chr.2: 27199822-27199822 on Build GRCh38
rs1054298062 Chr.2: 27199572-27199572 on Build GRCh38
rs7081 Chr.2: 27199737-27199737 on Build GRCh38
rs536460641 Chr.2: 27199832-27199832 on Build GRCh38
rs147636020 Chr.2: 27199642-27199642 on Build GRCh38
rs530891883 Chr.2: 27199685-27199685 on Build GRCh38
rs553288948 Chr.2: 27199913-27199913 on Build GRCh38
rs977420995 Chr.2: 27199830-27199830 on Build GRCh38
rs957307876 Chr.2: 27199865-27199865 on Build GRCh38
rs959041068 Chr.2: 27199738-27199738 on Build GRCh38
rs890823349 Chr.2: 27199664-27199664 on Build GRCh38
rs921007019 Chr.2: 27199589-27199589 on Build GRCh38
rs565913580 Chr.2: 27199748-27199748 on Build GRCh38
rs568295034 Chr.2: 27199978-27199978 on Build GRCh38
rs921120023 Chr.2: 27199926-27199927 on Build GRCh38
rs956657029 Chr.2: 27199807-27199807 on Build GRCh38
rs745866069 Chr.2: 27199618-27199618 on Build GRCh38
rs1006176115 Chr.2: 27199682-27199682 on Build GRCh38
rs538739460 Chr.2: 27199634-27199634 on Build GRCh38
rs900009847 Chr.2: 27199698-27199698 on Build GRCh38
rs935780466 Chr.2: 27199545-27199545 on Build GRCh38
rs971395382 Chr.2: 27199797-27199797 on Build GRCh38
rs932456051 Chr.2: 27199617-27199617 on Build GRCh38
rs906221497 Chr.2: 27199628-27199628 on Build GRCh38
rs980542562 Chr.2: 27199864-27199864 on Build GRCh38
rs867884391 Chr.2: 27199598-27199598 on Build GRCh38
rs529127907 Chr.2: 27199535-27199535 on Build GRCh38
rs1044833601 Chr.2: 27199532-27199532 on Build GRCh38
rs777777925 Chr.2: 27199599-27199599 on Build GRCh38
rs866109268 Chr.2: 27199552-27199552 on Build GRCh38
rs1046729059 Chr.2: 27199942-27199942 on Build GRCh38
rs994932759 Chr.2: 27199733-27199733 on Build GRCh38
rs932378851 Chr.2: 27199940-27199940 on Build GRCh38
rs939058132 Chr.2: 27199933-27199933 on Build GRCh38

View Full Product Details