Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31525198-31525708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRD5A2
Entrez Gene ID
6716
Gene Name
steroid 5 alpha-reductase 2
Gene Aliases
-
UniGene
Hs.458345
Gene Chromosome Location
Chr.2: 31522486-31665651 on Build GRCh38
Transcripts
NM_000348.3 XM_011533069.2 XM_011533072.2

SNP Detail

SNP ID Chromosome Location
rs886055951 Chr.2: 31525582-31525582 on Build GRCh38
rs909096851 Chr.2: 31525669-31525669 on Build GRCh38
rs977334726 Chr.2: 31525608-31525608 on Build GRCh38
rs887707501 Chr.2: 31525223-31525223 on Build GRCh38
rs572080553 Chr.2: 31525437-31525437 on Build GRCh38
rs931957697 Chr.2: 31525585-31525585 on Build GRCh38
rs9332975 Chr.2: 31525347-31525347 on Build GRCh38
rs940594569 Chr.2: 31525673-31525673 on Build GRCh38
rs566378490 Chr.2: 31525222-31525222 on Build GRCh38
rs567160341 Chr.2: 31525613-31525613 on Build GRCh38
rs1022515980 Chr.2: 31525404-31525404 on Build GRCh38
rs183366009 Chr.2: 31525555-31525555 on Build GRCh38
rs886055950 Chr.2: 31525510-31525510 on Build GRCh38
rs968715349 Chr.2: 31525425-31525425 on Build GRCh38
rs180835129 Chr.2: 31525391-31525391 on Build GRCh38
rs958331774 Chr.2: 31525376-31525376 on Build GRCh38
rs75984166 Chr.2: 31525524-31525524 on Build GRCh38
rs901162627 Chr.2: 31525439-31525439 on Build GRCh38
rs976123745 Chr.2: 31525520-31525520 on Build GRCh38
rs61750401 Chr.2: 31525375-31525375 on Build GRCh38
rs543095997 Chr.2: 31525603-31525603 on Build GRCh38
rs186133158 Chr.2: 31525432-31525432 on Build GRCh38
rs1003032029 Chr.2: 31525374-31525374 on Build GRCh38
rs575785628 Chr.2: 31525400-31525400 on Build GRCh38
rs1004677943 Chr.2: 31525230-31525230 on Build GRCh38
rs777069397 Chr.2: 31525508-31525508 on Build GRCh38
rs28383084 Chr.2: 31525533-31525533 on Build GRCh38
rs907069457 Chr.2: 31525350-31525350 on Build GRCh38
rs1049051226 Chr.2: 31525631-31525631 on Build GRCh38
rs578243673 Chr.2: 31525519-31525519 on Build GRCh38
rs1021457185 Chr.2: 31525553-31525553 on Build GRCh38
rs966262235 Chr.2: 31525510-31525510 on Build GRCh38
rs191053938 Chr.2: 31525487-31525487 on Build GRCh38

View Full Product Details