Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32948778-32949278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
UniGene
Hs.619315
Gene Chromosome Location
Chr.2: 32947129-33399509 on Build GRCh38
Transcripts
NM_206943.2 XM_005264317.3 XM_005264318.3 XM_011532853.2 XM_011532855.2 XM_011532856.2 XM_011532857.2 XM_011532858.2 XM_011532859.2 XM_011532860.2 XM_011532861.2 XM_011532862.2 XM_017004108.1 XM_017004109.1 XM_017004110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1214070 c.556T>C p.C186R Chr.2: 32948936-32948936 on Build GRCh38
6010069 c.512G>A p.G171E Chr.2: 32948892-32948892 on Build GRCh38
4583475 c.534T>C p.S178S Chr.2: 32948914-32948914 on Build GRCh38
1407865 c.565C>T p.P189S Chr.2: 32948945-32948945 on Build GRCh38
477352 c.565+2T>C p.? Chr.2: 32948947-32948947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554528264 Chr.2: 32948829-32948829 on Build GRCh38
rs753207754 Chr.2: 32948841-32948841 on Build GRCh38
rs1022140257 Chr.2: 32949017-32949017 on Build GRCh38
rs150654 Chr.2: 32949005-32949005 on Build GRCh38
rs754268048 Chr.2: 32948882-32948882 on Build GRCh38
rs776261971 Chr.2: 32948919-32948919 on Build GRCh38
rs926987957 Chr.2: 32949172-32949172 on Build GRCh38
rs549357995 Chr.2: 32949040-32949040 on Build GRCh38
rs570738596 Chr.2: 32949038-32949038 on Build GRCh38
rs956653624 Chr.2: 32949103-32949103 on Build GRCh38
rs72866295 Chr.2: 32949185-32949185 on Build GRCh38
rs528661711 Chr.2: 32949180-32949180 on Build GRCh38
rs765222409 Chr.2: 32949198-32949198 on Build GRCh38
rs941102267 Chr.2: 32949174-32949174 on Build GRCh38
rs529975050 Chr.2: 32949010-32949010 on Build GRCh38
rs985802991 Chr.2: 32949155-32949155 on Build GRCh38
rs368668567 Chr.2: 32948836-32948836 on Build GRCh38
rs754869743 Chr.2: 32948994-32948994 on Build GRCh38
rs547251529 Chr.2: 32948984-32948984 on Build GRCh38
rs542585090 Chr.2: 32948899-32948899 on Build GRCh38
rs1036514146 Chr.2: 32948961-32948961 on Build GRCh38
rs149392195 Chr.2: 32948946-32948946 on Build GRCh38
rs976409428 Chr.2: 32949104-32949104 on Build GRCh38
rs190499850 Chr.2: 32949235-32949235 on Build GRCh38
rs186792908 Chr.2: 32949196-32949196 on Build GRCh38
rs925795069 Chr.2: 32948828-32948828 on Build GRCh38
rs562390115 Chr.2: 32948904-32948904 on Build GRCh38
rs765896819 Chr.2: 32948967-32948967 on Build GRCh38
rs538297237 Chr.2: 32949152-32949152 on Build GRCh38
rs376301449 Chr.2: 32948914-32948914 on Build GRCh38
rs772707025 Chr.2: 32948831-32948831 on Build GRCh38
rs986304905 Chr.2: 32948834-32948834 on Build GRCh38
rs758508790 Chr.2: 32948989-32948989 on Build GRCh38
rs375693267 Chr.2: 32948832-32948832 on Build GRCh38
rs564688924 Chr.2: 32948980-32948980 on Build GRCh38
rs373808176 Chr.2: 32948978-32948978 on Build GRCh38
rs916912174 Chr.2: 32948884-32948884 on Build GRCh38
rs578224422 Chr.2: 32948797-32948797 on Build GRCh38
rs779332816 Chr.2: 32948894-32948894 on Build GRCh38
rs746946453 Chr.2: 32948908-32948908 on Build GRCh38
rs748043458 Chr.2: 32948928-32948928 on Build GRCh38
rs769996398 Chr.2: 32948988-32948989 on Build GRCh38
rs369323612 Chr.2: 32948986-32948986 on Build GRCh38
rs112352267 Chr.2: 32949091-32949091 on Build GRCh38
rs1047898704 Chr.2: 32949225-32949225 on Build GRCh38
rs764343043 Chr.2: 32948880-32948880 on Build GRCh38
rs13029145 Chr.2: 32948983-32948983 on Build GRCh38
rs756451429 Chr.2: 32948854-32948854 on Build GRCh38
rs917112914 Chr.2: 32949133-32949133 on Build GRCh38
rs750701861 Chr.2: 32948987-32948987 on Build GRCh38
rs1044019413 Chr.2: 32949162-32949162 on Build GRCh38
rs568314442 Chr.2: 32949168-32949168 on Build GRCh38
rs978863175 Chr.2: 32949089-32949089 on Build GRCh38
rs954987907 Chr.2: 32949014-32949014 on Build GRCh38
rs941757008 Chr.2: 32949184-32949184 on Build GRCh38
rs267599357 Chr.2: 32948878-32948878 on Build GRCh38
rs540881867 Chr.2: 32948812-32948812 on Build GRCh38
rs964766838 Chr.2: 32949068-32949068 on Build GRCh38
rs574285274 Chr.2: 32948896-32948896 on Build GRCh38
rs1030225695 Chr.2: 32949106-32949106 on Build GRCh38
rs1053475463 Chr.2: 32948857-32948857 on Build GRCh38
rs919434104 Chr.2: 32949192-32949192 on Build GRCh38
rs988741430 Chr.2: 32949107-32949107 on Build GRCh38
rs1017630806 Chr.2: 32949066-32949066 on Build GRCh38
rs370213558 Chr.2: 32948990-32948990 on Build GRCh38
rs1057509004 Chr.2: 32949242-32949242 on Build GRCh38
rs375096378 Chr.2: 32948956-32948956 on Build GRCh38
rs1000676476 Chr.2: 32949088-32949088 on Build GRCh38
rs776901646 Chr.2: 32949041-32949041 on Build GRCh38
rs772864792 Chr.2: 32948948-32948948 on Build GRCh38
rs531735967 Chr.2: 32949099-32949100 on Build GRCh38
rs769424829 Chr.2: 32948936-32948936 on Build GRCh38
rs757414734 Chr.2: 32948891-32948891 on Build GRCh38
rs75466330 Chr.2: 32948976-32948976 on Build GRCh38

View Full Product Details